Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121988996-121989590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TACC2
Entrez Gene ID
10579
Gene Name
transforming acidic coiled-coil containing protein 2
Gene Aliases
AZU-1, ECTACC
UniGene
Hs.501252
Gene Chromosome Location
Chr.10: 121989174-122254545 on Build GRCh38
Transcripts
NM_001291876.1 NM_001291877.1 NM_206861.2 NM_206862.3

SNP Detail

SNP ID Chromosome Location
rs776072316 Chr.10: 121989360-121989360 on Build GRCh38
rs1042233058 Chr.10: 121989487-121989487 on Build GRCh38
rs548916172 Chr.10: 121989286-121989286 on Build GRCh38
rs987429620 Chr.10: 121989060-121989060 on Build GRCh38
rs1049590222 Chr.10: 121989182-121989182 on Build GRCh38
rs973359600 Chr.10: 121989494-121989494 on Build GRCh38
rs2420980 Chr.10: 121989312-121989312 on Build GRCh38
rs572808375 Chr.10: 121989408-121989408 on Build GRCh38
rs907913264 Chr.10: 121989068-121989068 on Build GRCh38
rs762564570 Chr.10: 121989407-121989407 on Build GRCh38
rs575648701 Chr.10: 121989046-121989046 on Build GRCh38
rs981013029 Chr.10: 121989357-121989357 on Build GRCh38
rs35482691 Chr.10: 121989511-121989512 on Build GRCh38
rs908296000 Chr.10: 121989113-121989113 on Build GRCh38
rs890929596 Chr.10: 121989209-121989209 on Build GRCh38
rs544126169 Chr.10: 121989102-121989102 on Build GRCh38
rs890587664 Chr.10: 121989454-121989454 on Build GRCh38
rs7900585 Chr.10: 121989287-121989287 on Build GRCh38
rs751235609 Chr.10: 121989438-121989438 on Build GRCh38
rs747252814 Chr.10: 121989175-121989175 on Build GRCh38
rs192099598 Chr.10: 121989241-121989241 on Build GRCh38
rs906426419 Chr.10: 121989133-121989133 on Build GRCh38
rs576928358 Chr.10: 121989132-121989132 on Build GRCh38
rs969873075 Chr.10: 121989558-121989558 on Build GRCh38
rs374260725 Chr.10: 121989343-121989343 on Build GRCh38
rs932569076 Chr.10: 121989417-121989417 on Build GRCh38
rs761492335 Chr.10: 121989488-121989488 on Build GRCh38
rs1027697847 Chr.10: 121989556-121989556 on Build GRCh38
rs1009037273 Chr.10: 121989481-121989481 on Build GRCh38
rs771241346 Chr.10: 121989252-121989252 on Build GRCh38
rs749100229 Chr.10: 121989103-121989103 on Build GRCh38
rs908316190 Chr.10: 121989355-121989355 on Build GRCh38
rs961123404 Chr.10: 121989363-121989363 on Build GRCh38
rs948188297 Chr.10: 121989354-121989354 on Build GRCh38
rs187260120 Chr.10: 121989540-121989540 on Build GRCh38
rs899577667 Chr.10: 121989412-121989412 on Build GRCh38
rs368131504 Chr.10: 121989482-121989482 on Build GRCh38
rs1025108934 Chr.10: 121989333-121989333 on Build GRCh38
rs943876036 Chr.10: 121989477-121989477 on Build GRCh38
rs570687354 Chr.10: 121989377-121989377 on Build GRCh38
rs1054200655 Chr.10: 121989521-121989521 on Build GRCh38
rs115821605 Chr.10: 121989167-121989167 on Build GRCh38
rs56264039 Chr.10: 121989266-121989266 on Build GRCh38
rs34491926 Chr.10: 121989535-121989536 on Build GRCh38
rs2901298 Chr.10: 121989326-121989326 on Build GRCh38
rs542911910 Chr.10: 121989100-121989100 on Build GRCh38
rs1033032133 Chr.10: 121989067-121989067 on Build GRCh38
rs1043421301 Chr.10: 121989410-121989411 on Build GRCh38
rs561140197 Chr.10: 121989129-121989129 on Build GRCh38
rs566785052 Chr.10: 121989442-121989442 on Build GRCh38
rs1052311422 Chr.10: 121989378-121989378 on Build GRCh38
rs371604800 Chr.10: 121989208-121989208 on Build GRCh38
rs1000871774 Chr.10: 121989052-121989052 on Build GRCh38
rs1052094273 Chr.10: 121989187-121989187 on Build GRCh38
rs955121621 Chr.10: 121989271-121989271 on Build GRCh38
rs1022030967 Chr.10: 121989224-121989224 on Build GRCh38
rs959637491 Chr.10: 121989095-121989095 on Build GRCh38
rs1008056453 Chr.10: 121989239-121989239 on Build GRCh38
rs912481480 Chr.10: 121989469-121989469 on Build GRCh38
rs1010580459 Chr.10: 121989223-121989223 on Build GRCh38
rs973387312 Chr.10: 121989329-121989329 on Build GRCh38
rs970907229 Chr.10: 121989335-121989335 on Build GRCh38
rs553270419 Chr.10: 121989429-121989429 on Build GRCh38
rs375129852 Chr.10: 121989155-121989155 on Build GRCh38
rs1032317071 Chr.10: 121989311-121989311 on Build GRCh38
rs927855191 Chr.10: 121989375-121989376 on Build GRCh38
rs748235790 Chr.10: 121989101-121989101 on Build GRCh38
rs936747602 Chr.10: 121989519-121989519 on Build GRCh38

View Full Product Details