Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6457278-6457872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAPBPL
Entrez Gene ID
55080
Gene Name
TAP binding protein like
Gene Aliases
TAPBP-R, TAPBPR
UniGene
Hs.504597
Gene Chromosome Location
Chr.12: 6451655-6472006 on Build GRCh38
Transcripts
NM_018009.4 XM_005253700.3 XM_017019545.1 XM_017019546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5967784 c.789C>T p.A263A Chr.12: 6457629-6457629 on Build GRCh38
942564 c.683G>A p.R228Q Chr.12: 6457523-6457523 on Build GRCh38
5927691 c.800C>T p.S267F Chr.12: 6457640-6457640 on Build GRCh38
3464260 c.741G>A p.Q247Q Chr.12: 6457581-6457581 on Build GRCh38
6000529 c.566-7T>G p.? Chr.12: 6457399-6457399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80121838 Chr.12: 6457577-6457577 on Build GRCh38
rs759024231 Chr.12: 6457412-6457412 on Build GRCh38
rs753870144 Chr.12: 6457755-6457755 on Build GRCh38
rs758207098 Chr.12: 6457566-6457566 on Build GRCh38
rs780033985 Chr.12: 6457680-6457680 on Build GRCh38
rs972158285 Chr.12: 6457346-6457346 on Build GRCh38
rs771155357 Chr.12: 6457403-6457403 on Build GRCh38
rs145002394 Chr.12: 6457569-6457569 on Build GRCh38
rs908469827 Chr.12: 6457671-6457671 on Build GRCh38
rs759912011 Chr.12: 6457729-6457729 on Build GRCh38
rs778616024 Chr.12: 6457589-6457589 on Build GRCh38
rs368452478 Chr.12: 6457745-6457745 on Build GRCh38
rs943795453 Chr.12: 6457317-6457317 on Build GRCh38
rs776081237 Chr.12: 6457736-6457736 on Build GRCh38
rs887143158 Chr.12: 6457463-6457463 on Build GRCh38
rs752309726 Chr.12: 6457370-6457370 on Build GRCh38
rs757060906 Chr.12: 6457588-6457588 on Build GRCh38
rs370569616 Chr.12: 6457472-6457472 on Build GRCh38
rs756208076 Chr.12: 6457630-6457630 on Build GRCh38
rs777448228 Chr.12: 6457492-6457492 on Build GRCh38
rs148676522 Chr.12: 6457572-6457572 on Build GRCh38
rs1055643412 Chr.12: 6457347-6457347 on Build GRCh38
rs1041068917 Chr.12: 6457508-6457508 on Build GRCh38
rs369139309 Chr.12: 6457375-6457375 on Build GRCh38
rs780781991 Chr.12: 6457711-6457711 on Build GRCh38
rs781274274 Chr.12: 6457468-6457468 on Build GRCh38
rs888910391 Chr.12: 6457467-6457467 on Build GRCh38
rs918958027 Chr.12: 6457682-6457682 on Build GRCh38
rs1047045113 Chr.12: 6457462-6457462 on Build GRCh38
rs774954384 Chr.12: 6457439-6457440 on Build GRCh38
rs899944483 Chr.12: 6457517-6457517 on Build GRCh38
rs528810200 Chr.12: 6457518-6457518 on Build GRCh38
rs765025420 Chr.12: 6457309-6457309 on Build GRCh38
rs1051543915 Chr.12: 6457427-6457427 on Build GRCh38
rs756316246 Chr.12: 6457475-6457475 on Build GRCh38
rs773749731 Chr.12: 6457526-6457526 on Build GRCh38
rs762074490 Chr.12: 6457757-6457757 on Build GRCh38
rs536533776 Chr.12: 6457587-6457587 on Build GRCh38
rs763782394 Chr.12: 6457523-6457523 on Build GRCh38
rs750255175 Chr.12: 6457759-6457759 on Build GRCh38
rs146293321 Chr.12: 6457407-6457407 on Build GRCh38
rs367967500 Chr.12: 6457784-6457784 on Build GRCh38
rs202227888 Chr.12: 6457654-6457654 on Build GRCh38
rs771997905 Chr.12: 6457597-6457597 on Build GRCh38
rs367731473 Chr.12: 6457629-6457629 on Build GRCh38
rs139424342 Chr.12: 6457420-6457420 on Build GRCh38
rs1032536367 Chr.12: 6457545-6457545 on Build GRCh38
rs899897946 Chr.12: 6457707-6457707 on Build GRCh38
rs774914021 Chr.12: 6457607-6457607 on Build GRCh38
rs768225454 Chr.12: 6457708-6457708 on Build GRCh38
rs765552248 Chr.12: 6457396-6457396 on Build GRCh38
rs374333373 Chr.12: 6457622-6457622 on Build GRCh38
rs571120714 Chr.12: 6457450-6457450 on Build GRCh38
rs757434582 Chr.12: 6457647-6457647 on Build GRCh38
rs981156322 Chr.12: 6457302-6457302 on Build GRCh38
rs981125090 Chr.12: 6457592-6457592 on Build GRCh38
rs758196754 Chr.12: 6457764-6457764 on Build GRCh38
rs541269313 Chr.12: 6457522-6457522 on Build GRCh38
rs763864841 Chr.12: 6457613-6457613 on Build GRCh38
rs769518478 Chr.12: 6457720-6457720 on Build GRCh38
rs767049553 Chr.12: 6457549-6457549 on Build GRCh38
rs557638949 Chr.12: 6457814-6457814 on Build GRCh38
rs537438146 Chr.12: 6457722-6457722 on Build GRCh38
rs185881959 Chr.12: 6457596-6457596 on Build GRCh38
rs374216855 Chr.12: 6457672-6457672 on Build GRCh38
rs763404003 Chr.12: 6457542-6457542 on Build GRCh38
rs747829683 Chr.12: 6457712-6457712 on Build GRCh38
rs368603949 Chr.12: 6457669-6457669 on Build GRCh38
rs746499832 Chr.12: 6457610-6457610 on Build GRCh38
rs759142074 Chr.12: 6457357-6457357 on Build GRCh38
rs997411915 Chr.12: 6457530-6457530 on Build GRCh38
rs1009554705 Chr.12: 6457589-6457589 on Build GRCh38
rs894047719 Chr.12: 6457548-6457548 on Build GRCh38
rs761499216 Chr.12: 6457620-6457620 on Build GRCh38
rs770923977 Chr.12: 6457494-6457494 on Build GRCh38
rs372660549 Chr.12: 6457653-6457653 on Build GRCh38
rs1009856500 Chr.12: 6457563-6457563 on Build GRCh38
rs149945987 Chr.12: 6457493-6457493 on Build GRCh38
rs1010154404 Chr.12: 6457800-6457800 on Build GRCh38
rs765417644 Chr.12: 6457758-6457758 on Build GRCh38
rs200270358 Chr.12: 6457743-6457743 on Build GRCh38
rs751515199 Chr.12: 6457782-6457782 on Build GRCh38
rs141207979 Chr.12: 6457623-6457623 on Build GRCh38
rs941674798 Chr.12: 6457310-6457310 on Build GRCh38
rs146936905 Chr.12: 6457646-6457646 on Build GRCh38
rs771490726 Chr.12: 6457510-6457510 on Build GRCh38
rs774261669 Chr.12: 6457496-6457496 on Build GRCh38
rs141867731 Chr.12: 6457733-6457733 on Build GRCh38
rs772547574 Chr.12: 6457660-6457664 on Build GRCh38
rs910484344 Chr.12: 6457786-6457786 on Build GRCh38
rs780673708 Chr.12: 6457417-6457417 on Build GRCh38
rs1041035597 Chr.12: 6457681-6457681 on Build GRCh38
rs931213359 Chr.12: 6457624-6457624 on Build GRCh38
rs148937081 Chr.12: 6457616-6457616 on Build GRCh38
rs906798479 Chr.12: 6457823-6457823 on Build GRCh38
rs779652370 Chr.12: 6457777-6457777 on Build GRCh38
rs776179311 Chr.12: 6457722-6457722 on Build GRCh38
rs373833866 Chr.12: 6457692-6457692 on Build GRCh38
rs771705782 Chr.12: 6457373-6457373 on Build GRCh38
rs932791825 Chr.12: 6457739-6457739 on Build GRCh38
rs928373541 Chr.12: 6457323-6457323 on Build GRCh38
rs755094176 Chr.12: 6457440-6457440 on Build GRCh38
rs746402950 Chr.12: 6457655-6457656 on Build GRCh38
rs1021196429 Chr.12: 6457801-6457801 on Build GRCh38
rs188801141 Chr.12: 6457342-6457342 on Build GRCh38
rs745751036 Chr.12: 6457504-6457504 on Build GRCh38
rs752125188 Chr.12: 6457554-6457554 on Build GRCh38
rs752770027 Chr.12: 6457570-6457570 on Build GRCh38
rs764106054 Chr.12: 6457645-6457645 on Build GRCh38

View Full Product Details