Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34103032-34103595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGBD4
Entrez Gene ID
161779
Gene Name
piggyBac transposable element derived 4
Gene Aliases
-
UniGene
Hs.156317
Gene Chromosome Location
Chr.15: 34102073-34104390 on Build GRCh38
Transcripts
NM_152595.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
960787 c.854C>A p.S285Y Chr.15: 34103385-34103385 on Build GRCh38
960788 c.866G>A p.W289* Chr.15: 34103397-34103397 on Build GRCh38
6001711 c.896T>G p.M299R Chr.15: 34103427-34103427 on Build GRCh38
960786 c.734G>C p.R245T Chr.15: 34103265-34103265 on Build GRCh38
4611521 c.545_546delTT p.L183fs*12 Chr.15: 34103076-34103077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778306367 Chr.15: 34103463-34103463 on Build GRCh38
rs770341607 Chr.15: 34103491-34103491 on Build GRCh38
rs768428971 Chr.15: 34103370-34103370 on Build GRCh38
rs755332934 Chr.15: 34103332-34103332 on Build GRCh38
rs1053909363 Chr.15: 34103302-34103302 on Build GRCh38
rs1052564183 Chr.15: 34103192-34103192 on Build GRCh38
rs202024701 Chr.15: 34103172-34103172 on Build GRCh38
rs1044827368 Chr.15: 34103331-34103331 on Build GRCh38
rs146189712 Chr.15: 34103522-34103522 on Build GRCh38
rs199581079 Chr.15: 34103321-34103321 on Build GRCh38
rs768486530 Chr.15: 34103506-34103506 on Build GRCh38
rs370825077 Chr.15: 34103544-34103544 on Build GRCh38
rs202141977 Chr.15: 34103055-34103055 on Build GRCh38
rs762579307 Chr.15: 34103458-34103460 on Build GRCh38
rs779740799 Chr.15: 34103482-34103482 on Build GRCh38
rs148860727 Chr.15: 34103369-34103369 on Build GRCh38
rs776345687 Chr.15: 34103140-34103140 on Build GRCh38
rs760133722 Chr.15: 34103512-34103514 on Build GRCh38
rs75481258 Chr.15: 34103078-34103078 on Build GRCh38
rs34833601 Chr.15: 34103434-34103435 on Build GRCh38
rs780257243 Chr.15: 34103113-34103113 on Build GRCh38
rs775289821 Chr.15: 34103253-34103253 on Build GRCh38
rs142561291 Chr.15: 34103381-34103381 on Build GRCh38
rs776494909 Chr.15: 34103271-34103271 on Build GRCh38
rs773140925 Chr.15: 34103543-34103543 on Build GRCh38
rs146381896 Chr.15: 34103227-34103227 on Build GRCh38
rs781778443 Chr.15: 34103339-34103339 on Build GRCh38
rs750598314 Chr.15: 34103091-34103091 on Build GRCh38
rs946418983 Chr.15: 34103306-34103306 on Build GRCh38
rs773195843 Chr.15: 34103404-34103404 on Build GRCh38
rs753249085 Chr.15: 34103342-34103342 on Build GRCh38
rs746661880 Chr.15: 34103487-34103487 on Build GRCh38
rs760041549 Chr.15: 34103437-34103437 on Build GRCh38
rs769615144 Chr.15: 34103185-34103185 on Build GRCh38
rs779615523 Chr.15: 34103368-34103368 on Build GRCh38
rs763106066 Chr.15: 34103300-34103300 on Build GRCh38
rs747327538 Chr.15: 34103121-34103121 on Build GRCh38
rs775301641 Chr.15: 34103379-34103379 on Build GRCh38
rs767965674 Chr.15: 34103445-34103445 on Build GRCh38
rs756605245 Chr.15: 34103446-34103446 on Build GRCh38
rs764024087 Chr.15: 34103264-34103264 on Build GRCh38
rs935022293 Chr.15: 34103255-34103255 on Build GRCh38
rs768088366 Chr.15: 34103514-34103515 on Build GRCh38
rs147563138 Chr.15: 34103425-34103425 on Build GRCh38
rs765199469 Chr.15: 34103287-34103287 on Build GRCh38
rs536136648 Chr.15: 34103188-34103188 on Build GRCh38
rs766582033 Chr.15: 34103305-34103305 on Build GRCh38
rs905331328 Chr.15: 34103366-34103366 on Build GRCh38
rs751973731 Chr.15: 34103308-34103308 on Build GRCh38
rs759083171 Chr.15: 34103161-34103161 on Build GRCh38
rs560002445 Chr.15: 34103082-34103082 on Build GRCh38
rs527624224 Chr.15: 34103171-34103171 on Build GRCh38
rs141918057 Chr.15: 34103466-34103466 on Build GRCh38
rs377322220 Chr.15: 34103259-34103259 on Build GRCh38
rs201890653 Chr.15: 34103148-34103148 on Build GRCh38
rs879042878 Chr.15: 34103570-34103570 on Build GRCh38
rs765510999 Chr.15: 34103089-34103089 on Build GRCh38
rs897113333 Chr.15: 34103471-34103471 on Build GRCh38
rs771127773 Chr.15: 34103571-34103571 on Build GRCh38
rs751815133 Chr.15: 34103426-34103426 on Build GRCh38
rs771955148 Chr.15: 34103230-34103230 on Build GRCh38
rs377101769 Chr.15: 34103352-34103352 on Build GRCh38
rs374332052 Chr.15: 34103574-34103574 on Build GRCh38
rs769523483 Chr.15: 34103176-34103179 on Build GRCh38
rs757261104 Chr.15: 34103084-34103084 on Build GRCh38
rs547379354 Chr.15: 34103165-34103165 on Build GRCh38
rs774123573 Chr.15: 34103193-34103193 on Build GRCh38
rs377184439 Chr.15: 34103435-34103435 on Build GRCh38
rs150028814 Chr.15: 34103277-34103277 on Build GRCh38
rs778266868 Chr.15: 34103360-34103360 on Build GRCh38
rs758835595 Chr.15: 34103094-34103094 on Build GRCh38
rs145331237 Chr.15: 34103362-34103362 on Build GRCh38
rs757867616 Chr.15: 34103470-34103470 on Build GRCh38
rs761631964 Chr.15: 34103384-34103384 on Build GRCh38
rs769720800 Chr.15: 34103403-34103403 on Build GRCh38
rs760422907 Chr.15: 34103256-34103256 on Build GRCh38
rs780985754 Chr.15: 34103521-34103521 on Build GRCh38
rs370574731 Chr.15: 34103408-34103408 on Build GRCh38
rs773931267 Chr.15: 34103504-34103504 on Build GRCh38
rs772714037 Chr.15: 34103295-34103301 on Build GRCh38
rs750705018 Chr.15: 34103295-34103295 on Build GRCh38
rs771657889 Chr.15: 34103365-34103365 on Build GRCh38

View Full Product Details