Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58987487-58987855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF111
Entrez Gene ID
54778
Gene Name
ring finger protein 111
Gene Aliases
ARK
UniGene
Hs.404423
Gene Chromosome Location
Chr.15: 58987588-59097419 on Build GRCh38
Transcripts
NM_001270528.1 NM_001270529.1 NM_017610.7 XM_005254479.3 XM_006720575.3 XM_017022336.1 XM_017022337.1 XM_017022338.1 XM_017022339.1 XM_017022340.1

SNP Detail

SNP ID Chromosome Location
rs553057575 Chr.15: 58987687-58987687 on Build GRCh38
rs973120905 Chr.15: 58987511-58987511 on Build GRCh38
rs776656500 Chr.15: 58987698-58987698 on Build GRCh38
rs931798858 Chr.15: 58987534-58987534 on Build GRCh38
rs112968854 Chr.15: 58987672-58987672 on Build GRCh38
rs776536752 Chr.15: 58987699-58987700 on Build GRCh38
rs188755309 Chr.15: 58987582-58987582 on Build GRCh38
rs946679412 Chr.15: 58987725-58987725 on Build GRCh38
rs529815971 Chr.15: 58987678-58987678 on Build GRCh38
rs1052826238 Chr.15: 58987573-58987573 on Build GRCh38
rs890342829 Chr.15: 58987570-58987570 on Build GRCh38
rs1017175621 Chr.15: 58987660-58987660 on Build GRCh38
rs570717167 Chr.15: 58987754-58987754 on Build GRCh38
rs1004096351 Chr.15: 58987618-58987618 on Build GRCh38
rs539056473 Chr.15: 58987626-58987626 on Build GRCh38
rs973067035 Chr.15: 58987668-58987668 on Build GRCh38
rs200091286 Chr.15: 58987717-58987718 on Build GRCh38
rs1010943689 Chr.15: 58987594-58987594 on Build GRCh38
rs192188341 Chr.15: 58987784-58987784 on Build GRCh38
rs921713995 Chr.15: 58987533-58987533 on Build GRCh38
rs983052803 Chr.15: 58987659-58987659 on Build GRCh38
rs915187974 Chr.15: 58987705-58987705 on Build GRCh38
rs116750621 Chr.15: 58987549-58987549 on Build GRCh38
rs935271842 Chr.15: 58987572-58987572 on Build GRCh38
rs558119560 Chr.15: 58987587-58987587 on Build GRCh38
rs771871144 Chr.15: 58987671-58987671 on Build GRCh38
rs542029990 Chr.15: 58987680-58987680 on Build GRCh38
rs762047849 Chr.15: 58987701-58987717 on Build GRCh38
rs905301722 Chr.15: 58987613-58987613 on Build GRCh38
rs543925060 Chr.15: 58987627-58987627 on Build GRCh38
rs572826364 Chr.15: 58987775-58987775 on Build GRCh38
rs963001684 Chr.15: 58987665-58987665 on Build GRCh38
rs376025450 Chr.15: 58987686-58987686 on Build GRCh38
rs1023710635 Chr.15: 58987596-58987596 on Build GRCh38
rs572380679 Chr.15: 58987528-58987528 on Build GRCh38
rs550343012 Chr.15: 58987770-58987770 on Build GRCh38
rs760648212 Chr.15: 58987676-58987676 on Build GRCh38
rs552546904 Chr.15: 58987745-58987745 on Build GRCh38
rs768382770 Chr.15: 58987540-58987540 on Build GRCh38
rs765246636 Chr.15: 58987700-58987700 on Build GRCh38

View Full Product Details