Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78984998-78985359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGRF1
Entrez Gene ID
5923
Gene Name
Ras protein specific guanine nucleotide releasing factor 1
Gene Aliases
CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
UniGene
Hs.459035
Gene Chromosome Location
Chr.15: 78959947-79090873 on Build GRCh38
Transcripts
NM_001145648.1 NM_002891.4 NM_153815.2 XM_011521866.2 XM_011521867.2 XM_017022455.1 XM_017022456.1 XM_017022457.1 XM_017022458.1 XM_017022459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6501391 c.3301G>A p.E1101K Chr.15: 78985168-78985168 on Build GRCh38
1293544 c.3321G>A p.V1107V Chr.15: 78985148-78985148 on Build GRCh38
701704 c.3304G>T p.D1102Y Chr.15: 78985165-78985165 on Build GRCh38
6481069 c.3303G>T p.E1101D Chr.15: 78985166-78985166 on Build GRCh38
5680007 c.3313G>A p.A1105T Chr.15: 78985156-78985156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183855437 Chr.15: 78985324-78985324 on Build GRCh38
rs763095425 Chr.15: 78985041-78985041 on Build GRCh38
rs770093406 Chr.15: 78985031-78985031 on Build GRCh38
rs762403523 Chr.15: 78985192-78985192 on Build GRCh38
rs200705194 Chr.15: 78985217-78985217 on Build GRCh38
rs772432320 Chr.15: 78985213-78985213 on Build GRCh38
rs908065950 Chr.15: 78985292-78985292 on Build GRCh38
rs745899859 Chr.15: 78985271-78985271 on Build GRCh38
rs534779427 Chr.15: 78985268-78985268 on Build GRCh38
rs2230518 Chr.15: 78985169-78985169 on Build GRCh38
rs372576578 Chr.15: 78985123-78985123 on Build GRCh38
rs752361913 Chr.15: 78985238-78985238 on Build GRCh38
rs774934199 Chr.15: 78985063-78985063 on Build GRCh38
rs764836166 Chr.15: 78985115-78985115 on Build GRCh38
rs4778751 Chr.15: 78985266-78985266 on Build GRCh38
rs368264976 Chr.15: 78985293-78985293 on Build GRCh38
rs980068014 Chr.15: 78985257-78985257 on Build GRCh38
rs148440502 Chr.15: 78985145-78985145 on Build GRCh38
rs552256734 Chr.15: 78985239-78985239 on Build GRCh38
rs774255043 Chr.15: 78985174-78985174 on Build GRCh38
rs889428370 Chr.15: 78985019-78985019 on Build GRCh38
rs551294752 Chr.15: 78985147-78985147 on Build GRCh38
rs368529149 Chr.15: 78985118-78985118 on Build GRCh38
rs939749278 Chr.15: 78985313-78985313 on Build GRCh38
rs767955928 Chr.15: 78985073-78985073 on Build GRCh38
rs891594333 Chr.15: 78985304-78985304 on Build GRCh38
rs930771848 Chr.15: 78985277-78985277 on Build GRCh38
rs775761540 Chr.15: 78985032-78985032 on Build GRCh38
rs753120852 Chr.15: 78985028-78985028 on Build GRCh38
rs117481060 Chr.15: 78985049-78985049 on Build GRCh38
rs1028667615 Chr.15: 78985079-78985079 on Build GRCh38
rs763694663 Chr.15: 78985253-78985253 on Build GRCh38
rs542075208 Chr.15: 78985139-78985139 on Build GRCh38
rs749462256 Chr.15: 78985173-78985173 on Build GRCh38
rs983608489 Chr.15: 78985288-78985288 on Build GRCh38
rs889471831 Chr.15: 78985335-78985335 on Build GRCh38
rs528622544 Chr.15: 78985124-78985124 on Build GRCh38
rs376602426 Chr.15: 78985157-78985157 on Build GRCh38
rs764924189 Chr.15: 78985234-78985234 on Build GRCh38
rs939783290 Chr.15: 78985026-78985026 on Build GRCh38
rs368452140 Chr.15: 78985085-78985085 on Build GRCh38
rs569376580 Chr.15: 78985214-78985214 on Build GRCh38
rs868393463 Chr.15: 78985138-78985138 on Build GRCh38
rs116883402 Chr.15: 78985144-78985144 on Build GRCh38
rs779570128 Chr.15: 78985269-78985269 on Build GRCh38
rs754670907 Chr.15: 78985107-78985107 on Build GRCh38
rs143238097 Chr.15: 78985067-78985067 on Build GRCh38

View Full Product Details