Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 110636555-110637120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PITX2
Entrez Gene ID
5308
Gene Name
paired like homeodomain 2
Gene Aliases
ARP1, Brx1, IDG2, IGDS, IGDS2, IHG2, IRID2, Otlx2, PTX2, RGS, RIEG, RIEG1, RS
UniGene
Hs.643588
Gene Chromosome Location
Chr.4: 110617423-110642123 on Build GRCh38
Transcripts
NM_001204397.1 NM_001204398.1 NM_001204399.1 NM_153426.2 NM_153427.2 XM_011532027.2

SNP Detail

SNP ID Chromosome Location
rs548434234 Chr.4: 110636577-110636577 on Build GRCh38
rs776489864 Chr.4: 110637082-110637082 on Build GRCh38
rs999225014 Chr.4: 110636890-110636890 on Build GRCh38
rs1034672162 Chr.4: 110636991-110636991 on Build GRCh38
rs1010571247 Chr.4: 110636963-110636963 on Build GRCh38
rs942783447 Chr.4: 110637071-110637071 on Build GRCh38
rs892666724 Chr.4: 110636944-110636944 on Build GRCh38
rs1052607338 Chr.4: 110636942-110636942 on Build GRCh38
rs972039726 Chr.4: 110637007-110637007 on Build GRCh38
rs113717851 Chr.4: 110637057-110637057 on Build GRCh38
rs904839822 Chr.4: 110636977-110636977 on Build GRCh38
rs1026147392 Chr.4: 110637054-110637054 on Build GRCh38
rs901076483 Chr.4: 110636879-110636879 on Build GRCh38
rs952174424 Chr.4: 110637024-110637024 on Build GRCh38
rs980621377 Chr.4: 110636692-110636692 on Build GRCh38
rs1022006991 Chr.4: 110636964-110636964 on Build GRCh38
rs982747899 Chr.4: 110636725-110636725 on Build GRCh38
rs149097993 Chr.4: 110637064-110637064 on Build GRCh38
rs367939004 Chr.4: 110636758-110636758 on Build GRCh38
rs930071307 Chr.4: 110636770-110636770 on Build GRCh38
rs889586740 Chr.4: 110636854-110636854 on Build GRCh38
rs953622075 Chr.4: 110636615-110636615 on Build GRCh38
rs1031608496 Chr.4: 110636627-110636627 on Build GRCh38
rs535573526 Chr.4: 110636975-110636975 on Build GRCh38
rs919114266 Chr.4: 110636750-110636750 on Build GRCh38
rs570712814 Chr.4: 110636799-110636799 on Build GRCh38
rs969028859 Chr.4: 110636688-110636688 on Build GRCh38
rs901311950 Chr.4: 110636617-110636617 on Build GRCh38
rs971904388 Chr.4: 110636744-110636744 on Build GRCh38
rs922551206 Chr.4: 110637065-110637065 on Build GRCh38
rs893565979 Chr.4: 110636901-110636901 on Build GRCh38
rs574619224 Chr.4: 110636909-110636909 on Build GRCh38
rs1003704385 Chr.4: 110636952-110636952 on Build GRCh38
rs111733107 Chr.4: 110636981-110636981 on Build GRCh38
rs1041005863 Chr.4: 110636869-110636869 on Build GRCh38
rs774880258 Chr.4: 110637053-110637054 on Build GRCh38
rs942651729 Chr.4: 110636860-110636860 on Build GRCh38
rs1011106793 Chr.4: 110636679-110636679 on Build GRCh38
rs1023824651 Chr.4: 110636918-110636918 on Build GRCh38
rs1019300985 Chr.4: 110636601-110636601 on Build GRCh38
rs969680926 Chr.4: 110636721-110636721 on Build GRCh38
rs566046542 Chr.4: 110636691-110636691 on Build GRCh38
rs2739203 Chr.4: 110636790-110636790 on Build GRCh38
rs909984465 Chr.4: 110637069-110637069 on Build GRCh38
rs920560326 Chr.4: 110637017-110637017 on Build GRCh38
rs767726449 Chr.4: 110636680-110636680 on Build GRCh38
rs957764914 Chr.4: 110636686-110636686 on Build GRCh38
rs984247043 Chr.4: 110636781-110636781 on Build GRCh38
rs569889228 Chr.4: 110636726-110636726 on Build GRCh38
rs998381102 Chr.4: 110636622-110636622 on Build GRCh38
rs909847452 Chr.4: 110636788-110636788 on Build GRCh38
rs552456920 Chr.4: 110636766-110636766 on Build GRCh38
rs910618670 Chr.4: 110637043-110637044 on Build GRCh38
rs956467443 Chr.4: 110636655-110636655 on Build GRCh38
rs1039983974 Chr.4: 110636609-110636609 on Build GRCh38
rs553173018 Chr.4: 110636892-110636892 on Build GRCh38
rs371965728 Chr.4: 110636961-110636961 on Build GRCh38
rs901404159 Chr.4: 110636903-110636903 on Build GRCh38
rs972032996 Chr.4: 110636755-110636755 on Build GRCh38
rs189715917 Chr.4: 110636739-110636739 on Build GRCh38
rs374074728 Chr.4: 110636621-110636621 on Build GRCh38
rs900815169 Chr.4: 110636608-110636608 on Build GRCh38
rs972427230 Chr.4: 110637028-110637028 on Build GRCh38
rs986595215 Chr.4: 110637036-110637036 on Build GRCh38
rs960643451 Chr.4: 110637021-110637021 on Build GRCh38
rs1034914666 Chr.4: 110636700-110636700 on Build GRCh38
rs930681180 Chr.4: 110636820-110636820 on Build GRCh38
rs535056127 Chr.4: 110636849-110636849 on Build GRCh38
rs942688607 Chr.4: 110636810-110636810 on Build GRCh38
rs1024190501 Chr.4: 110636697-110636697 on Build GRCh38
rs935202329 Chr.4: 110637075-110637075 on Build GRCh38

View Full Product Details