Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59654292-59654808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A9
Entrez Gene ID
220963
Gene Name
solute carrier family 16 member 9
Gene Aliases
C10orf36, MCT9
UniGene
Hs.499709
Gene Chromosome Location
Chr.10: 59650764-59710079 on Build GRCh38
Transcripts
NM_001323977.1 NM_001323978.1 NM_001323979.1 NM_001323980.1 NM_001323981.1 NM_194298.2 XM_017015883.1 XM_017015884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1348400 c.604delA p.I202fs*1 Chr.10: 59654422-59654422 on Build GRCh38
6779871 c.657G>T p.K219N Chr.10: 59654369-59654369 on Build GRCh38
4396940 c.571C>T p.L191F Chr.10: 59654455-59654455 on Build GRCh38
1348401 c.564G>A p.M188I Chr.10: 59654462-59654462 on Build GRCh38
684811 c.541A>G p.I181V Chr.10: 59654485-59654485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768442955 Chr.10: 59654521-59654521 on Build GRCh38
rs775979449 Chr.10: 59654408-59654408 on Build GRCh38
rs548227657 Chr.10: 59654422-59654422 on Build GRCh38
rs900860365 Chr.10: 59654553-59654553 on Build GRCh38
rs1006940777 Chr.10: 59654344-59654344 on Build GRCh38
rs777872049 Chr.10: 59654605-59654605 on Build GRCh38
rs752137198 Chr.10: 59654439-59654439 on Build GRCh38
rs770049784 Chr.10: 59654682-59654682 on Build GRCh38
rs762653515 Chr.10: 59654562-59654562 on Build GRCh38
rs56370926 Chr.10: 59654418-59654418 on Build GRCh38
rs763613570 Chr.10: 59654401-59654401 on Build GRCh38
rs948294238 Chr.10: 59654724-59654724 on Build GRCh38
rs757446582 Chr.10: 59654620-59654620 on Build GRCh38
rs752716124 Chr.10: 59654448-59654448 on Build GRCh38
rs779861267 Chr.10: 59654621-59654621 on Build GRCh38
rs145850404 Chr.10: 59654372-59654372 on Build GRCh38
rs537376284 Chr.10: 59654589-59654589 on Build GRCh38
rs534894384 Chr.10: 59654699-59654699 on Build GRCh38
rs964607994 Chr.10: 59654455-59654455 on Build GRCh38
rs939382895 Chr.10: 59654771-59654771 on Build GRCh38
rs999940296 Chr.10: 59654314-59654314 on Build GRCh38
rs746994628 Chr.10: 59654519-59654519 on Build GRCh38
rs770439309 Chr.10: 59654358-59654358 on Build GRCh38
rs761563562 Chr.10: 59654409-59654409 on Build GRCh38
rs777875410 Chr.10: 59654480-59654480 on Build GRCh38
rs151099964 Chr.10: 59654561-59654561 on Build GRCh38
rs769876769 Chr.10: 59654316-59654316 on Build GRCh38
rs952268987 Chr.10: 59654565-59654565 on Build GRCh38
rs368640774 Chr.10: 59654582-59654582 on Build GRCh38
rs11006658 Chr.10: 59654652-59654652 on Build GRCh38
rs753831803 Chr.10: 59654740-59654740 on Build GRCh38
rs756157160 Chr.10: 59654604-59654604 on Build GRCh38
rs1031926137 Chr.10: 59654318-59654318 on Build GRCh38
rs764792405 Chr.10: 59654415-59654415 on Build GRCh38
rs142913788 Chr.10: 59654549-59654549 on Build GRCh38
rs760554253 Chr.10: 59654615-59654615 on Build GRCh38
rs754083929 Chr.10: 59654619-59654619 on Build GRCh38
rs771888471 Chr.10: 59654493-59654493 on Build GRCh38
rs768695997 Chr.10: 59654633-59654633 on Build GRCh38
rs182954890 Chr.10: 59654386-59654386 on Build GRCh38
rs951152506 Chr.10: 59654598-59654598 on Build GRCh38
rs755520435 Chr.10: 59654443-59654443 on Build GRCh38
rs759214728 Chr.10: 59654474-59654474 on Build GRCh38
rs745641085 Chr.10: 59654381-59654381 on Build GRCh38
rs746798987 Chr.10: 59654632-59654632 on Build GRCh38
rs779773338 Chr.10: 59654499-59654499 on Build GRCh38
rs766901996 Chr.10: 59654568-59654568 on Build GRCh38
rs974772864 Chr.10: 59654458-59654458 on Build GRCh38
rs761216589 Chr.10: 59654554-59654554 on Build GRCh38
rs376826336 Chr.10: 59654581-59654581 on Build GRCh38
rs769288819 Chr.10: 59654558-59654558 on Build GRCh38
rs756420720 Chr.10: 59654478-59654478 on Build GRCh38
rs371337386 Chr.10: 59654446-59654446 on Build GRCh38
rs372622560 Chr.10: 59654601-59654601 on Build GRCh38
rs868563722 Chr.10: 59654373-59654373 on Build GRCh38
rs559499490 Chr.10: 59654391-59654391 on Build GRCh38
rs749448585 Chr.10: 59654485-59654485 on Build GRCh38
rs775414333 Chr.10: 59654390-59654390 on Build GRCh38
rs571005294 Chr.10: 59654691-59654691 on Build GRCh38
rs371109066 Chr.10: 59654592-59654592 on Build GRCh38
rs927983882 Chr.10: 59654756-59654756 on Build GRCh38
rs766782417 Chr.10: 59654430-59654430 on Build GRCh38
rs142544901 Chr.10: 59654573-59654573 on Build GRCh38

View Full Product Details