Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45240202-45240730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT13
Entrez Gene ID
57586
Gene Name
synaptotagmin 13
Gene Aliases
-
UniGene
Hs.436643
Gene Chromosome Location
Chr.11: 45240302-45286333 on Build GRCh38
Transcripts
NM_001247987.1 NM_020826.2

SNP Detail

SNP ID Chromosome Location
rs545730736 Chr.11: 45240268-45240268 on Build GRCh38
rs766299972 Chr.11: 45240291-45240291 on Build GRCh38
rs995770174 Chr.11: 45240357-45240357 on Build GRCh38
rs951496324 Chr.11: 45240224-45240224 on Build GRCh38
rs1001557918 Chr.11: 45240572-45240572 on Build GRCh38
rs544931723 Chr.11: 45240675-45240675 on Build GRCh38
rs931255626 Chr.11: 45240440-45240440 on Build GRCh38
rs8929 Chr.11: 45240575-45240575 on Build GRCh38
rs970476907 Chr.11: 45240669-45240669 on Build GRCh38
rs938184025 Chr.11: 45240351-45240351 on Build GRCh38
rs780828455 Chr.11: 45240344-45240344 on Build GRCh38
rs755779963 Chr.11: 45240402-45240402 on Build GRCh38
rs573759036 Chr.11: 45240535-45240535 on Build GRCh38
rs934614640 Chr.11: 45240601-45240601 on Build GRCh38
rs145041027 Chr.11: 45240273-45240273 on Build GRCh38
rs141207837 Chr.11: 45240460-45240460 on Build GRCh38
rs901899606 Chr.11: 45240534-45240534 on Build GRCh38
rs2863173 Chr.11: 45240691-45240691 on Build GRCh38
rs567435191 Chr.11: 45240261-45240261 on Build GRCh38
rs757601399 Chr.11: 45240236-45240238 on Build GRCh38
rs577518093 Chr.11: 45240335-45240335 on Build GRCh38
rs972445951 Chr.11: 45240679-45240679 on Build GRCh38
rs562357930 Chr.11: 45240503-45240503 on Build GRCh38
rs2902427 Chr.11: 45240705-45240705 on Build GRCh38
rs943092257 Chr.11: 45240666-45240666 on Build GRCh38
rs915744257 Chr.11: 45240496-45240496 on Build GRCh38
rs575720491 Chr.11: 45240566-45240566 on Build GRCh38
rs752416805 Chr.11: 45240401-45240401 on Build GRCh38
rs573042610 Chr.11: 45240583-45240583 on Build GRCh38
rs966037714 Chr.11: 45240276-45240276 on Build GRCh38
rs777201410 Chr.11: 45240431-45240431 on Build GRCh38
rs960093911 Chr.11: 45240462-45240462 on Build GRCh38
rs1035670240 Chr.11: 45240432-45240432 on Build GRCh38
rs151149235 Chr.11: 45240412-45240412 on Build GRCh38
rs1051410741 Chr.11: 45240603-45240603 on Build GRCh38
rs951669002 Chr.11: 45240378-45240378 on Build GRCh38
rs917677384 Chr.11: 45240656-45240659 on Build GRCh38
rs149113991 Chr.11: 45240304-45240304 on Build GRCh38
rs771369622 Chr.11: 45240690-45240690 on Build GRCh38
rs890186528 Chr.11: 45240313-45240313 on Build GRCh38
rs928685932 Chr.11: 45240252-45240252 on Build GRCh38
rs546765909 Chr.11: 45240279-45240279 on Build GRCh38
rs896809935 Chr.11: 45240417-45240417 on Build GRCh38
rs143009829 Chr.11: 45240306-45240306 on Build GRCh38
rs559248255 Chr.11: 45240325-45240325 on Build GRCh38
rs947171606 Chr.11: 45240512-45240512 on Build GRCh38
rs949062282 Chr.11: 45240370-45240370 on Build GRCh38
rs982812301 Chr.11: 45240249-45240249 on Build GRCh38
rs1051442551 Chr.11: 45240307-45240307 on Build GRCh38
rs1027259188 Chr.11: 45240258-45240258 on Build GRCh38
rs978627134 Chr.11: 45240550-45240550 on Build GRCh38
rs947320016 Chr.11: 45240274-45240274 on Build GRCh38
rs959806680 Chr.11: 45240599-45240599 on Build GRCh38
rs1027610122 Chr.11: 45240366-45240366 on Build GRCh38
rs890155418 Chr.11: 45240616-45240616 on Build GRCh38
rs1021230858 Chr.11: 45240302-45240302 on Build GRCh38
rs991559690 Chr.11: 45240477-45240477 on Build GRCh38
rs754711833 Chr.11: 45240334-45240334 on Build GRCh38
rs979689895 Chr.11: 45240313-45240314 on Build GRCh38
rs900034993 Chr.11: 45240345-45240345 on Build GRCh38
rs542778243 Chr.11: 45240557-45240557 on Build GRCh38

View Full Product Details