Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39847807-39848318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIT1
Entrez Gene ID
54802
Gene Name
tRNA isopentenyltransferase 1
Gene Aliases
GRO1, IPPT, IPT, IPTase, MOD5, hGRO1
UniGene
Hs.356554
Gene Chromosome Location
Chr.1: 39841031-39883511 on Build GRCh38
Transcripts
NM_001312691.1 NM_001312692.1 NM_017646.5 NR_132401.1 NR_132402.1 NR_132403.1 NR_132404.1 NR_132405.1 NR_132406.1 NR_132407.1 NR_132408.1 NR_132409.1 NR_132410.1 NR_132412.1 NR_132413.1 NR_132414.1 NR_132415.1 XM_005270954.2 XM_006710706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6808924 c.815+1G>A p.? Chr.1: 39847985-39847985 on Build GRCh38
6994349 c.742C>T p.L248F Chr.1: 39848059-39848059 on Build GRCh38
131052 c.733G>T p.D245Y Chr.1: 39848068-39848068 on Build GRCh38
3741280 c.784C>T p.R262C Chr.1: 39848017-39848017 on Build GRCh38
6056664 c.730G>T p.V244L Chr.1: 39848071-39848071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889268715 Chr.1: 39848235-39848235 on Build GRCh38
rs752309680 Chr.1: 39847939-39847939 on Build GRCh38
rs953557538 Chr.1: 39847934-39847934 on Build GRCh38
rs750845995 Chr.1: 39847960-39847960 on Build GRCh38
rs528431829 Chr.1: 39847919-39847919 on Build GRCh38
rs140406438 Chr.1: 39848086-39848086 on Build GRCh38
rs775171278 Chr.1: 39848022-39848022 on Build GRCh38
rs757577525 Chr.1: 39847940-39847940 on Build GRCh38
rs1000324503 Chr.1: 39847921-39847921 on Build GRCh38
rs948430265 Chr.1: 39848257-39848257 on Build GRCh38
rs201156716 Chr.1: 39848017-39848017 on Build GRCh38
rs1008158751 Chr.1: 39848295-39848295 on Build GRCh38
rs975558410 Chr.1: 39847967-39847967 on Build GRCh38
rs749150087 Chr.1: 39848013-39848013 on Build GRCh38
rs894231672 Chr.1: 39847845-39847845 on Build GRCh38
rs751672103 Chr.1: 39847977-39847977 on Build GRCh38
rs149918734 Chr.1: 39848105-39848105 on Build GRCh38
rs765554362 Chr.1: 39847948-39847948 on Build GRCh38
rs769581003 Chr.1: 39848004-39848004 on Build GRCh38
rs771524834 Chr.1: 39848135-39848135 on Build GRCh38
rs750744539 Chr.1: 39848062-39848062 on Build GRCh38
rs375323353 Chr.1: 39848281-39848281 on Build GRCh38
rs549894930 Chr.1: 39847923-39847923 on Build GRCh38
rs539069256 Chr.1: 39848053-39848053 on Build GRCh38
rs950054587 Chr.1: 39848163-39848163 on Build GRCh38
rs567143261 Chr.1: 39847852-39847852 on Build GRCh38
rs768177032 Chr.1: 39848142-39848142 on Build GRCh38
rs555173540 Chr.1: 39848176-39848176 on Build GRCh38
rs1333742 Chr.1: 39848177-39848177 on Build GRCh38
rs760296667 Chr.1: 39848140-39848140 on Build GRCh38
rs1045880594 Chr.1: 39847891-39847891 on Build GRCh38
rs536311622 Chr.1: 39847918-39847918 on Build GRCh38
rs918476473 Chr.1: 39848074-39848074 on Build GRCh38
rs372034946 Chr.1: 39847988-39847988 on Build GRCh38
rs903221409 Chr.1: 39848199-39848199 on Build GRCh38
rs769920760 Chr.1: 39848227-39848244 on Build GRCh38
rs771577841 Chr.1: 39848020-39848020 on Build GRCh38
rs1056318501 Chr.1: 39847851-39847851 on Build GRCh38
rs868291294 Chr.1: 39848023-39848023 on Build GRCh38
rs367853894 Chr.1: 39847867-39847882 on Build GRCh38
rs145359484 Chr.1: 39848085-39848085 on Build GRCh38
rs1033289422 Chr.1: 39848233-39848233 on Build GRCh38
rs906925220 Chr.1: 39848095-39848095 on Build GRCh38
rs756261635 Chr.1: 39848099-39848099 on Build GRCh38
rs895806400 Chr.1: 39847875-39847875 on Build GRCh38
rs758797367 Chr.1: 39847961-39847961 on Build GRCh38
rs749025150 Chr.1: 39848113-39848113 on Build GRCh38
rs575718388 Chr.1: 39848162-39848162 on Build GRCh38
rs763465310 Chr.1: 39848066-39848066 on Build GRCh38
rs899861024 Chr.1: 39848146-39848146 on Build GRCh38
rs766934787 Chr.1: 39848069-39848069 on Build GRCh38
rs781497494 Chr.1: 39847994-39847994 on Build GRCh38
rs369974880 Chr.1: 39848134-39848134 on Build GRCh38
rs965050390 Chr.1: 39848001-39848001 on Build GRCh38
rs748666632 Chr.1: 39847899-39847899 on Build GRCh38
rs752872268 Chr.1: 39848090-39848090 on Build GRCh38
rs745840577 Chr.1: 39848132-39848132 on Build GRCh38
rs373052302 Chr.1: 39848050-39848050 on Build GRCh38
rs777410226 Chr.1: 39848012-39848012 on Build GRCh38
rs572413509 Chr.1: 39847864-39847879 on Build GRCh38
rs974014661 Chr.1: 39848244-39848244 on Build GRCh38
rs1030148276 Chr.1: 39847973-39847973 on Build GRCh38
rs774920510 Chr.1: 39847887-39847887 on Build GRCh38
rs759908563 Chr.1: 39848136-39848136 on Build GRCh38
rs12239482 Chr.1: 39848203-39848203 on Build GRCh38
rs939550358 Chr.1: 39848118-39848118 on Build GRCh38
rs770852136 Chr.1: 39848014-39848014 on Build GRCh38
rs981486991 Chr.1: 39848287-39848287 on Build GRCh38
rs752116890 Chr.1: 39848070-39848070 on Build GRCh38
rs1020513401 Chr.1: 39847917-39847917 on Build GRCh38
rs767697080 Chr.1: 39848159-39848159 on Build GRCh38
rs1022216185 Chr.1: 39847938-39847938 on Build GRCh38
rs1033173885 Chr.1: 39847926-39847926 on Build GRCh38
rs752755323 Chr.1: 39848172-39848172 on Build GRCh38
rs113012982 Chr.1: 39848100-39848100 on Build GRCh38
rs149680997 Chr.1: 39847995-39847995 on Build GRCh38
rs776356550 Chr.1: 39848143-39848143 on Build GRCh38
rs906728707 Chr.1: 39847893-39847893 on Build GRCh38
rs770729723 Chr.1: 39848121-39848121 on Build GRCh38
rs780769551 Chr.1: 39847964-39847964 on Build GRCh38
rs575935794 Chr.1: 39848249-39848249 on Build GRCh38
rs770243858 Chr.1: 39847900-39847900 on Build GRCh38
rs143719619 Chr.1: 39848209-39848209 on Build GRCh38
rs753609051 Chr.1: 39848248-39848248 on Build GRCh38

View Full Product Details