Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51242222-51242713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAZAP2
Entrez Gene ID
9802
Gene Name
DAZ associated protein 2
Gene Aliases
PRTB
UniGene
Hs.369761
Gene Chromosome Location
Chr.12: 51238724-51246717 on Build GRCh38
Transcripts
NM_001136264.1 NM_001136266.1 NM_001136267.1 NM_001136268.1 NM_001136269.1 NM_014764.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4042671 c.361C>T p.R121C Chr.12: 51242389-51242389 on Build GRCh38
3671099 c.459G>T p.K153N Chr.12: 51242410-51242410 on Build GRCh38
3935995 c.304T>A p.S102T Chr.12: 51242332-51242332 on Build GRCh38
3935997 c.363T>G p.R121R Chr.12: 51242391-51242391 on Build GRCh38
3671098 c.382G>T p.G128W Chr.12: 51242410-51242410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768045110 Chr.12: 51242405-51242405 on Build GRCh38
rs762089686 Chr.12: 51242331-51242331 on Build GRCh38
rs778733375 Chr.12: 51242279-51242279 on Build GRCh38
rs747921155 Chr.12: 51242280-51242280 on Build GRCh38
rs776111164 Chr.12: 51242413-51242413 on Build GRCh38
rs747907569 Chr.12: 51242368-51242368 on Build GRCh38
rs1015602736 Chr.12: 51242663-51242663 on Build GRCh38
rs751901959 Chr.12: 51242323-51242323 on Build GRCh38
rs144235964 Chr.12: 51242390-51242390 on Build GRCh38
rs373137618 Chr.12: 51242515-51242515 on Build GRCh38
rs1049467 Chr.12: 51242475-51242475 on Build GRCh38
rs549110284 Chr.12: 51242334-51242334 on Build GRCh38
rs778732954 Chr.12: 51242366-51242366 on Build GRCh38
rs201854240 Chr.12: 51242481-51242481 on Build GRCh38
rs759605132 Chr.12: 51242301-51242301 on Build GRCh38
rs748645128 Chr.12: 51242483-51242483 on Build GRCh38
rs750160340 Chr.12: 51242339-51242339 on Build GRCh38
rs758186736 Chr.12: 51242449-51242449 on Build GRCh38
rs752647176 Chr.12: 51242533-51242533 on Build GRCh38
rs746134764 Chr.12: 51242388-51242388 on Build GRCh38
rs772739085 Chr.12: 51242500-51242500 on Build GRCh38
rs754446793 Chr.12: 51242432-51242432 on Build GRCh38
rs776086673 Chr.12: 51242423-51242423 on Build GRCh38
rs1044302342 Chr.12: 51242252-51242252 on Build GRCh38
rs764040405 Chr.12: 51242542-51242542 on Build GRCh38
rs771571804 Chr.12: 51242306-51242306 on Build GRCh38
rs751297697 Chr.12: 51242561-51242561 on Build GRCh38
rs866151968 Chr.12: 51242480-51242480 on Build GRCh38
rs746420669 Chr.12: 51242598-51242598 on Build GRCh38
rs768400345 Chr.12: 51242585-51242586 on Build GRCh38
rs775952142 Chr.12: 51242312-51242312 on Build GRCh38
rs774787428 Chr.12: 51242395-51242395 on Build GRCh38
rs201345162 Chr.12: 51242294-51242294 on Build GRCh38
rs776209061 Chr.12: 51242516-51242516 on Build GRCh38
rs764700099 Chr.12: 51242526-51242526 on Build GRCh38
rs138210383 Chr.12: 51242360-51242360 on Build GRCh38
rs567234272 Chr.12: 51242685-51242685 on Build GRCh38
rs377252029 Chr.12: 51242463-51242463 on Build GRCh38
rs938626245 Chr.12: 51242270-51242270 on Build GRCh38
rs948590565 Chr.12: 51242247-51242247 on Build GRCh38
rs771123020 Chr.12: 51242285-51242285 on Build GRCh38
rs11555475 Chr.12: 51242410-51242410 on Build GRCh38
rs769916188 Chr.12: 51242306-51242306 on Build GRCh38
rs374168708 Chr.12: 51242482-51242482 on Build GRCh38
rs759069912 Chr.12: 51242519-51242519 on Build GRCh38
rs763553723 Chr.12: 51242314-51242314 on Build GRCh38
rs748284553 Chr.12: 51242668-51242668 on Build GRCh38
rs377433711 Chr.12: 51242631-51242631 on Build GRCh38
rs771825570 Chr.12: 51242382-51242382 on Build GRCh38
rs772970562 Chr.12: 51242406-51242406 on Build GRCh38
rs1011272989 Chr.12: 51242295-51242295 on Build GRCh38
rs755664500 Chr.12: 51242341-51242341 on Build GRCh38
rs749383352 Chr.12: 51242478-51242478 on Build GRCh38
rs571090536 Chr.12: 51242396-51242396 on Build GRCh38
rs779908029 Chr.12: 51242472-51242472 on Build GRCh38
rs780242085 Chr.12: 51242581-51242581 on Build GRCh38
rs754899504 Chr.12: 51242361-51242361 on Build GRCh38
rs1053166394 Chr.12: 51242281-51242281 on Build GRCh38
rs773665805 Chr.12: 51242511-51242511 on Build GRCh38
rs753334286 Chr.12: 51242417-51242417 on Build GRCh38
rs766156924 Chr.12: 51242415-51242415 on Build GRCh38
rs764539554 Chr.12: 51242319-51242319 on Build GRCh38
rs758289456 Chr.12: 51242536-51242536 on Build GRCh38
rs776378700 Chr.12: 51242646-51242647 on Build GRCh38
rs566133877 Chr.12: 51242384-51242384 on Build GRCh38
rs35097159 Chr.12: 51242264-51242265 on Build GRCh38
rs746969932 Chr.12: 51242564-51242566 on Build GRCh38
rs780268345 Chr.12: 51242477-51242477 on Build GRCh38
rs190665763 Chr.12: 51242538-51242538 on Build GRCh38
rs749523180 Chr.12: 51242592-51242592 on Build GRCh38
rs907092203 Chr.12: 51242263-51242263 on Build GRCh38
rs765212285 Chr.12: 51242445-51242445 on Build GRCh38
rs529245061 Chr.12: 51242262-51242262 on Build GRCh38
rs752632409 Chr.12: 51242447-51242447 on Build GRCh38
rs551887665 Chr.12: 51242393-51242393 on Build GRCh38
rs755057505 Chr.12: 51242607-51242607 on Build GRCh38
rs757111801 Chr.12: 51242578-51242578 on Build GRCh38
rs987071954 Chr.12: 51242619-51242619 on Build GRCh38
rs149578797 Chr.12: 51242389-51242389 on Build GRCh38
rs779753588 Chr.12: 51242349-51242349 on Build GRCh38

View Full Product Details