Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57335236-57335593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPO11
Entrez Gene ID
23626
Gene Name
SPO11, initiator of meiotic double stranded breaks
Gene Aliases
CT35, SPATA43, TOPVIA
UniGene
Hs.159737
Gene Chromosome Location
Chr.20: 57329757-57343993 on Build GRCh38
Transcripts
NM_012444.2 NM_198265.1 XM_005260379.3 XM_005260380.3 XM_005260381.3 XM_005260382.4 XM_011528756.2 XM_011528757.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
578070 c.598-6T>A p.? Chr.20: 57335413-57335413 on Build GRCh38
5035290 c.598-5A>T p.? Chr.20: 57335414-57335414 on Build GRCh38
3548082 c.633G>A p.R211R Chr.20: 57335454-57335454 on Build GRCh38
7391795 c.602T>C p.V201A Chr.20: 57335423-57335423 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1044797374 Chr.20: 57335462-57335462 on Build GRCh38
rs753266523 Chr.20: 57335385-57335385 on Build GRCh38
rs763196022 Chr.20: 57335327-57335327 on Build GRCh38
rs1030240743 Chr.20: 57335539-57335539 on Build GRCh38
rs756701075 Chr.20: 57335423-57335423 on Build GRCh38
rs780342271 Chr.20: 57335459-57335459 on Build GRCh38
rs751737152 Chr.20: 57335405-57335405 on Build GRCh38
rs775256954 Chr.20: 57335274-57335274 on Build GRCh38
rs773040526 Chr.20: 57335493-57335493 on Build GRCh38
rs946978056 Chr.20: 57335383-57335383 on Build GRCh38
rs903607349 Chr.20: 57335466-57335466 on Build GRCh38
rs1052718069 Chr.20: 57335524-57335524 on Build GRCh38
rs28368083 Chr.20: 57335533-57335533 on Build GRCh38
rs764628667 Chr.20: 57335404-57335405 on Build GRCh38
rs754164090 Chr.20: 57335435-57335435 on Build GRCh38
rs181329544 Chr.20: 57335526-57335526 on Build GRCh38
rs541651106 Chr.20: 57335322-57335322 on Build GRCh38
rs140465905 Chr.20: 57335310-57335310 on Build GRCh38
rs764361442 Chr.20: 57335427-57335427 on Build GRCh38
rs750506809 Chr.20: 57335519-57335519 on Build GRCh38
rs1007519577 Chr.20: 57335563-57335563 on Build GRCh38
rs758531602 Chr.20: 57335453-57335453 on Build GRCh38
rs953408215 Chr.20: 57335269-57335269 on Build GRCh38
rs749058928 Chr.20: 57335494-57335494 on Build GRCh38
rs561827166 Chr.20: 57335366-57335366 on Build GRCh38
rs991105384 Chr.20: 57335352-57335352 on Build GRCh38
rs1016271074 Chr.20: 57335564-57335564 on Build GRCh38
rs757585150 Chr.20: 57335436-57335436 on Build GRCh38
rs759003934 Chr.20: 57335413-57335413 on Build GRCh38
rs28368082 Chr.20: 57335452-57335452 on Build GRCh38
rs17406460 Chr.20: 57335426-57335426 on Build GRCh38
rs748034051 Chr.20: 57335469-57335469 on Build GRCh38
rs3764674 Chr.20: 57335486-57335486 on Build GRCh38
rs371443486 Chr.20: 57335406-57335406 on Build GRCh38
rs150374893 Chr.20: 57335565-57335565 on Build GRCh38
rs758789088 Chr.20: 57335404-57335404 on Build GRCh38
rs6025499 Chr.20: 57335557-57335557 on Build GRCh38
rs761869741 Chr.20: 57335471-57335471 on Build GRCh38
rs28368081 Chr.20: 57335414-57335414 on Build GRCh38
rs371863196 Chr.20: 57335535-57335535 on Build GRCh38
rs370477685 Chr.20: 57335506-57335506 on Build GRCh38
rs953378723 Chr.20: 57335550-57335550 on Build GRCh38
rs879184032 Chr.20: 57335495-57335495 on Build GRCh38
rs779238568 Chr.20: 57335441-57335441 on Build GRCh38
rs915504247 Chr.20: 57335361-57335361 on Build GRCh38

View Full Product Details