Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50517188-50517706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAPH2
Entrez Gene ID
29781
Gene Name
non-SMC condensin II complex subunit H2
Gene Aliases
CAPH2
UniGene
Hs.730607
Gene Chromosome Location
Chr.22: 50508216-50524780 on Build GRCh38
Transcripts
NM_001185011.1 NM_014551.4 NM_152299.3 XM_005261912.4 XM_011530685.2 XM_017028793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035492 c.267-2A>G p.? Chr.22: 50517575-50517575 on Build GRCh38
6230523 c.231C>T p.L77L Chr.22: 50517447-50517447 on Build GRCh38
3065921 c.263A>G p.K88R Chr.22: 50517479-50517479 on Build GRCh38
6872041 c.331C>T p.P111S Chr.22: 50517641-50517641 on Build GRCh38
3065922 c.263A>G p.K88R Chr.22: 50517479-50517479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745977314 Chr.22: 50517456-50517456 on Build GRCh38
rs988277532 Chr.22: 50517290-50517290 on Build GRCh38
rs779404124 Chr.22: 50517348-50517348 on Build GRCh38
rs758751077 Chr.22: 50517563-50517563 on Build GRCh38
rs113288865 Chr.22: 50517662-50517662 on Build GRCh38
rs529394061 Chr.22: 50517331-50517331 on Build GRCh38
rs773949592 Chr.22: 50517493-50517493 on Build GRCh38
rs973804277 Chr.22: 50517222-50517222 on Build GRCh38
rs976187762 Chr.22: 50517312-50517312 on Build GRCh38
rs368250304 Chr.22: 50517614-50517614 on Build GRCh38
rs1013426189 Chr.22: 50517354-50517354 on Build GRCh38
rs749111633 Chr.22: 50517534-50517534 on Build GRCh38
rs950199829 Chr.22: 50517317-50517317 on Build GRCh38
rs377723640 Chr.22: 50517447-50517447 on Build GRCh38
rs920647844 Chr.22: 50517236-50517236 on Build GRCh38
rs372044931 Chr.22: 50517635-50517635 on Build GRCh38
rs752216231 Chr.22: 50517501-50517501 on Build GRCh38
rs770278461 Chr.22: 50517545-50517545 on Build GRCh38
rs201599631 Chr.22: 50517642-50517642 on Build GRCh38
rs199689462 Chr.22: 50517597-50517597 on Build GRCh38
rs756618804 Chr.22: 50517424-50517424 on Build GRCh38
rs779301926 Chr.22: 50517519-50517519 on Build GRCh38
rs867295725 Chr.22: 50517281-50517281 on Build GRCh38
rs905140475 Chr.22: 50517378-50517378 on Build GRCh38
rs937344538 Chr.22: 50517249-50517249 on Build GRCh38
rs772231508 Chr.22: 50517459-50517459 on Build GRCh38
rs770819683 Chr.22: 50517585-50517585 on Build GRCh38
rs562206785 Chr.22: 50517276-50517276 on Build GRCh38
rs745695877 Chr.22: 50517653-50517653 on Build GRCh38
rs753094280 Chr.22: 50517422-50517422 on Build GRCh38
rs189560836 Chr.22: 50517353-50517353 on Build GRCh38
rs141239310 Chr.22: 50517522-50517522 on Build GRCh38
rs769089097 Chr.22: 50517344-50517344 on Build GRCh38
rs200663937 Chr.22: 50517687-50517687 on Build GRCh38
rs765827468 Chr.22: 50517615-50517615 on Build GRCh38
rs750201844 Chr.22: 50517399-50517399 on Build GRCh38
rs940812751 Chr.22: 50517349-50517349 on Build GRCh38
rs776252361 Chr.22: 50517609-50517609 on Build GRCh38
rs373870538 Chr.22: 50517577-50517577 on Build GRCh38
rs765737574 Chr.22: 50517560-50517560 on Build GRCh38
rs1136705 Chr.22: 50517443-50517443 on Build GRCh38
rs780264028 Chr.22: 50517567-50517567 on Build GRCh38
rs750830683 Chr.22: 50517619-50517619 on Build GRCh38
rs766538568 Chr.22: 50517626-50517626 on Build GRCh38
rs768040574 Chr.22: 50517420-50517420 on Build GRCh38
rs762750604 Chr.22: 50517486-50517486 on Build GRCh38
rs771631683 Chr.22: 50517591-50517591 on Build GRCh38
rs771854678 Chr.22: 50517658-50517658 on Build GRCh38
rs749518820 Chr.22: 50517376-50517376 on Build GRCh38
rs762390370 Chr.22: 50517557-50517557 on Build GRCh38
rs62239486 Chr.22: 50517554-50517554 on Build GRCh38
rs771184206 Chr.22: 50517382-50517382 on Build GRCh38
rs756641153 Chr.22: 50517505-50517505 on Build GRCh38
rs769765812 Chr.22: 50517471-50517471 on Build GRCh38
rs750814257 Chr.22: 50517562-50517562 on Build GRCh38
rs760261671 Chr.22: 50517512-50517513 on Build GRCh38
rs755043609 Chr.22: 50517578-50517578 on Build GRCh38
rs372646427 Chr.22: 50517589-50517589 on Build GRCh38
rs779692981 Chr.22: 50517675-50517675 on Build GRCh38
rs761692582 Chr.22: 50517390-50517390 on Build GRCh38
rs775617474 Chr.22: 50517466-50517466 on Build GRCh38
rs1036692820 Chr.22: 50517603-50517603 on Build GRCh38
rs199513142 Chr.22: 50517582-50517582 on Build GRCh38
rs754323391 Chr.22: 50517556-50517556 on Build GRCh38
rs760022279 Chr.22: 50517567-50517568 on Build GRCh38
rs747097527 Chr.22: 50517527-50517527 on Build GRCh38
rs749654643 Chr.22: 50517444-50517444 on Build GRCh38
rs200222785 Chr.22: 50517551-50517551 on Build GRCh38
rs373229326 Chr.22: 50517386-50517386 on Build GRCh38
rs140833290 Chr.22: 50517453-50517453 on Build GRCh38
rs991471228 Chr.22: 50517498-50517498 on Build GRCh38
rs965059019 Chr.22: 50517499-50517499 on Build GRCh38
rs377259008 Chr.22: 50517526-50517526 on Build GRCh38
rs374607010 Chr.22: 50517598-50517598 on Build GRCh38
rs569238129 Chr.22: 50517351-50517351 on Build GRCh38
rs768191519 Chr.22: 50517680-50517680 on Build GRCh38
rs1008858456 Chr.22: 50517590-50517590 on Build GRCh38
rs758663307 Chr.22: 50517523-50517523 on Build GRCh38
rs150150677 Chr.22: 50517631-50517631 on Build GRCh38
rs35488039 Chr.22: 50517292-50517293 on Build GRCh38
rs776198405 Chr.22: 50517416-50517416 on Build GRCh38
rs759222844 Chr.22: 50517550-50517550 on Build GRCh38
rs764608213 Chr.22: 50517510-50517510 on Build GRCh38
rs760167692 Chr.22: 50517553-50517553 on Build GRCh38
rs1050311517 Chr.22: 50517373-50517373 on Build GRCh38
rs995094234 Chr.22: 50517674-50517674 on Build GRCh38
rs778070413 Chr.22: 50517425-50517425 on Build GRCh38
rs1008724072 Chr.22: 50517389-50517389 on Build GRCh38
rs75088399 Chr.22: 50517210-50517210 on Build GRCh38
rs774879640 Chr.22: 50517552-50517552 on Build GRCh38
rs777135387 Chr.22: 50517549-50517550 on Build GRCh38
rs535638620 Chr.22: 50517495-50517495 on Build GRCh38
rs956591417 Chr.22: 50517448-50517448 on Build GRCh38
rs772938145 Chr.22: 50517473-50517473 on Build GRCh38
rs956510086 Chr.22: 50517250-50517250 on Build GRCh38
rs749678153 Chr.22: 50517298-50517298 on Build GRCh38
rs777591184 Chr.22: 50517579-50517579 on Build GRCh38
rs962182734 Chr.22: 50517220-50517221 on Build GRCh38
rs763342233 Chr.22: 50517623-50517623 on Build GRCh38
rs147758819 Chr.22: 50517404-50517404 on Build GRCh38
rs922191584 Chr.22: 50517328-50517328 on Build GRCh38
rs746714891 Chr.22: 50517678-50517678 on Build GRCh38
rs759067436 Chr.22: 50517494-50517494 on Build GRCh38
rs374588731 Chr.22: 50517531-50517531 on Build GRCh38
rs768262618 Chr.22: 50517607-50517607 on Build GRCh38
rs368713408 Chr.22: 50517394-50517394 on Build GRCh38
rs772644324 Chr.22: 50517364-50517364 on Build GRCh38
rs985430239 Chr.22: 50517533-50517533 on Build GRCh38
rs752766206 Chr.22: 50517640-50517640 on Build GRCh38
rs552455534 Chr.22: 50517383-50517383 on Build GRCh38
rs574846073 Chr.22: 50517638-50517638 on Build GRCh38
rs949439055 Chr.22: 50517564-50517564 on Build GRCh38
rs770660298 Chr.22: 50517541-50517541 on Build GRCh38
rs766001548 Chr.22: 50517406-50517406 on Build GRCh38
rs868561263 Chr.22: 50517261-50517261 on Build GRCh38
rs113818181 Chr.22: 50517664-50517664 on Build GRCh38
rs759634125 Chr.22: 50517384-50517384 on Build GRCh38
rs368611781 Chr.22: 50517588-50517588 on Build GRCh38
rs762848166 Chr.22: 50517542-50517542 on Build GRCh38
rs932223632 Chr.22: 50517335-50517335 on Build GRCh38
rs370859039 Chr.22: 50517528-50517528 on Build GRCh38
rs550934220 Chr.22: 50517342-50517342 on Build GRCh38
rs192993579 Chr.22: 50517517-50517517 on Build GRCh38
rs754416891 Chr.22: 50517419-50517419 on Build GRCh38
rs748077061 Chr.22: 50517469-50517469 on Build GRCh38
rs766023747 Chr.22: 50517492-50517492 on Build GRCh38
rs375327095 Chr.22: 50517387-50517387 on Build GRCh38
rs754160402 Chr.22: 50517511-50517511 on Build GRCh38
rs774108172 Chr.22: 50517549-50517549 on Build GRCh38
rs912289279 Chr.22: 50517294-50517294 on Build GRCh38
rs138641341 Chr.22: 50517637-50517637 on Build GRCh38

View Full Product Details