Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108469522-108470117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCC2
Entrez Gene ID
9648
Gene Name
GRIP and coiled-coil domain containing 2
Gene Aliases
GCC185, RANBP2L4, REN53
UniGene
Hs.436505
Gene Chromosome Location
Chr.2: 108448561-108509398 on Build GRCh38
Transcripts
NM_181453.3 NR_028063.1 XM_006712870.3 XM_006712871.1 XM_006712872.3 XM_011512213.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6086544 c.350A>T p.H117L Chr.2: 108469679-108469679 on Build GRCh38
6669884 c.540_541insA p.L183fs*6 Chr.2: 108469869-108469870 on Build GRCh38
1564873 c.490T>C p.L164L Chr.2: 108469819-108469819 on Build GRCh38
6086545 c.328G>T p.V110L Chr.2: 108469657-108469657 on Build GRCh38
5576672 c.336G>A p.K112K Chr.2: 108469665-108469665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747569256 Chr.2: 108469846-108469846 on Build GRCh38
rs372290189 Chr.2: 108469689-108469689 on Build GRCh38
rs763201366 Chr.2: 108469876-108469876 on Build GRCh38
rs766165187 Chr.2: 108469658-108469658 on Build GRCh38
rs751554456 Chr.2: 108469888-108469888 on Build GRCh38
rs748848872 Chr.2: 108469690-108469690 on Build GRCh38
rs769292941 Chr.2: 108469683-108469683 on Build GRCh38
rs149070749 Chr.2: 108470071-108470071 on Build GRCh38
rs761036599 Chr.2: 108469626-108469626 on Build GRCh38
rs766179585 Chr.2: 108470058-108470058 on Build GRCh38
rs963398140 Chr.2: 108469591-108469591 on Build GRCh38
rs757912028 Chr.2: 108470086-108470086 on Build GRCh38
rs1057397524 Chr.2: 108470040-108470040 on Build GRCh38
rs373554463 Chr.2: 108469753-108469753 on Build GRCh38
rs776808644 Chr.2: 108469714-108469714 on Build GRCh38
rs938366734 Chr.2: 108469712-108469712 on Build GRCh38
rs1007980786 Chr.2: 108469949-108469949 on Build GRCh38
rs1015316852 Chr.2: 108469631-108469631 on Build GRCh38
rs1017644891 Chr.2: 108469587-108469587 on Build GRCh38
rs573626780 Chr.2: 108469592-108469592 on Build GRCh38
rs771300150 Chr.2: 108469860-108469860 on Build GRCh38
rs927482380 Chr.2: 108470009-108470009 on Build GRCh38
rs376279109 Chr.2: 108470091-108470091 on Build GRCh38
rs903042861 Chr.2: 108469803-108469803 on Build GRCh38
rs754092294 Chr.2: 108469609-108469609 on Build GRCh38
rs759864429 Chr.2: 108469863-108469863 on Build GRCh38
rs376738248 Chr.2: 108469661-108469661 on Build GRCh38
rs150351597 Chr.2: 108469884-108469884 on Build GRCh38
rs751925348 Chr.2: 108469611-108469614 on Build GRCh38
rs868566268 Chr.2: 108469721-108469721 on Build GRCh38
rs755932248 Chr.2: 108469917-108469917 on Build GRCh38
rs564390284 Chr.2: 108469994-108469994 on Build GRCh38
rs775920739 Chr.2: 108469868-108469868 on Build GRCh38
rs370827700 Chr.2: 108469674-108469674 on Build GRCh38
rs368534626 Chr.2: 108469678-108469678 on Build GRCh38
rs915570315 Chr.2: 108469754-108469754 on Build GRCh38
rs752749562 Chr.2: 108469781-108469781 on Build GRCh38
rs758608147 Chr.2: 108469766-108469766 on Build GRCh38
rs759830991 Chr.2: 108469740-108469740 on Build GRCh38
rs746381453 Chr.2: 108469966-108469966 on Build GRCh38
rs766775912 Chr.2: 108469710-108469710 on Build GRCh38
rs552450339 Chr.2: 108469916-108469916 on Build GRCh38
rs899169070 Chr.2: 108469998-108469998 on Build GRCh38
rs768163382 Chr.2: 108469831-108469831 on Build GRCh38
rs937580013 Chr.2: 108470029-108470029 on Build GRCh38
rs767632667 Chr.2: 108470007-108470007 on Build GRCh38
rs1017594058 Chr.2: 108469955-108469955 on Build GRCh38
rs755093633 Chr.2: 108469615-108469615 on Build GRCh38
rs1043247481 Chr.2: 108469973-108469973 on Build GRCh38
rs761016613 Chr.2: 108469706-108469706 on Build GRCh38
rs761923647 Chr.2: 108469895-108469895 on Build GRCh38
rs779761244 Chr.2: 108469921-108469921 on Build GRCh38
rs778423603 Chr.2: 108469948-108469948 on Build GRCh38
rs752781355 Chr.2: 108469638-108469638 on Build GRCh38
rs539963771 Chr.2: 108469881-108469881 on Build GRCh38
rs755348339 Chr.2: 108470021-108470021 on Build GRCh38
rs764264510 Chr.2: 108469779-108469779 on Build GRCh38
rs751556782 Chr.2: 108469780-108469780 on Build GRCh38
rs1026821005 Chr.2: 108469736-108469736 on Build GRCh38
rs757141762 Chr.2: 108469672-108469672 on Build GRCh38
rs781607007 Chr.2: 108469964-108469964 on Build GRCh38
rs755917409 Chr.2: 108469815-108469815 on Build GRCh38
rs912540674 Chr.2: 108469790-108469790 on Build GRCh38
rs765415439 Chr.2: 108469751-108469751 on Build GRCh38
rs35216570 Chr.2: 108469717-108469718 on Build GRCh38
rs201591332 Chr.2: 108469867-108469867 on Build GRCh38
rs747649752 Chr.2: 108469956-108469956 on Build GRCh38
rs781073297 Chr.2: 108469807-108469807 on Build GRCh38
rs773788157 Chr.2: 108469842-108469842 on Build GRCh38
rs775949291 Chr.2: 108469974-108469974 on Build GRCh38
rs753586906 Chr.2: 108470068-108470068 on Build GRCh38
rs994906398 Chr.2: 108470021-108470021 on Build GRCh38
rs745692153 Chr.2: 108469814-108469814 on Build GRCh38
rs763300179 Chr.2: 108469980-108469980 on Build GRCh38
rs774722092 Chr.2: 108470044-108470044 on Build GRCh38
rs973452697 Chr.2: 108469603-108469603 on Build GRCh38
rs746930580 Chr.2: 108469669-108469669 on Build GRCh38
rs753498571 Chr.2: 108469926-108469926 on Build GRCh38
rs367729142 Chr.2: 108469947-108469947 on Build GRCh38
rs771587101 Chr.2: 108469962-108469962 on Build GRCh38
rs371673633 Chr.2: 108469959-108469959 on Build GRCh38
rs758519494 Chr.2: 108469646-108469646 on Build GRCh38
rs181774123 Chr.2: 108469600-108469600 on Build GRCh38
rs769277550 Chr.2: 108469856-108469856 on Build GRCh38
rs750335313 Chr.2: 108470039-108470039 on Build GRCh38
rs761858711 Chr.2: 108469997-108469997 on Build GRCh38
rs375071388 Chr.2: 108470066-108470066 on Build GRCh38
rs1030233979 Chr.2: 108469576-108469576 on Build GRCh38
rs765378096 Chr.2: 108469629-108469629 on Build GRCh38
rs868736677 Chr.2: 108469655-108469655 on Build GRCh38
rs752379605 Chr.2: 108470084-108470084 on Build GRCh38
rs778583148 Chr.2: 108470082-108470082 on Build GRCh38
rs748938912 Chr.2: 108469824-108469824 on Build GRCh38
rs141520578 Chr.2: 108469700-108469700 on Build GRCh38
rs540390097 Chr.2: 108469571-108469571 on Build GRCh38
rs757094933 Chr.2: 108469799-108469799 on Build GRCh38
rs768145104 Chr.2: 108469695-108469695 on Build GRCh38
rs147242602 Chr.2: 108469577-108469577 on Build GRCh38
rs1007534000 Chr.2: 108469582-108469582 on Build GRCh38
rs758253345 Chr.2: 108469970-108469970 on Build GRCh38
rs779742639 Chr.2: 108469817-108469817 on Build GRCh38
rs767406173 Chr.2: 108469899-108469899 on Build GRCh38

View Full Product Details