Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151802497-151802895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5A
Entrez Gene ID
26225
Gene Name
ADP ribosylation factor like GTPase 5A
Gene Aliases
ARFLP5, ARL5
UniGene
Hs.470233
Gene Chromosome Location
Chr.2: 151800966-151828495 on Build GRCh38
Transcripts
NM_001037174.1 NM_012097.3 NM_177985.2

SNP Detail

SNP ID Chromosome Location
rs79886308 Chr.2: 151802579-151802579 on Build GRCh38
rs138368813 Chr.2: 151802838-151802838 on Build GRCh38
rs976316504 Chr.2: 151802833-151802833 on Build GRCh38
rs907033483 Chr.2: 151802721-151802721 on Build GRCh38
rs868670084 Chr.2: 151802746-151802746 on Build GRCh38
rs1047797469 Chr.2: 151802671-151802671 on Build GRCh38
rs751611733 Chr.2: 151802803-151802803 on Build GRCh38
rs547481913 Chr.2: 151802605-151802607 on Build GRCh38
rs923423685 Chr.2: 151802645-151802645 on Build GRCh38
rs894408806 Chr.2: 151802680-151802680 on Build GRCh38
rs547140895 Chr.2: 151802798-151802798 on Build GRCh38
rs942043460 Chr.2: 151802674-151802674 on Build GRCh38
rs957309951 Chr.2: 151802739-151802739 on Build GRCh38
rs570066059 Chr.2: 151802622-151802622 on Build GRCh38
rs532653981 Chr.2: 151802813-151802813 on Build GRCh38
rs1055023795 Chr.2: 151802675-151802675 on Build GRCh38
rs964980986 Chr.2: 151802811-151802811 on Build GRCh38
rs943579786 Chr.2: 151802594-151802594 on Build GRCh38
rs755633320 Chr.2: 151802605-151802605 on Build GRCh38
rs369549359 Chr.2: 151802526-151802526 on Build GRCh38
rs775537632 Chr.2: 151802805-151802806 on Build GRCh38
rs115325184 Chr.2: 151802602-151802602 on Build GRCh38
rs1017254134 Chr.2: 151802804-151802804 on Build GRCh38
rs909313826 Chr.2: 151802848-151802848 on Build GRCh38
rs999045853 Chr.2: 151802722-151802722 on Build GRCh38
rs990190152 Chr.2: 151802542-151802542 on Build GRCh38
rs763972572 Chr.2: 151802710-151802710 on Build GRCh38
rs560381883 Chr.2: 151802734-151802734 on Build GRCh38
rs887823649 Chr.2: 151802673-151802673 on Build GRCh38
rs183322511 Chr.2: 151802564-151802564 on Build GRCh38
rs140218885 Chr.2: 151802854-151802854 on Build GRCh38
rs531718120 Chr.2: 151802844-151802844 on Build GRCh38
rs929407242 Chr.2: 151802670-151802670 on Build GRCh38
rs757301567 Chr.2: 151802862-151802862 on Build GRCh38
rs1012772353 Chr.2: 151802699-151802699 on Build GRCh38
rs188725618 Chr.2: 151802845-151802845 on Build GRCh38
rs77715973 Chr.2: 151802834-151802834 on Build GRCh38

View Full Product Details