Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1388556-1389119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFS7
Entrez Gene ID
374291
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
Gene Aliases
CI-20, CI-20KD, MY017, PSST
UniGene
Hs.211914
Gene Chromosome Location
Chr.19: 1383571-1395589 on Build GRCh38
Transcripts
NM_024407.4 XM_005259556.4 XM_017026768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833631 c.158C>T p.A53V Chr.19: 1388868-1388868 on Build GRCh38
6723828 c.99C>T p.S33S Chr.19: 1388570-1388570 on Build GRCh38
7146921 c.94C>A p.Q32K Chr.19: 1388565-1388565 on Build GRCh38
5343207 c.176G>A p.R59Q Chr.19: 1388886-1388886 on Build GRCh38
3822072 c.158C>T p.A53V Chr.19: 1388868-1388868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370279289 Chr.19: 1388629-1388629 on Build GRCh38
rs770205498 Chr.19: 1388696-1388696 on Build GRCh38
rs764362456 Chr.19: 1388921-1388921 on Build GRCh38
rs750760204 Chr.19: 1389030-1389030 on Build GRCh38
rs759573368 Chr.19: 1388902-1388902 on Build GRCh38
rs375442011 Chr.19: 1388579-1388579 on Build GRCh38
rs533688474 Chr.19: 1388670-1388670 on Build GRCh38
rs989806082 Chr.19: 1388760-1388760 on Build GRCh38
rs1028082230 Chr.19: 1388644-1388644 on Build GRCh38
rs745366485 Chr.19: 1388691-1388692 on Build GRCh38
rs750259721 Chr.19: 1388824-1388824 on Build GRCh38
rs775188712 Chr.19: 1388733-1388733 on Build GRCh38
rs752069239 Chr.19: 1389050-1389050 on Build GRCh38
rs1056050076 Chr.19: 1388775-1388775 on Build GRCh38
rs556060098 Chr.19: 1388759-1388759 on Build GRCh38
rs753548878 Chr.19: 1388756-1388756 on Build GRCh38
rs73515044 Chr.19: 1389046-1389046 on Build GRCh38
rs758876232 Chr.19: 1388622-1388623 on Build GRCh38
rs780556136 Chr.19: 1389048-1389048 on Build GRCh38
rs768221863 Chr.19: 1389036-1389040 on Build GRCh38
rs753037012 Chr.19: 1388588-1388596 on Build GRCh38
rs147710123 Chr.19: 1388848-1388848 on Build GRCh38
rs765488756 Chr.19: 1389029-1389029 on Build GRCh38
rs746339724 Chr.19: 1388941-1388941 on Build GRCh38
rs781512047 Chr.19: 1388675-1388675 on Build GRCh38
rs867795397 Chr.19: 1388623-1388623 on Build GRCh38
rs984936506 Chr.19: 1388834-1388834 on Build GRCh38
rs749786231 Chr.19: 1388793-1388793 on Build GRCh38
rs199959713 Chr.19: 1389077-1389077 on Build GRCh38
rs974885285 Chr.19: 1388708-1388708 on Build GRCh38
rs565395435 Chr.19: 1388868-1388868 on Build GRCh38
rs751213511 Chr.19: 1388761-1388761 on Build GRCh38
rs7258846 Chr.19: 1389065-1389065 on Build GRCh38
rs1029591568 Chr.19: 1388861-1388861 on Build GRCh38
rs775589766 Chr.19: 1389000-1389000 on Build GRCh38
rs763618091 Chr.19: 1388727-1388727 on Build GRCh38
rs779830951 Chr.19: 1388999-1389000 on Build GRCh38
rs766259287 Chr.19: 1388840-1388840 on Build GRCh38
rs745999411 Chr.19: 1389035-1389035 on Build GRCh38
rs969169978 Chr.19: 1388660-1388660 on Build GRCh38
rs764272458 Chr.19: 1389009-1389009 on Build GRCh38
rs771605832 Chr.19: 1388794-1388794 on Build GRCh38
rs748416030 Chr.19: 1388690-1388690 on Build GRCh38
rs771691675 Chr.19: 1388731-1388731 on Build GRCh38
rs141532719 Chr.19: 1389064-1389064 on Build GRCh38
rs774952544 Chr.19: 1388805-1388805 on Build GRCh38
rs763612447 Chr.19: 1388754-1388754 on Build GRCh38
rs776254971 Chr.19: 1388873-1388873 on Build GRCh38
rs771776554 Chr.19: 1389093-1389093 on Build GRCh38
rs765623994 Chr.19: 1388589-1388589 on Build GRCh38
rs376280861 Chr.19: 1388619-1388619 on Build GRCh38
rs11551666 Chr.19: 1388856-1388856 on Build GRCh38
rs774247730 Chr.19: 1388982-1388982 on Build GRCh38
rs752392841 Chr.19: 1388618-1388618 on Build GRCh38
rs778771182 Chr.19: 1388631-1388631 on Build GRCh38
rs943412607 Chr.19: 1388591-1388591 on Build GRCh38
rs758471816 Chr.19: 1388643-1388643 on Build GRCh38
rs762772605 Chr.19: 1388835-1388835 on Build GRCh38
rs773786251 Chr.19: 1388716-1388716 on Build GRCh38
rs201793033 Chr.19: 1388818-1388818 on Build GRCh38
rs748404828 Chr.19: 1388774-1388774 on Build GRCh38
rs762680917 Chr.19: 1388890-1388890 on Build GRCh38
rs377418064 Chr.19: 1388820-1388820 on Build GRCh38
rs981357356 Chr.19: 1388767-1388767 on Build GRCh38
rs781216362 Chr.19: 1388770-1388770 on Build GRCh38
rs770617436 Chr.19: 1388980-1388980 on Build GRCh38
rs565395922 Chr.19: 1388854-1388854 on Build GRCh38
rs544819281 Chr.19: 1388963-1388963 on Build GRCh38
rs761523590 Chr.19: 1389063-1389080 on Build GRCh38
rs780169395 Chr.19: 1388659-1388659 on Build GRCh38
rs538064847 Chr.19: 1388755-1388755 on Build GRCh38
rs766203571 Chr.19: 1388896-1388896 on Build GRCh38
rs532472723 Chr.19: 1388888-1388888 on Build GRCh38
rs551856533 Chr.19: 1389063-1389063 on Build GRCh38
rs763922735 Chr.19: 1388624-1388624 on Build GRCh38
rs562729616 Chr.19: 1388967-1388967 on Build GRCh38
rs757717527 Chr.19: 1388864-1388864 on Build GRCh38
rs375830930 Chr.19: 1388968-1388968 on Build GRCh38
rs768040127 Chr.19: 1388872-1388872 on Build GRCh38
rs619693 Chr.19: 1388813-1388813 on Build GRCh38
rs772876915 Chr.19: 1388885-1388885 on Build GRCh38
rs778337126 Chr.19: 1388790-1388790 on Build GRCh38
rs570415132 Chr.19: 1388724-1388724 on Build GRCh38
rs776145250 Chr.19: 1388815-1388815 on Build GRCh38
rs767448532 Chr.19: 1388984-1388984 on Build GRCh38
rs112045351 Chr.19: 1388599-1388599 on Build GRCh38
rs758379713 Chr.19: 1388762-1388762 on Build GRCh38
rs200080426 Chr.19: 1389045-1389046 on Build GRCh38
rs550324102 Chr.19: 1388605-1388605 on Build GRCh38
rs756293384 Chr.19: 1388917-1388917 on Build GRCh38
rs754125936 Chr.19: 1388925-1388925 on Build GRCh38
rs757603144 Chr.19: 1389019-1389019 on Build GRCh38
rs774148682 Chr.19: 1388899-1388899 on Build GRCh38
rs1002462419 Chr.19: 1389086-1389086 on Build GRCh38
rs916641133 Chr.19: 1388789-1388789 on Build GRCh38
rs745812336 Chr.19: 1388636-1388636 on Build GRCh38
rs754101592 Chr.19: 1389010-1389010 on Build GRCh38
rs772788714 Chr.19: 1388976-1388976 on Build GRCh38
rs745627211 Chr.19: 1389069-1389069 on Build GRCh38
rs759056901 Chr.19: 1388603-1388603 on Build GRCh38
rs767469432 Chr.19: 1388905-1388905 on Build GRCh38
rs751556495 Chr.19: 1388768-1388768 on Build GRCh38
rs112547983 Chr.19: 1389007-1389007 on Build GRCh38
rs757956854 Chr.19: 1388630-1388630 on Build GRCh38
rs770275334 Chr.19: 1388778-1388778 on Build GRCh38
rs558627153 Chr.19: 1388722-1388722 on Build GRCh38
rs752857982 Chr.19: 1388908-1388908 on Build GRCh38
rs757627492 Chr.19: 1388933-1388933 on Build GRCh38
rs368271480 Chr.19: 1388971-1388971 on Build GRCh38
rs142519186 Chr.19: 1388855-1388855 on Build GRCh38
rs779228151 Chr.19: 1388866-1388866 on Build GRCh38
rs750011146 Chr.19: 1388602-1388602 on Build GRCh38
rs1030902336 Chr.19: 1388625-1388625 on Build GRCh38
rs776909226 Chr.19: 1388580-1388580 on Build GRCh38
rs769467649 Chr.19: 1388955-1388956 on Build GRCh38
rs779394590 Chr.19: 1388936-1388936 on Build GRCh38
rs778480411 Chr.19: 1388692-1388692 on Build GRCh38
rs760343474 Chr.19: 1388749-1388749 on Build GRCh38
rs958883478 Chr.19: 1388920-1388920 on Build GRCh38
rs530310236 Chr.19: 1388616-1388616 on Build GRCh38
rs749010384 Chr.19: 1388979-1388979 on Build GRCh38
rs758984505 Chr.19: 1388956-1388956 on Build GRCh38
rs779890539 Chr.19: 1388686-1388686 on Build GRCh38
rs778251548 Chr.19: 1388860-1388860 on Build GRCh38
rs754948521 Chr.19: 1388849-1388849 on Build GRCh38
rs761450030 Chr.19: 1388758-1388758 on Build GRCh38
rs748918090 Chr.19: 1389004-1389007 on Build GRCh38
rs762352035 Chr.19: 1388584-1388584 on Build GRCh38
rs781397518 Chr.19: 1388851-1388851 on Build GRCh38
rs889218427 Chr.19: 1388587-1388587 on Build GRCh38
rs143192592 Chr.19: 1389005-1389005 on Build GRCh38
rs879550646 Chr.19: 1389076-1389076 on Build GRCh38
rs886657046 Chr.19: 1388654-1388654 on Build GRCh38
rs140236960 Chr.19: 1388863-1388863 on Build GRCh38
rs574820199 Chr.19: 1388766-1388766 on Build GRCh38
rs960390737 Chr.19: 1388679-1388679 on Build GRCh38
rs747232341 Chr.19: 1388669-1388669 on Build GRCh38
rs755619725 Chr.19: 1389059-1389059 on Build GRCh38
rs747767440 Chr.19: 1388877-1388877 on Build GRCh38
rs755110912 Chr.19: 1388769-1388769 on Build GRCh38
rs934714296 Chr.19: 1389032-1389033 on Build GRCh38
rs370831422 Chr.19: 1388878-1388878 on Build GRCh38
rs113461980 Chr.19: 1388940-1388940 on Build GRCh38

View Full Product Details