Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 62368436-62368982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF11A
Entrez Gene ID
8792
Gene Name
TNF receptor superfamily member 11a
Gene Aliases
CD265, FEO, LOH18CR1, ODFR, OFE, OPTB7, OSTS, PDB2, RANK, TRANCER
UniGene
Hs.204044
Gene Chromosome Location
Chr.18: 62325287-62388096 on Build GRCh38
Transcripts
NM_001270949.1 NM_001270950.1 NM_001270951.1 NM_001278268.1 NM_003839.3 XM_011526244.2 XM_011526245.2 XM_017026064.1 XM_017026065.1 XM_017026066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6557091 c.830G>T p.G277V Chr.18: 62368747-62368747 on Build GRCh38
7318197 c.868C>T p.L290L Chr.18: 62368785-62368785 on Build GRCh38
6804976 c.997_998insC p.E334fs*15 Chr.18: 62368914-62368915 on Build GRCh38
303420 c.964G>A p.E322K Chr.18: 62368881-62368881 on Build GRCh38
6390090 c.869T>C p.L290P Chr.18: 62368786-62368786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182849554 Chr.18: 62368902-62368902 on Build GRCh38
rs200952751 Chr.18: 62368871-62368871 on Build GRCh38
rs553992512 Chr.18: 62368736-62368736 on Build GRCh38
rs765856121 Chr.18: 62368716-62368716 on Build GRCh38
rs745367315 Chr.18: 62368737-62368737 on Build GRCh38
rs749292753 Chr.18: 62368881-62368881 on Build GRCh38
rs779851655 Chr.18: 62368775-62368775 on Build GRCh38
rs1040260233 Chr.18: 62368469-62368469 on Build GRCh38
rs758054643 Chr.18: 62368477-62368477 on Build GRCh38
rs375650076 Chr.18: 62368888-62368888 on Build GRCh38
rs112149607 Chr.18: 62368702-62368702 on Build GRCh38
rs759786718 Chr.18: 62368917-62368917 on Build GRCh38
rs1012179260 Chr.18: 62368653-62368653 on Build GRCh38
rs138204772 Chr.18: 62368916-62368916 on Build GRCh38
rs756426313 Chr.18: 62368498-62368499 on Build GRCh38
rs773991517 Chr.18: 62368795-62368795 on Build GRCh38
rs369507616 Chr.18: 62368695-62368695 on Build GRCh38
rs769235050 Chr.18: 62368751-62368751 on Build GRCh38
rs749136654 Chr.18: 62368783-62368783 on Build GRCh38
rs747314306 Chr.18: 62368797-62368797 on Build GRCh38
rs868606619 Chr.18: 62368926-62368926 on Build GRCh38
rs540191294 Chr.18: 62368897-62368897 on Build GRCh38
rs780982821 Chr.18: 62368944-62368944 on Build GRCh38
rs768443075 Chr.18: 62368883-62368883 on Build GRCh38
rs866828586 Chr.18: 62368858-62368858 on Build GRCh38
rs765372255 Chr.18: 62368821-62368821 on Build GRCh38
rs562689512 Chr.18: 62368860-62368860 on Build GRCh38
rs773511368 Chr.18: 62368660-62368660 on Build GRCh38
rs764498229 Chr.18: 62368935-62368935 on Build GRCh38
rs780834651 Chr.18: 62368870-62368870 on Build GRCh38
rs775402989 Chr.18: 62368921-62368921 on Build GRCh38
rs141296714 Chr.18: 62368585-62368585 on Build GRCh38
rs1025726517 Chr.18: 62368793-62368793 on Build GRCh38
rs762168916 Chr.18: 62368949-62368949 on Build GRCh38
rs143978208 Chr.18: 62368499-62368502 on Build GRCh38
rs766698670 Chr.18: 62368801-62368801 on Build GRCh38
rs935112085 Chr.18: 62368894-62368894 on Build GRCh38
rs1051470553 Chr.18: 62368520-62368520 on Build GRCh38
rs984101155 Chr.18: 62368817-62368817 on Build GRCh38
rs201372520 Chr.18: 62368632-62368632 on Build GRCh38
rs539985099 Chr.18: 62368613-62368613 on Build GRCh38
rs572449155 Chr.18: 62368696-62368696 on Build GRCh38
rs777951423 Chr.18: 62368730-62368730 on Build GRCh38
rs755652460 Chr.18: 62368879-62368879 on Build GRCh38
rs555319249 Chr.18: 62368685-62368685 on Build GRCh38
rs758825585 Chr.18: 62368722-62368722 on Build GRCh38
rs1042572034 Chr.18: 62368614-62368614 on Build GRCh38
rs975707224 Chr.18: 62368869-62368869 on Build GRCh38
rs189818158 Chr.18: 62368648-62368648 on Build GRCh38
rs556358673 Chr.18: 62368634-62368634 on Build GRCh38
rs370984926 Chr.18: 62368813-62368813 on Build GRCh38
rs970590296 Chr.18: 62368774-62368774 on Build GRCh38
rs8092336 Chr.18: 62368850-62368850 on Build GRCh38
rs199871080 Chr.18: 62368709-62368709 on Build GRCh38
rs933046033 Chr.18: 62368502-62368502 on Build GRCh38
rs747910982 Chr.18: 62368891-62368891 on Build GRCh38
rs752044497 Chr.18: 62368942-62368942 on Build GRCh38
rs753270663 Chr.18: 62368836-62368836 on Build GRCh38
rs12721431 Chr.18: 62368767-62368767 on Build GRCh38
rs902306940 Chr.18: 62368541-62368541 on Build GRCh38
rs756829516 Chr.18: 62368735-62368735 on Build GRCh38
rs376997742 Chr.18: 62368726-62368726 on Build GRCh38
rs764561352 Chr.18: 62368849-62368849 on Build GRCh38
rs760995497 Chr.18: 62368800-62368800 on Build GRCh38
rs779486519 Chr.18: 62368880-62368880 on Build GRCh38
rs750094340 Chr.18: 62368958-62368958 on Build GRCh38
rs934154488 Chr.18: 62368872-62368872 on Build GRCh38
rs372651523 Chr.18: 62368650-62368650 on Build GRCh38
rs780503624 Chr.18: 62368769-62368769 on Build GRCh38
rs141157565 Chr.18: 62368720-62368720 on Build GRCh38
rs770043265 Chr.18: 62368919-62368919 on Build GRCh38
rs914337428 Chr.18: 62368582-62368582 on Build GRCh38
rs775410414 Chr.18: 62368770-62368770 on Build GRCh38
rs763347757 Chr.18: 62368840-62368840 on Build GRCh38
rs768515742 Chr.18: 62368787-62368787 on Build GRCh38
rs766612724 Chr.18: 62368694-62368694 on Build GRCh38
rs746724864 Chr.18: 62368733-62368733 on Build GRCh38
rs548117602 Chr.18: 62368533-62368533 on Build GRCh38
rs577066104 Chr.18: 62368819-62368819 on Build GRCh38
rs9675625 Chr.18: 62368531-62368531 on Build GRCh38
rs763586080 Chr.18: 62368934-62368934 on Build GRCh38
rs143775083 Chr.18: 62368954-62368954 on Build GRCh38

View Full Product Details