Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70925457-70926056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A3
Entrez Gene ID
84889
Gene Name
solute carrier family 7 member 3
Gene Aliases
ATRC3, CAT-3, CAT3
UniGene
Hs.175220
Gene Chromosome Location
Chr.X: 70925579-70931125 on Build GRCh38
Transcripts
NM_001048164.2 NM_032803.5 XM_017029912.1

SNP Detail

SNP ID Chromosome Location
rs759970627 Chr.X: 70925883-70925883 on Build GRCh38
rs374374371 Chr.X: 70925840-70925840 on Build GRCh38
rs773048078 Chr.X: 70925827-70925827 on Build GRCh38
rs750168357 Chr.X: 70925957-70925957 on Build GRCh38
rs979758268 Chr.X: 70925740-70925740 on Build GRCh38
rs183949880 Chr.X: 70926004-70926004 on Build GRCh38
rs193167949 Chr.X: 70925735-70925735 on Build GRCh38
rs747729341 Chr.X: 70925819-70925819 on Build GRCh38
rs769062447 Chr.X: 70925978-70925978 on Build GRCh38
rs138954830 Chr.X: 70925875-70925875 on Build GRCh38
rs1014829750 Chr.X: 70925499-70925499 on Build GRCh38
rs770139501 Chr.X: 70925803-70925803 on Build GRCh38
rs1040548838 Chr.X: 70925895-70925895 on Build GRCh38
rs149447856 Chr.X: 70925907-70925907 on Build GRCh38
rs751055612 Chr.X: 70925695-70925695 on Build GRCh38
rs978103355 Chr.X: 70925809-70925809 on Build GRCh38
rs771753027 Chr.X: 70925983-70925983 on Build GRCh38
rs1031455781 Chr.X: 70925663-70925663 on Build GRCh38
rs968094456 Chr.X: 70925645-70925645 on Build GRCh38
rs199657967 Chr.X: 70925995-70925995 on Build GRCh38
rs185505099 Chr.X: 70925889-70925889 on Build GRCh38
rs188253454 Chr.X: 70925712-70925712 on Build GRCh38
rs1040998227 Chr.X: 70925510-70925510 on Build GRCh38
rs771695858 Chr.X: 70925824-70925824 on Build GRCh38
rs756498269 Chr.X: 70925967-70925967 on Build GRCh38
rs756840747 Chr.X: 70925724-70925724 on Build GRCh38
rs370603171 Chr.X: 70925796-70925796 on Build GRCh38
rs766232079 Chr.X: 70925854-70925854 on Build GRCh38
rs746685423 Chr.X: 70926031-70926031 on Build GRCh38
rs999603234 Chr.X: 70925650-70925650 on Build GRCh38
rs996539962 Chr.X: 70925571-70925571 on Build GRCh38
rs953114811 Chr.X: 70925730-70925730 on Build GRCh38
rs965730509 Chr.X: 70925795-70925795 on Build GRCh38
rs191422299 Chr.X: 70925912-70925912 on Build GRCh38
rs749790512 Chr.X: 70925982-70925982 on Build GRCh38
rs201510794 Chr.X: 70925927-70925927 on Build GRCh38
rs1027458958 Chr.X: 70925716-70925716 on Build GRCh38
rs746407923 Chr.X: 70925764-70925764 on Build GRCh38
rs147665669 Chr.X: 70925874-70925874 on Build GRCh38
rs924886149 Chr.X: 70925829-70925829 on Build GRCh38
rs776243764 Chr.X: 70925855-70925855 on Build GRCh38
rs961895601 Chr.X: 70925679-70925679 on Build GRCh38
rs1022258929 Chr.X: 70925632-70925632 on Build GRCh38
rs373957093 Chr.X: 70925808-70925808 on Build GRCh38
rs930362776 Chr.X: 70925826-70925826 on Build GRCh38
rs911584325 Chr.X: 70925767-70925767 on Build GRCh38
rs900771355 Chr.X: 70925547-70925547 on Build GRCh38
rs944846341 Chr.X: 70925882-70925882 on Build GRCh38
rs368524198 Chr.X: 70925885-70925885 on Build GRCh38
rs766507451 Chr.X: 70925772-70925772 on Build GRCh38
rs777382174 Chr.X: 70926023-70926023 on Build GRCh38
rs371854122 Chr.X: 70925835-70925835 on Build GRCh38

View Full Product Details