Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139367137-139367657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAIP2
Entrez Gene ID
51247
Gene Name
poly(A) binding protein interacting protein 2
Gene Aliases
PAIP-2, PAIP2A
UniGene
Hs.396644
Gene Chromosome Location
Chr.5: 139341832-139369720 on Build GRCh38
Transcripts
NM_001033112.2 NM_016480.4 NR_109830.1 XM_011543428.1 XM_017009522.1 XM_017009523.1 NM_005847.4 NM_152685.3 XM_005272148.3 XM_005272149.3 XM_006714741.2 XM_011543766.1 XM_011543767.1 XM_011543768.1 XM_011543769.1

Gene Symbol
SLC23A1
Entrez Gene ID
9963
Gene Name
solute carrier family 23 member 1
Gene Aliases
SLC23A2, SVCT1, YSPL3
UniGene
Hs.643467
Gene Chromosome Location
Chr.5: 139367196-139384553 on Build GRCh38
Transcripts
NM_001033112.2 NM_016480.4 NR_109830.1 XM_011543428.1 XM_017009522.1 XM_017009523.1 NM_005847.4 NM_152685.3 XM_005272148.3 XM_005272149.3 XM_006714741.2 XM_011543766.1 XM_011543767.1 XM_011543768.1 XM_011543769.1

SNP Detail

SNP ID Chromosome Location
rs1030257154 Chr.5: 139367322-139367322 on Build GRCh38
rs144815785 Chr.5: 139367228-139367228 on Build GRCh38
rs759570577 Chr.5: 139367584-139367584 on Build GRCh38
rs893055632 Chr.5: 139367185-139367185 on Build GRCh38
rs947005087 Chr.5: 139367625-139367625 on Build GRCh38
rs749693799 Chr.5: 139367432-139367432 on Build GRCh38
rs955478440 Chr.5: 139367481-139367481 on Build GRCh38
rs374941537 Chr.5: 139367455-139367455 on Build GRCh38
rs72552249 Chr.5: 139367230-139367230 on Build GRCh38
rs767497925 Chr.5: 139367613-139367613 on Build GRCh38
rs982368762 Chr.5: 139367471-139367471 on Build GRCh38
rs772413815 Chr.5: 139367252-139367252 on Build GRCh38
rs955583324 Chr.5: 139367285-139367285 on Build GRCh38
rs73255268 Chr.5: 139367327-139367327 on Build GRCh38
rs539845141 Chr.5: 139367568-139367568 on Build GRCh38
rs773445564 Chr.5: 139367358-139367358 on Build GRCh38
rs1038943593 Chr.5: 139367472-139367472 on Build GRCh38
rs998062197 Chr.5: 139367273-139367273 on Build GRCh38
rs928407375 Chr.5: 139367545-139367546 on Build GRCh38
rs548009946 Chr.5: 139367505-139367505 on Build GRCh38
rs899685755 Chr.5: 139367530-139367530 on Build GRCh38
rs537959586 Chr.5: 139367531-139367532 on Build GRCh38
rs1030202224 Chr.5: 139367480-139367480 on Build GRCh38
rs568411721 Chr.5: 139367437-139367437 on Build GRCh38
rs116682228 Chr.5: 139367470-139367470 on Build GRCh38
rs865815081 Chr.5: 139367543-139367543 on Build GRCh38
rs566648751 Chr.5: 139367422-139367422 on Build GRCh38
rs892771599 Chr.5: 139367469-139367469 on Build GRCh38
rs534871649 Chr.5: 139367397-139367397 on Build GRCh38
rs1015552339 Chr.5: 139367493-139367493 on Build GRCh38
rs916788277 Chr.5: 139367454-139367454 on Build GRCh38
rs916083611 Chr.5: 139367390-139367390 on Build GRCh38
rs914967369 Chr.5: 139367498-139367498 on Build GRCh38
rs1004220328 Chr.5: 139367481-139367481 on Build GRCh38
rs1052762459 Chr.5: 139367468-139367469 on Build GRCh38
rs1011448491 Chr.5: 139367186-139367186 on Build GRCh38
rs962663617 Chr.5: 139367496-139367497 on Build GRCh38
rs916151931 Chr.5: 139367526-139367526 on Build GRCh38
rs928385565 Chr.5: 139367434-139367434 on Build GRCh38
rs1017455490 Chr.5: 139367214-139367214 on Build GRCh38
rs1039903937 Chr.5: 139367513-139367513 on Build GRCh38
rs770393589 Chr.5: 139367338-139367338 on Build GRCh38
rs1038995898 Chr.5: 139367631-139367631 on Build GRCh38
rs1052131094 Chr.5: 139367497-139367497 on Build GRCh38
rs927584622 Chr.5: 139367475-139367475 on Build GRCh38
rs74704674 Chr.5: 139367386-139367386 on Build GRCh38
rs963521075 Chr.5: 139367418-139367418 on Build GRCh38
rs1029715623 Chr.5: 139367254-139367254 on Build GRCh38
rs557642206 Chr.5: 139367512-139367512 on Build GRCh38
rs948887006 Chr.5: 139367395-139367395 on Build GRCh38
rs546815077 Chr.5: 139367426-139367426 on Build GRCh38
rs1004948185 Chr.5: 139367378-139367378 on Build GRCh38
rs66721307 Chr.5: 139367576-139367576 on Build GRCh38
rs141928669 Chr.5: 139367446-139367446 on Build GRCh38
rs532952723 Chr.5: 139367283-139367283 on Build GRCh38
rs188304462 Chr.5: 139367563-139367563 on Build GRCh38
rs962941794 Chr.5: 139367362-139367362 on Build GRCh38
rs371452181 Chr.5: 139367452-139367452 on Build GRCh38
rs935009684 Chr.5: 139367489-139367489 on Build GRCh38

View Full Product Details