Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30448199-30448691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOD1
Entrez Gene ID
10392
Gene Name
nucleotide binding oligomerization domain containing 1
Gene Aliases
CARD4, CLR7.1, NLRC1
UniGene
Hs.405153
Gene Chromosome Location
Chr.7: 30424527-30478840 on Build GRCh38
Transcripts
NM_006092.2 XM_005249568.1 XM_005249572.1 XM_005249576.1 XM_006715633.2 XM_011515079.1 XM_011515080.2 XM_011515081.2 XM_011515083.1 XM_011515084.1 XM_011515085.1 XM_011515087.1 XM_011515088.2 XM_017011674.1 XM_017011675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1259274 c.2202-1G>C p.? Chr.7: 30448382-30448382 on Build GRCh38
78182 c.2237A>C p.K746T Chr.7: 30448346-30448346 on Build GRCh38
6727596 c.2208C>T p.S736S Chr.7: 30448375-30448375 on Build GRCh38
5629182 c.2271T>A p.I757I Chr.7: 30448312-30448312 on Build GRCh38
1259273 c.2256G>T p.L752L Chr.7: 30448327-30448327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770292624 Chr.7: 30448262-30448303 on Build GRCh38
rs977524590 Chr.7: 30448282-30448282 on Build GRCh38
rs375665569 Chr.7: 30448430-30448430 on Build GRCh38
rs967106860 Chr.7: 30448556-30448556 on Build GRCh38
rs987167845 Chr.7: 30448271-30448271 on Build GRCh38
rs957613123 Chr.7: 30448624-30448624 on Build GRCh38
rs749111479 Chr.7: 30448369-30448369 on Build GRCh38
rs553752426 Chr.7: 30448465-30448465 on Build GRCh38
rs147319394 Chr.7: 30448335-30448335 on Build GRCh38
rs147294540 Chr.7: 30448356-30448356 on Build GRCh38
rs5743349 Chr.7: 30448607-30448607 on Build GRCh38
rs199476266 Chr.7: 30448267-30448267 on Build GRCh38
rs916208167 Chr.7: 30448659-30448659 on Build GRCh38
rs189249798 Chr.7: 30448278-30448278 on Build GRCh38
rs190839337 Chr.7: 30448521-30448521 on Build GRCh38
rs139288894 Chr.7: 30448410-30448410 on Build GRCh38
rs377256233 Chr.7: 30448415-30448415 on Build GRCh38
rs376229776 Chr.7: 30448286-30448286 on Build GRCh38
rs545419778 Chr.7: 30448316-30448316 on Build GRCh38
rs767995510 Chr.7: 30448431-30448431 on Build GRCh38
rs748057160 Chr.7: 30448386-30448386 on Build GRCh38
rs5743351 Chr.7: 30448416-30448416 on Build GRCh38
rs973306714 Chr.7: 30448596-30448596 on Build GRCh38
rs369269480 Chr.7: 30448287-30448287 on Build GRCh38
rs995820269 Chr.7: 30448498-30448498 on Build GRCh38
rs761422072 Chr.7: 30448294-30448294 on Build GRCh38
rs537172994 Chr.7: 30448375-30448375 on Build GRCh38
rs753174542 Chr.7: 30448326-30448326 on Build GRCh38
rs751894924 Chr.7: 30448429-30448429 on Build GRCh38
rs763163398 Chr.7: 30448331-30448331 on Build GRCh38
rs1028895887 Chr.7: 30448495-30448495 on Build GRCh38
rs559250887 Chr.7: 30448621-30448621 on Build GRCh38
rs1029261595 Chr.7: 30448225-30448225 on Build GRCh38
rs772605825 Chr.7: 30448463-30448463 on Build GRCh38
rs199476265 Chr.7: 30448408-30448408 on Build GRCh38
rs551226017 Chr.7: 30448628-30448628 on Build GRCh38
rs565321339 Chr.7: 30448554-30448554 on Build GRCh38
rs928758046 Chr.7: 30448647-30448647 on Build GRCh38
rs180712416 Chr.7: 30448336-30448336 on Build GRCh38
rs776092228 Chr.7: 30448413-30448413 on Build GRCh38
rs757271582 Chr.7: 30448252-30448252 on Build GRCh38
rs143956406 Chr.7: 30448345-30448345 on Build GRCh38
rs771567280 Chr.7: 30448374-30448374 on Build GRCh38
rs528524681 Chr.7: 30448606-30448606 on Build GRCh38
rs764178949 Chr.7: 30448661-30448661 on Build GRCh38
rs563890272 Chr.7: 30448539-30448539 on Build GRCh38
rs372998851 Chr.7: 30448277-30448277 on Build GRCh38
rs772915247 Chr.7: 30448404-30448404 on Build GRCh38
rs1038798970 Chr.7: 30448307-30448307 on Build GRCh38
rs561181917 Chr.7: 30448562-30448562 on Build GRCh38
rs34750559 Chr.7: 30448426-30448427 on Build GRCh38
rs759166672 Chr.7: 30448397-30448397 on Build GRCh38
rs770143611 Chr.7: 30448229-30448229 on Build GRCh38
rs953196334 Chr.7: 30448499-30448499 on Build GRCh38
rs76568682 Chr.7: 30448618-30448618 on Build GRCh38
rs543108293 Chr.7: 30448561-30448561 on Build GRCh38
rs926190068 Chr.7: 30448292-30448292 on Build GRCh38
rs140015596 Chr.7: 30448357-30448357 on Build GRCh38
rs575719156 Chr.7: 30448553-30448553 on Build GRCh38
rs780131661 Chr.7: 30448366-30448366 on Build GRCh38
rs749021114 Chr.7: 30448279-30448279 on Build GRCh38
rs963652979 Chr.7: 30448559-30448559 on Build GRCh38
rs772113099 Chr.7: 30448393-30448393 on Build GRCh38
rs113419138 Chr.7: 30448640-30448640 on Build GRCh38
rs539294978 Chr.7: 30448325-30448325 on Build GRCh38
rs781286859 Chr.7: 30448344-30448344 on Build GRCh38
rs907815347 Chr.7: 30448246-30448246 on Build GRCh38
rs772818623 Chr.7: 30448318-30448318 on Build GRCh38
rs759965860 Chr.7: 30448438-30448438 on Build GRCh38
rs5743350 Chr.7: 30448492-30448492 on Build GRCh38
rs775571589 Chr.7: 30448475-30448475 on Build GRCh38
rs1008723314 Chr.7: 30448538-30448538 on Build GRCh38
rs533040940 Chr.7: 30448623-30448623 on Build GRCh38
rs767086231 Chr.7: 30448301-30448301 on Build GRCh38
rs776197726 Chr.7: 30448396-30448397 on Build GRCh38
rs950191416 Chr.7: 30448667-30448667 on Build GRCh38
rs746054225 Chr.7: 30448269-30448269 on Build GRCh38

View Full Product Details