Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29146177-29146764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHN2
Entrez Gene ID
1124
Gene Name
chimerin 2
Gene Aliases
ARHGAP3, BCH, CHN2-3, RHOGAP3
UniGene
Hs.654611
Gene Chromosome Location
Chr.7: 29146547-29514335 on Build GRCh38
Transcripts
NM_001293069.1 XM_011515105.2 XM_011515106.2 XM_017011721.1 NM_019029.2 NM_031311.3 XM_011515437.1 XM_017012366.1

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
UniGene
Hs.233389
Gene Chromosome Location
Chr.7: 28995630-29146537 on Build GRCh38
Transcripts
NM_001293069.1 XM_011515105.2 XM_011515106.2 XM_017011721.1 NM_019029.2 NM_031311.3 XM_011515437.1 XM_017012366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4162131 c.153T>G p.G51G Chr.7: 29146736-29146736 on Build GRCh38
6039025 c.4A>G p.T2A Chr.7: 29146587-29146587 on Build GRCh38
6201982 c.155C>T p.A52V Chr.7: 29146738-29146738 on Build GRCh38
1635267 c.36G>C p.S12S Chr.7: 29146619-29146619 on Build GRCh38
7425265 c.62C>T p.T21I Chr.7: 29146645-29146645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904080620 Chr.7: 29146222-29146222 on Build GRCh38
rs954216404 Chr.7: 29146681-29146681 on Build GRCh38
rs992377619 Chr.7: 29146547-29146547 on Build GRCh38
rs974158962 Chr.7: 29146653-29146653 on Build GRCh38
rs891279724 Chr.7: 29146285-29146286 on Build GRCh38
rs974170532 Chr.7: 29146439-29146439 on Build GRCh38
rs149094510 Chr.7: 29146389-29146389 on Build GRCh38
rs568736513 Chr.7: 29146631-29146631 on Build GRCh38
rs753834181 Chr.7: 29146296-29146296 on Build GRCh38
rs1040072572 Chr.7: 29146556-29146556 on Build GRCh38
rs1012870082 Chr.7: 29146408-29146408 on Build GRCh38
rs762806698 Chr.7: 29146673-29146673 on Build GRCh38
rs932919558 Chr.7: 29146328-29146328 on Build GRCh38
rs1027551012 Chr.7: 29146669-29146669 on Build GRCh38
rs571012210 Chr.7: 29146452-29146452 on Build GRCh38
rs576660849 Chr.7: 29146340-29146340 on Build GRCh38
rs760486853 Chr.7: 29146522-29146522 on Build GRCh38
rs998656166 Chr.7: 29146391-29146391 on Build GRCh38
rs570153400 Chr.7: 29146428-29146428 on Build GRCh38
rs1038514309 Chr.7: 29146309-29146309 on Build GRCh38
rs555617352 Chr.7: 29146261-29146261 on Build GRCh38
rs887442623 Chr.7: 29146503-29146503 on Build GRCh38
rs887473568 Chr.7: 29146680-29146680 on Build GRCh38
rs897541260 Chr.7: 29146442-29146442 on Build GRCh38
rs992284971 Chr.7: 29146382-29146382 on Build GRCh38
rs558382517 Chr.7: 29146629-29146629 on Build GRCh38
rs757908730 Chr.7: 29146620-29146620 on Build GRCh38
rs750725701 Chr.7: 29146436-29146436 on Build GRCh38
rs575702666 Chr.7: 29146298-29146299 on Build GRCh38
rs1008634280 Chr.7: 29146385-29146385 on Build GRCh38
rs113631333 Chr.7: 29146326-29146326 on Build GRCh38
rs111863920 Chr.7: 29146238-29146238 on Build GRCh38
rs141813055 Chr.7: 29146254-29146255 on Build GRCh38
rs866557417 Chr.7: 29146450-29146450 on Build GRCh38
rs912826388 Chr.7: 29146548-29146548 on Build GRCh38
rs944326629 Chr.7: 29146552-29146552 on Build GRCh38
rs985757203 Chr.7: 29146605-29146605 on Build GRCh38
rs960584063 Chr.7: 29146538-29146538 on Build GRCh38
rs1027532074 Chr.7: 29146315-29146315 on Build GRCh38
rs1050035127 Chr.7: 29146490-29146490 on Build GRCh38
rs763980926 Chr.7: 29146736-29146736 on Build GRCh38
rs1022857906 Chr.7: 29146612-29146612 on Build GRCh38
rs112503596 Chr.7: 29146242-29146242 on Build GRCh38
rs187292682 Chr.7: 29146601-29146601 on Build GRCh38
rs971444692 Chr.7: 29146416-29146416 on Build GRCh38
rs979386986 Chr.7: 29146410-29146410 on Build GRCh38
rs1034001276 Chr.7: 29146533-29146533 on Build GRCh38
rs957514131 Chr.7: 29146404-29146404 on Build GRCh38
rs2687633 Chr.7: 29146558-29146558 on Build GRCh38
rs971298662 Chr.7: 29146621-29146621 on Build GRCh38
rs552029947 Chr.7: 29146443-29146443 on Build GRCh38
rs245880 Chr.7: 29146640-29146640 on Build GRCh38
rs527380851 Chr.7: 29146419-29146419 on Build GRCh38
rs544078140 Chr.7: 29146346-29146346 on Build GRCh38
rs1055068376 Chr.7: 29146734-29146734 on Build GRCh38
rs986047830 Chr.7: 29146682-29146682 on Build GRCh38
rs540081749 Chr.7: 29146386-29146386 on Build GRCh38
rs888819156 Chr.7: 29146381-29146381 on Build GRCh38
rs1054687170 Chr.7: 29146584-29146584 on Build GRCh38
rs1050508138 Chr.7: 29146344-29146344 on Build GRCh38
rs998646172 Chr.7: 29146564-29146564 on Build GRCh38
rs549958548 Chr.7: 29146415-29146415 on Build GRCh38
rs960804154 Chr.7: 29146371-29146371 on Build GRCh38
rs76609459 Chr.7: 29146294-29146294 on Build GRCh38
rs759332913 Chr.7: 29146532-29146532 on Build GRCh38
rs559892986 Chr.7: 29146418-29146418 on Build GRCh38
rs991859507 Chr.7: 29146211-29146211 on Build GRCh38
rs1002151010 Chr.7: 29146531-29146531 on Build GRCh38
rs200559057 Chr.7: 29146256-29146257 on Build GRCh38
rs892997339 Chr.7: 29146589-29146589 on Build GRCh38
rs925531811 Chr.7: 29146414-29146414 on Build GRCh38
rs554088638 Chr.7: 29146729-29146729 on Build GRCh38
rs1037574798 Chr.7: 29146627-29146627 on Build GRCh38
rs1050545559 Chr.7: 29146420-29146420 on Build GRCh38
rs116302767 Chr.7: 29146402-29146402 on Build GRCh38
rs770946319 Chr.7: 29146599-29146599 on Build GRCh38
rs908603612 Chr.7: 29146430-29146430 on Build GRCh38
rs535813152 Chr.7: 29146485-29146485 on Build GRCh38
rs769641932 Chr.7: 29146635-29146635 on Build GRCh38
rs957317508 Chr.7: 29146202-29146202 on Build GRCh38
rs764807330 Chr.7: 29146555-29146558 on Build GRCh38
rs891316393 Chr.7: 29146427-29146427 on Build GRCh38

View Full Product Details