Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160677065-160677648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL14
Entrez Gene ID
80117
Gene Name
ADP ribosylation factor like GTPase 14
Gene Aliases
ARF7
UniGene
Hs.287702
Gene Chromosome Location
Chr.3: 160677160-160678448 on Build GRCh38
Transcripts
NM_025047.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183603 c.181A>G p.T61A Chr.3: 160677527-160677527 on Build GRCh38
4449335 c.206delA p.M71fs*1 Chr.3: 160677552-160677552 on Build GRCh38
1040648 c.277G>T p.D93Y Chr.3: 160677623-160677623 on Build GRCh38
4429933 c.27G>A p.P9P Chr.3: 160677373-160677373 on Build GRCh38
6988496 c.202C>T p.Q68* Chr.3: 160677548-160677548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913306835 Chr.3: 160677538-160677538 on Build GRCh38
rs780494931 Chr.3: 160677573-160677573 on Build GRCh38
rs139181474 Chr.3: 160677592-160677592 on Build GRCh38
rs752720930 Chr.3: 160677440-160677440 on Build GRCh38
rs1879797 Chr.3: 160677305-160677305 on Build GRCh38
rs573150490 Chr.3: 160677503-160677503 on Build GRCh38
rs548220537 Chr.3: 160677356-160677356 on Build GRCh38
rs772368827 Chr.3: 160677309-160677310 on Build GRCh38
rs996440684 Chr.3: 160677457-160677457 on Build GRCh38
rs781128689 Chr.3: 160677386-160677386 on Build GRCh38
rs146866643 Chr.3: 160677373-160677373 on Build GRCh38
rs780602593 Chr.3: 160677326-160677326 on Build GRCh38
rs533454381 Chr.3: 160677498-160677498 on Build GRCh38
rs146619115 Chr.3: 160677419-160677419 on Build GRCh38
rs962518983 Chr.3: 160677170-160677170 on Build GRCh38
rs147660859 Chr.3: 160677313-160677313 on Build GRCh38
rs774047450 Chr.3: 160677424-160677424 on Build GRCh38
rs766233582 Chr.3: 160677620-160677623 on Build GRCh38
rs796098392 Chr.3: 160677302-160677302 on Build GRCh38
rs768593688 Chr.3: 160677315-160677315 on Build GRCh38
rs748739938 Chr.3: 160677465-160677465 on Build GRCh38
rs796186575 Chr.3: 160677302-160677303 on Build GRCh38
rs115969922 Chr.3: 160677281-160677281 on Build GRCh38
rs751320155 Chr.3: 160677375-160677375 on Build GRCh38
rs116525449 Chr.3: 160677154-160677154 on Build GRCh38
rs763260093 Chr.3: 160677595-160677595 on Build GRCh38
rs757755203 Chr.3: 160677305-160677306 on Build GRCh38
rs543602720 Chr.3: 160677307-160677308 on Build GRCh38
rs368653565 Chr.3: 160677625-160677625 on Build GRCh38
rs1003933696 Chr.3: 160677129-160677129 on Build GRCh38
rs781709360 Chr.3: 160677586-160677586 on Build GRCh38
rs374528704 Chr.3: 160677337-160677337 on Build GRCh38
rs760666218 Chr.3: 160677561-160677561 on Build GRCh38
rs1013296642 Chr.3: 160677165-160677165 on Build GRCh38
rs376680744 Chr.3: 160677340-160677340 on Build GRCh38
rs767213992 Chr.3: 160677438-160677438 on Build GRCh38
rs1033510945 Chr.3: 160677141-160677141 on Build GRCh38
rs756894023 Chr.3: 160677569-160677569 on Build GRCh38
rs139090920 Chr.3: 160677354-160677354 on Build GRCh38
rs914690092 Chr.3: 160677535-160677535 on Build GRCh38
rs755983383 Chr.3: 160677463-160677463 on Build GRCh38
rs200111812 Chr.3: 160677422-160677422 on Build GRCh38
rs969244931 Chr.3: 160677540-160677540 on Build GRCh38
rs773491530 Chr.3: 160677517-160677517 on Build GRCh38
rs79817049 Chr.3: 160677309-160677309 on Build GRCh38
rs755718463 Chr.3: 160677399-160677399 on Build GRCh38
rs772977295 Chr.3: 160677338-160677338 on Build GRCh38
rs765380940 Chr.3: 160677428-160677428 on Build GRCh38
rs115464520 Chr.3: 160677217-160677217 on Build GRCh38
rs151085412 Chr.3: 160677471-160677471 on Build GRCh38
rs754862377 Chr.3: 160677433-160677433 on Build GRCh38
rs76426610 Chr.3: 160677312-160677313 on Build GRCh38
rs750581360 Chr.3: 160677563-160677563 on Build GRCh38
rs369705585 Chr.3: 160677609-160677609 on Build GRCh38
rs1051186698 Chr.3: 160677107-160677107 on Build GRCh38
rs750136438 Chr.3: 160677571-160677571 on Build GRCh38
rs775691418 Chr.3: 160677482-160677482 on Build GRCh38
rs528242311 Chr.3: 160677248-160677248 on Build GRCh38
rs758252711 Chr.3: 160677308-160677308 on Build GRCh38
rs753812333 Chr.3: 160677567-160677567 on Build GRCh38
rs111444152 Chr.3: 160677304-160677305 on Build GRCh38
rs754044705 Chr.3: 160677307-160677307 on Build GRCh38
rs756289289 Chr.3: 160677302-160677309 on Build GRCh38
rs774859720 Chr.3: 160677408-160677408 on Build GRCh38
rs748999128 Chr.3: 160677405-160677405 on Build GRCh38
rs770665963 Chr.3: 160677406-160677406 on Build GRCh38
rs748652938 Chr.3: 160677591-160677591 on Build GRCh38
rs546151722 Chr.3: 160677100-160677100 on Build GRCh38
rs747630242 Chr.3: 160677328-160677328 on Build GRCh38
rs142708665 Chr.3: 160677112-160677112 on Build GRCh38
rs747273775 Chr.3: 160677579-160677579 on Build GRCh38
rs762555229 Chr.3: 160677575-160677575 on Build GRCh38
rs772524730 Chr.3: 160677480-160677480 on Build GRCh38
rs769755563 Chr.3: 160677408-160677408 on Build GRCh38
rs557175663 Chr.3: 160677417-160677417 on Build GRCh38
rs754095868 Chr.3: 160677372-160677372 on Build GRCh38
rs144011121 Chr.3: 160677355-160677355 on Build GRCh38
rs756294186 Chr.3: 160677302-160677302 on Build GRCh38
rs752387195 Chr.3: 160677553-160677553 on Build GRCh38
rs751371454 Chr.3: 160677429-160677429 on Build GRCh38
rs764279237 Chr.3: 160677423-160677423 on Build GRCh38
rs1047454023 Chr.3: 160677381-160677381 on Build GRCh38
rs761721721 Chr.3: 160677331-160677331 on Build GRCh38
rs199795781 Chr.3: 160677410-160677410 on Build GRCh38
rs760529385 Chr.3: 160677479-160677479 on Build GRCh38
rs146704564 Chr.3: 160677351-160677351 on Build GRCh38
rs373695187 Chr.3: 160677624-160677624 on Build GRCh38
rs764980611 Chr.3: 160677301-160677302 on Build GRCh38
rs145894570 Chr.3: 160677292-160677292 on Build GRCh38
rs201732930 Chr.3: 160677420-160677420 on Build GRCh38
rs772811944 Chr.3: 160677477-160677477 on Build GRCh38
rs746994462 Chr.3: 160677314-160677314 on Build GRCh38
rs767395235 Chr.3: 160677530-160677530 on Build GRCh38
rs1043629893 Chr.3: 160677110-160677110 on Build GRCh38
rs77496186 Chr.3: 160677272-160677272 on Build GRCh38
rs149008200 Chr.3: 160677379-160677379 on Build GRCh38
rs764074683 Chr.3: 160677360-160677360 on Build GRCh38
rs370792201 Chr.3: 160677449-160677449 on Build GRCh38
rs777386557 Chr.3: 160677404-160677404 on Build GRCh38
rs898240747 Chr.3: 160677436-160677436 on Build GRCh38
rs990240252 Chr.3: 160677332-160677332 on Build GRCh38
rs772964719 Chr.3: 160677410-160677411 on Build GRCh38
rs536528754 Chr.3: 160677302-160677303 on Build GRCh38
rs762057724 Chr.3: 160677425-160677425 on Build GRCh38
rs777455458 Chr.3: 160677464-160677464 on Build GRCh38
rs201353845 Chr.3: 160677313-160677313 on Build GRCh38
rs187614470 Chr.3: 160677473-160677473 on Build GRCh38
rs770991598 Chr.3: 160677602-160677602 on Build GRCh38
rs757934795 Chr.3: 160677572-160677572 on Build GRCh38
rs945259190 Chr.3: 160677106-160677106 on Build GRCh38
rs146546740 Chr.3: 160677524-160677524 on Build GRCh38
rs898272129 Chr.3: 160677617-160677617 on Build GRCh38
rs748023818 Chr.3: 160677390-160677390 on Build GRCh38
rs531088682 Chr.3: 160677285-160677285 on Build GRCh38
rs200837427 Chr.3: 160677491-160677491 on Build GRCh38
rs761826350 Chr.3: 160677468-160677468 on Build GRCh38
rs974905530 Chr.3: 160677300-160677300 on Build GRCh38
rs750226309 Chr.3: 160677302-160677305 on Build GRCh38
rs768315352 Chr.3: 160677401-160677401 on Build GRCh38
rs950431316 Chr.3: 160677222-160677222 on Build GRCh38
rs34154432 Chr.3: 160677604-160677604 on Build GRCh38
rs760863756 Chr.3: 160677357-160677357 on Build GRCh38
rs776701439 Chr.3: 160677502-160677502 on Build GRCh38
rs762844930 Chr.3: 160677565-160677568 on Build GRCh38
rs747570888 Chr.3: 160677487-160677487 on Build GRCh38
rs35634980 Chr.3: 160677593-160677593 on Build GRCh38
rs79644959 Chr.3: 160677123-160677123 on Build GRCh38
rs760603451 Chr.3: 160677607-160677607 on Build GRCh38
rs17853289 Chr.3: 160677518-160677518 on Build GRCh38
rs923449097 Chr.3: 160677588-160677588 on Build GRCh38
rs778531724 Chr.3: 160677570-160677570 on Build GRCh38
rs769169425 Chr.3: 160677580-160677580 on Build GRCh38

View Full Product Details