Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18315899-18316333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSM4
Entrez Gene ID
25804
Gene Name
LSM4 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
GRP, YER112W
UniGene
Hs.515255
Gene Chromosome Location
Chr.19: 18306230-18323191 on Build GRCh38
Transcripts
NM_001252129.1 NM_012321.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712052 c.40C>T p.P14S Chr.19: 18316029-18316029 on Build GRCh38
711117 c.8C>T p.P3L Chr.19: 18316061-18316061 on Build GRCh38
438969 c.35delA p.N12fs*8 Chr.19: 18316034-18316034 on Build GRCh38
343503 c.24G>A p.K8K Chr.19: 18316045-18316045 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035614881 Chr.19: 18316293-18316293 on Build GRCh38
rs142711541 Chr.19: 18316030-18316030 on Build GRCh38
rs781724426 Chr.19: 18316127-18316127 on Build GRCh38
rs905905089 Chr.19: 18316204-18316204 on Build GRCh38
rs757029358 Chr.19: 18316168-18316168 on Build GRCh38
rs754918610 Chr.19: 18316097-18316097 on Build GRCh38
rs199963944 Chr.19: 18316101-18316101 on Build GRCh38
rs1002102279 Chr.19: 18315965-18315965 on Build GRCh38
rs752211582 Chr.19: 18316100-18316100 on Build GRCh38
rs560908507 Chr.19: 18316198-18316198 on Build GRCh38
rs911440581 Chr.19: 18316172-18316172 on Build GRCh38
rs1014816072 Chr.19: 18316299-18316299 on Build GRCh38
rs2232965 Chr.19: 18316140-18316140 on Build GRCh38
rs767702721 Chr.19: 18316156-18316156 on Build GRCh38
rs1022980789 Chr.19: 18315963-18315963 on Build GRCh38
rs749836187 Chr.19: 18316183-18316183 on Build GRCh38
rs760162322 Chr.19: 18316019-18316019 on Build GRCh38
rs780236548 Chr.19: 18316180-18316180 on Build GRCh38
rs574363669 Chr.19: 18316220-18316220 on Build GRCh38
rs760158389 Chr.19: 18316078-18316079 on Build GRCh38
rs534942965 Chr.19: 18315949-18315949 on Build GRCh38
rs907724951 Chr.19: 18316128-18316128 on Build GRCh38
rs1044857799 Chr.19: 18316201-18316201 on Build GRCh38
rs753597235 Chr.19: 18316096-18316096 on Build GRCh38
rs879716723 Chr.19: 18316085-18316085 on Build GRCh38
rs1034890356 Chr.19: 18315987-18315987 on Build GRCh38
rs761978097 Chr.19: 18316084-18316084 on Build GRCh38
rs539235634 Chr.19: 18315980-18315980 on Build GRCh38
rs775491191 Chr.19: 18316042-18316042 on Build GRCh38
rs371218311 Chr.19: 18316013-18316013 on Build GRCh38
rs906985733 Chr.19: 18316262-18316262 on Build GRCh38
rs764017733 Chr.19: 18316078-18316078 on Build GRCh38
rs766195945 Chr.19: 18316088-18316088 on Build GRCh38
rs1010433225 Chr.19: 18316229-18316229 on Build GRCh38
rs747669103 Chr.19: 18315989-18315989 on Build GRCh38
rs751797136 Chr.19: 18316079-18316079 on Build GRCh38
rs1037169479 Chr.19: 18315939-18315939 on Build GRCh38
rs559233199 Chr.19: 18315998-18315998 on Build GRCh38
rs536718275 Chr.19: 18315921-18315921 on Build GRCh38
rs556681258 Chr.19: 18315948-18315948 on Build GRCh38
rs764869953 Chr.19: 18316161-18316161 on Build GRCh38
rs758218871 Chr.19: 18316162-18316162 on Build GRCh38
rs1009409935 Chr.19: 18316306-18316306 on Build GRCh38
rs763314973 Chr.19: 18316141-18316141 on Build GRCh38
rs769122653 Chr.19: 18316184-18316184 on Build GRCh38
rs774377966 Chr.19: 18316147-18316147 on Build GRCh38
rs753688153 Chr.19: 18316157-18316157 on Build GRCh38
rs1002835635 Chr.19: 18316235-18316235 on Build GRCh38
rs2232966 Chr.19: 18316048-18316048 on Build GRCh38
rs373801881 Chr.19: 18316053-18316053 on Build GRCh38
rs1004377957 Chr.19: 18315935-18315935 on Build GRCh38
rs745445451 Chr.19: 18316169-18316169 on Build GRCh38
rs777359881 Chr.19: 18316108-18316108 on Build GRCh38
rs796478752 Chr.19: 18315982-18315982 on Build GRCh38
rs754907337 Chr.19: 18315973-18315973 on Build GRCh38
rs762064255 Chr.19: 18316149-18316149 on Build GRCh38
rs766316223 Chr.19: 18316151-18316152 on Build GRCh38
rs1034438158 Chr.19: 18316281-18316281 on Build GRCh38
rs922704199 Chr.19: 18316152-18316152 on Build GRCh38
rs746223367 Chr.19: 18316131-18316131 on Build GRCh38
rs898583901 Chr.19: 18315943-18315943 on Build GRCh38
rs60931638 Chr.19: 18315981-18315981 on Build GRCh38
rs1001168986 Chr.19: 18316267-18316267 on Build GRCh38
rs777085451 Chr.19: 18316018-18316018 on Build GRCh38
rs181725157 Chr.19: 18316294-18316294 on Build GRCh38
rs970451929 Chr.19: 18315964-18315964 on Build GRCh38
rs898632514 Chr.19: 18316194-18316194 on Build GRCh38
rs770272158 Chr.19: 18316132-18316132 on Build GRCh38
rs773352798 Chr.19: 18316000-18316000 on Build GRCh38
rs778892934 Chr.19: 18316098-18316098 on Build GRCh38
rs967837440 Chr.19: 18316118-18316118 on Build GRCh38
rs891987485 Chr.19: 18316208-18316208 on Build GRCh38
rs190460229 Chr.19: 18316245-18316245 on Build GRCh38
rs750782945 Chr.19: 18316057-18316057 on Build GRCh38
rs768806352 Chr.19: 18316143-18316143 on Build GRCh38
rs763726479 Chr.19: 18316164-18316164 on Build GRCh38
rs770665734 Chr.19: 18316111-18316111 on Build GRCh38
rs771434408 Chr.19: 18316009-18316009 on Build GRCh38
rs746681277 Chr.19: 18316109-18316109 on Build GRCh38

View Full Product Details