Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 36961836-36962272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPBL
Entrez Gene ID
25836
Gene Name
NIPBL, cohesin loading factor
Gene Aliases
CDLS, CDLS1, IDN3, IDN3-B, Scc2
UniGene
Hs.481927
Gene Chromosome Location
Chr.5: 36876759-37065819 on Build GRCh38
Transcripts
NM_015384.4 NM_133433.3 XM_005248280.3 XM_006714467.2 XM_006714468.2 XM_011514015.1 XM_017009329.1 XM_017009331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1067642 c.462G>A p.R154R Chr.5: 36962126-36962126 on Build GRCh38
4881587 c.529C>G p.P177A Chr.5: 36962193-36962193 on Build GRCh38
6347901 c.494G>T p.R165I Chr.5: 36962158-36962158 on Build GRCh38
4495801 c.460C>T p.R154W Chr.5: 36962124-36962124 on Build GRCh38
3855001 c.467T>C p.V156A Chr.5: 36962131-36962131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753137703 Chr.5: 36962095-36962095 on Build GRCh38
rs377385598 Chr.5: 36962074-36962074 on Build GRCh38
rs536895037 Chr.5: 36962147-36962147 on Build GRCh38
rs761663409 Chr.5: 36962183-36962183 on Build GRCh38
rs780261645 Chr.5: 36962089-36962089 on Build GRCh38
rs774880166 Chr.5: 36962099-36962099 on Build GRCh38
rs550326012 Chr.5: 36961993-36961993 on Build GRCh38
rs748523012 Chr.5: 36962202-36962202 on Build GRCh38
rs1052544132 Chr.5: 36962136-36962136 on Build GRCh38
rs1015312566 Chr.5: 36961998-36961998 on Build GRCh38
rs1049123061 Chr.5: 36961911-36961911 on Build GRCh38
rs778285743 Chr.5: 36962228-36962228 on Build GRCh38
rs80358360 Chr.5: 36962249-36962249 on Build GRCh38
rs181226022 Chr.5: 36961983-36961983 on Build GRCh38
rs901853977 Chr.5: 36962019-36962019 on Build GRCh38
rs1015296942 Chr.5: 36961968-36961968 on Build GRCh38
rs576458890 Chr.5: 36962193-36962193 on Build GRCh38
rs200692598 Chr.5: 36962125-36962125 on Build GRCh38
rs749472246 Chr.5: 36962093-36962093 on Build GRCh38
rs763783306 Chr.5: 36962143-36962143 on Build GRCh38
rs756481603 Chr.5: 36962117-36962117 on Build GRCh38
rs1029279171 Chr.5: 36962070-36962070 on Build GRCh38
rs887886821 Chr.5: 36961966-36961966 on Build GRCh38
rs539760615 Chr.5: 36962116-36962116 on Build GRCh38
rs146321563 Chr.5: 36962130-36962130 on Build GRCh38
rs139514413 Chr.5: 36962124-36962124 on Build GRCh38
rs568557020 Chr.5: 36962058-36962058 on Build GRCh38
rs962377978 Chr.5: 36961976-36961976 on Build GRCh38
rs748576180 Chr.5: 36962100-36962100 on Build GRCh38
rs750337367 Chr.5: 36962192-36962192 on Build GRCh38
rs780026320 Chr.5: 36962195-36962195 on Build GRCh38
rs372605856 Chr.5: 36962223-36962223 on Build GRCh38
rs939532340 Chr.5: 36962131-36962131 on Build GRCh38
rs1051015636 Chr.5: 36962245-36962245 on Build GRCh38
rs536085162 Chr.5: 36962072-36962072 on Build GRCh38
rs767425012 Chr.5: 36962186-36962186 on Build GRCh38
rs998139636 Chr.5: 36962067-36962067 on Build GRCh38
rs367997192 Chr.5: 36962081-36962081 on Build GRCh38
rs148542094 Chr.5: 36962198-36962198 on Build GRCh38
rs754045462 Chr.5: 36962197-36962197 on Build GRCh38
rs762615203 Chr.5: 36962137-36962137 on Build GRCh38
rs769203117 Chr.5: 36962126-36962126 on Build GRCh38
rs142923613 Chr.5: 36962199-36962199 on Build GRCh38
rs1004942671 Chr.5: 36961971-36961971 on Build GRCh38

View Full Product Details