Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53148613-53149187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCM1
Entrez Gene ID
8521
Gene Name
glial cells missing homolog 1
Gene Aliases
GCMA, hGCMa
UniGene
Hs.23589
Gene Chromosome Location
Chr.6: 53125644-53262803 on Build GRCh38
Transcripts
NM_003643.3 XM_017011390.1

SNP Detail

SNP ID Chromosome Location
rs1042578782 Chr.6: 53148945-53148945 on Build GRCh38
rs981780717 Chr.6: 53149129-53149129 on Build GRCh38
rs901688793 Chr.6: 53148918-53148918 on Build GRCh38
rs182663758 Chr.6: 53149080-53149080 on Build GRCh38
rs552852811 Chr.6: 53148703-53148703 on Build GRCh38
rs1017447777 Chr.6: 53149061-53149061 on Build GRCh38
rs116576029 Chr.6: 53148791-53148791 on Build GRCh38
rs1035074357 Chr.6: 53149148-53149148 on Build GRCh38
rs1010560023 Chr.6: 53148719-53148719 on Build GRCh38
rs1000170273 Chr.6: 53148702-53148702 on Build GRCh38
rs574882021 Chr.6: 53148907-53148907 on Build GRCh38
rs905882741 Chr.6: 53148665-53148665 on Build GRCh38
rs546570751 Chr.6: 53149041-53149041 on Build GRCh38
rs951785044 Chr.6: 53148737-53148738 on Build GRCh38
rs186160862 Chr.6: 53148732-53148732 on Build GRCh38
rs963305674 Chr.6: 53148776-53148776 on Build GRCh38
rs563342645 Chr.6: 53149153-53149153 on Build GRCh38
rs927436479 Chr.6: 53149155-53149155 on Build GRCh38
rs950357915 Chr.6: 53148792-53148792 on Build GRCh38
rs1051641202 Chr.6: 53149033-53149033 on Build GRCh38
rs973284250 Chr.6: 53148784-53148784 on Build GRCh38
rs760451463 Chr.6: 53149071-53149071 on Build GRCh38
rs1029740210 Chr.6: 53148713-53148713 on Build GRCh38
rs891765188 Chr.6: 53148863-53148863 on Build GRCh38
rs898961988 Chr.6: 53149070-53149070 on Build GRCh38
rs936065799 Chr.6: 53148839-53148839 on Build GRCh38
rs529688107 Chr.6: 53148953-53148953 on Build GRCh38
rs769744028 Chr.6: 53148639-53148639 on Build GRCh38
rs538317608 Chr.6: 53148645-53148645 on Build GRCh38
rs146572983 Chr.6: 53148707-53148707 on Build GRCh38
rs990940898 Chr.6: 53149158-53149158 on Build GRCh38
rs762977160 Chr.6: 53148801-53148801 on Build GRCh38
rs917462348 Chr.6: 53149076-53149076 on Build GRCh38
rs181693845 Chr.6: 53148699-53148699 on Build GRCh38
rs938199880 Chr.6: 53148881-53148881 on Build GRCh38
rs543563564 Chr.6: 53149119-53149119 on Build GRCh38
rs867730376 Chr.6: 53148849-53148849 on Build GRCh38
rs142362601 Chr.6: 53148951-53148951 on Build GRCh38
rs1007359483 Chr.6: 53149055-53149055 on Build GRCh38
rs1026226253 Chr.6: 53149072-53149072 on Build GRCh38
rs974536757 Chr.6: 53148733-53148733 on Build GRCh38
rs554700367 Chr.6: 53148646-53148646 on Build GRCh38
rs1007375627 Chr.6: 53148734-53148734 on Build GRCh38
rs574767488 Chr.6: 53148673-53148673 on Build GRCh38
rs191011973 Chr.6: 53148924-53148924 on Build GRCh38
rs917429887 Chr.6: 53148787-53148787 on Build GRCh38
rs1010156840 Chr.6: 53148897-53148897 on Build GRCh38
rs1017834219 Chr.6: 53148768-53148768 on Build GRCh38
rs927720761 Chr.6: 53148850-53148850 on Build GRCh38
rs149172766 Chr.6: 53148730-53148730 on Build GRCh38
rs940312615 Chr.6: 53148769-53148769 on Build GRCh38
rs766559330 Chr.6: 53148902-53148902 on Build GRCh38

View Full Product Details