Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68158319-68158814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KMT5B
Entrez Gene ID
51111
Gene Name
lysine methyltransferase 5B
Gene Aliases
CGI-85, CGI85, SUV420H1
UniGene
Hs.632120
Gene Chromosome Location
Chr.11: 68154863-68213904 on Build GRCh38
Transcripts
NM_001300907.1 NM_001300908.1 NM_017635.4 XM_005274035.3 XM_005274036.3 XM_006718581.1 XM_011545091.1 XM_011545092.2 XM_011545093.2 XM_017017876.1 XM_017017877.1 XM_017017878.1 XM_017017879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4474855 c.1949C>T p.S650F Chr.11: 68158397-68158397 on Build GRCh38
5475564 c.1784C>T p.A595V Chr.11: 68158562-68158562 on Build GRCh38
1356502 c.1707G>A p.T569T Chr.11: 68158639-68158639 on Build GRCh38
4865200 c.1615C>T p.R539W Chr.11: 68158731-68158731 on Build GRCh38
4574724 c.1619G>A p.R540Q Chr.11: 68158727-68158727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922405902 Chr.11: 68158498-68158498 on Build GRCh38
rs751653184 Chr.11: 68158671-68158671 on Build GRCh38
rs374890131 Chr.11: 68158717-68158717 on Build GRCh38
rs757362302 Chr.11: 68158687-68158687 on Build GRCh38
rs780554987 Chr.11: 68158788-68158788 on Build GRCh38
rs367596003 Chr.11: 68158730-68158730 on Build GRCh38
rs772536209 Chr.11: 68158490-68158490 on Build GRCh38
rs779903947 Chr.11: 68158575-68158575 on Build GRCh38
rs750243156 Chr.11: 68158565-68158565 on Build GRCh38
rs377489614 Chr.11: 68158340-68158340 on Build GRCh38
rs763718036 Chr.11: 68158553-68158553 on Build GRCh38
rs3824853 Chr.11: 68158700-68158700 on Build GRCh38
rs777943905 Chr.11: 68158726-68158728 on Build GRCh38
rs760069840 Chr.11: 68158525-68158525 on Build GRCh38
rs776905134 Chr.11: 68158524-68158524 on Build GRCh38
rs183505938 Chr.11: 68158675-68158675 on Build GRCh38
rs144513563 Chr.11: 68158446-68158446 on Build GRCh38
rs752309170 Chr.11: 68158439-68158439 on Build GRCh38
rs756056561 Chr.11: 68158487-68158487 on Build GRCh38
rs139658126 Chr.11: 68158627-68158627 on Build GRCh38
rs370541174 Chr.11: 68158348-68158348 on Build GRCh38
rs766136425 Chr.11: 68158754-68158754 on Build GRCh38
rs781083698 Chr.11: 68158444-68158446 on Build GRCh38
rs778314058 Chr.11: 68158349-68158349 on Build GRCh38
rs761165640 Chr.11: 68158737-68158737 on Build GRCh38
rs745667169 Chr.11: 68158447-68158447 on Build GRCh38
rs778951109 Chr.11: 68158495-68158495 on Build GRCh38
rs746122050 Chr.11: 68158714-68158714 on Build GRCh38
rs780882627 Chr.11: 68158477-68158477 on Build GRCh38
rs199581094 Chr.11: 68158783-68158783 on Build GRCh38
rs776530029 Chr.11: 68158418-68158418 on Build GRCh38
rs758426671 Chr.11: 68158588-68158588 on Build GRCh38
rs138240281 Chr.11: 68158396-68158396 on Build GRCh38
rs745816889 Chr.11: 68158397-68158397 on Build GRCh38
rs760334121 Chr.11: 68158644-68158644 on Build GRCh38
rs1012452486 Chr.11: 68158649-68158649 on Build GRCh38
rs962026772 Chr.11: 68158742-68158742 on Build GRCh38
rs113932704 Chr.11: 68158626-68158626 on Build GRCh38
rs759998936 Chr.11: 68158428-68158428 on Build GRCh38
rs759460249 Chr.11: 68158761-68158761 on Build GRCh38
rs1028680975 Chr.11: 68158625-68158625 on Build GRCh38
rs781214162 Chr.11: 68158713-68158713 on Build GRCh38
rs562653357 Chr.11: 68158639-68158639 on Build GRCh38
rs776088670 Chr.11: 68158623-68158623 on Build GRCh38
rs772693534 Chr.11: 68158750-68158750 on Build GRCh38
rs144521985 Chr.11: 68158640-68158640 on Build GRCh38
rs199946263 Chr.11: 68158536-68158536 on Build GRCh38
rs751219442 Chr.11: 68158554-68158554 on Build GRCh38
rs146143706 Chr.11: 68158766-68158766 on Build GRCh38
rs747260401 Chr.11: 68158521-68158521 on Build GRCh38
rs149239650 Chr.11: 68158429-68158429 on Build GRCh38
rs575248505 Chr.11: 68158424-68158424 on Build GRCh38
rs988475410 Chr.11: 68158383-68158383 on Build GRCh38
rs757026529 Chr.11: 68158462-68158462 on Build GRCh38
rs148005738 Chr.11: 68158503-68158503 on Build GRCh38
rs745788625 Chr.11: 68158485-68158485 on Build GRCh38
rs150879582 Chr.11: 68158609-68158609 on Build GRCh38
rs978515685 Chr.11: 68158354-68158354 on Build GRCh38
rs201627982 Chr.11: 68158753-68158753 on Build GRCh38
rs754710480 Chr.11: 68158387-68158388 on Build GRCh38
rs748297446 Chr.11: 68158502-68158502 on Build GRCh38
rs746151863 Chr.11: 68158618-68158618 on Build GRCh38
rs751550288 Chr.11: 68158403-68158403 on Build GRCh38
rs756078170 Chr.11: 68158572-68158572 on Build GRCh38
rs773367069 Chr.11: 68158507-68158507 on Build GRCh38
rs140037372 Chr.11: 68158784-68158784 on Build GRCh38
rs201173636 Chr.11: 68158726-68158726 on Build GRCh38
rs560196983 Chr.11: 68158759-68158759 on Build GRCh38
rs146086325 Chr.11: 68158425-68158425 on Build GRCh38
rs770635858 Chr.11: 68158388-68158388 on Build GRCh38
rs201400802 Chr.11: 68158361-68158361 on Build GRCh38
rs748348152 Chr.11: 68158412-68158412 on Build GRCh38
rs777813543 Chr.11: 68158666-68158666 on Build GRCh38
rs756901018 Chr.11: 68158558-68158558 on Build GRCh38
rs774055599 Chr.11: 68158630-68158630 on Build GRCh38
rs777696815 Chr.11: 68158366-68158366 on Build GRCh38
rs138680984 Chr.11: 68158426-68158426 on Build GRCh38
rs142276849 Chr.11: 68158420-68158420 on Build GRCh38
rs754307757 Chr.11: 68158343-68158343 on Build GRCh38
rs777868705 Chr.11: 68158599-68158599 on Build GRCh38
rs141548205 Chr.11: 68158581-68158581 on Build GRCh38
rs371880484 Chr.11: 68158663-68158663 on Build GRCh38
rs150905417 Chr.11: 68158751-68158751 on Build GRCh38
rs187695236 Chr.11: 68158560-68158560 on Build GRCh38
rs773565168 Chr.11: 68158423-68158423 on Build GRCh38
rs771629925 Chr.11: 68158749-68158749 on Build GRCh38
rs771292792 Chr.11: 68158523-68158523 on Build GRCh38
rs781361096 Chr.11: 68158778-68158778 on Build GRCh38
rs748061265 Chr.11: 68158507-68158509 on Build GRCh38
rs373030307 Chr.11: 68158621-68158623 on Build GRCh38
rs765088696 Chr.11: 68158762-68158762 on Build GRCh38
rs766185889 Chr.11: 68158655-68158655 on Build GRCh38
rs769344868 Chr.11: 68158728-68158728 on Build GRCh38
rs368394871 Chr.11: 68158461-68158461 on Build GRCh38
rs770295770 Chr.11: 68158620-68158620 on Build GRCh38
rs771112482 Chr.11: 68158613-68158613 on Build GRCh38
rs746867205 Chr.11: 68158380-68158380 on Build GRCh38
rs377537908 Chr.11: 68158365-68158365 on Build GRCh38
rs768156559 Chr.11: 68158617-68158617 on Build GRCh38
rs753741394 Chr.11: 68158658-68158658 on Build GRCh38
rs565603169 Chr.11: 68158727-68158727 on Build GRCh38
rs776143791 Chr.11: 68158538-68158538 on Build GRCh38
rs761514479 Chr.11: 68158634-68158634 on Build GRCh38

View Full Product Details