Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123630057-123630606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN3B
Entrez Gene ID
55800
Gene Name
sodium voltage-gated channel beta subunit 3
Gene Aliases
ATFB16, BRGDA7, HSA243396, SCNB3
UniGene
Hs.4865
Gene Chromosome Location
Chr.11: 123629187-123654607 on Build GRCh38
Transcripts
NM_001040151.1 NM_018400.3 XM_011542897.1

SNP Detail

SNP ID Chromosome Location
rs539098373 Chr.11: 123630342-123630342 on Build GRCh38
rs967998949 Chr.11: 123630268-123630268 on Build GRCh38
rs908832957 Chr.11: 123630467-123630467 on Build GRCh38
rs956740297 Chr.11: 123630334-123630334 on Build GRCh38
rs371488737 Chr.11: 123630299-123630299 on Build GRCh38
rs148818205 Chr.11: 123630329-123630329 on Build GRCh38
rs944419524 Chr.11: 123630535-123630535 on Build GRCh38
rs72552192 Chr.11: 123630282-123630282 on Build GRCh38
rs890144545 Chr.11: 123630267-123630267 on Build GRCh38
rs145531082 Chr.11: 123630158-123630158 on Build GRCh38
rs553697978 Chr.11: 123630314-123630314 on Build GRCh38
rs926832310 Chr.11: 123630326-123630326 on Build GRCh38
rs760875714 Chr.11: 123630241-123630241 on Build GRCh38
rs377271838 Chr.11: 123630355-123630355 on Build GRCh38
rs969429245 Chr.11: 123630373-123630373 on Build GRCh38
rs1005147462 Chr.11: 123630558-123630558 on Build GRCh38
rs72552191 Chr.11: 123630187-123630187 on Build GRCh38
rs1046759919 Chr.11: 123630466-123630466 on Build GRCh38
rs530374610 Chr.11: 123630125-123630125 on Build GRCh38
rs915365601 Chr.11: 123630095-123630095 on Build GRCh38
rs979686450 Chr.11: 123630283-123630283 on Build GRCh38
rs867908034 Chr.11: 123630523-123630523 on Build GRCh38
rs955473638 Chr.11: 123630199-123630199 on Build GRCh38
rs1010786624 Chr.11: 123630346-123630346 on Build GRCh38
rs974177213 Chr.11: 123630478-123630478 on Build GRCh38
rs927932571 Chr.11: 123630417-123630417 on Build GRCh38
rs117167932 Chr.11: 123630191-123630191 on Build GRCh38
rs980866675 Chr.11: 123630398-123630398 on Build GRCh38
rs1050020661 Chr.11: 123630259-123630259 on Build GRCh38
rs202081953 Chr.11: 123630343-123630343 on Build GRCh38
rs921445383 Chr.11: 123630484-123630484 on Build GRCh38
rs567925139 Chr.11: 123630293-123630293 on Build GRCh38
rs969377156 Chr.11: 123630099-123630099 on Build GRCh38
rs886047889 Chr.11: 123630557-123630557 on Build GRCh38
rs931707471 Chr.11: 123630509-123630509 on Build GRCh38
rs898880092 Chr.11: 123630225-123630225 on Build GRCh38
rs538356578 Chr.11: 123630238-123630238 on Build GRCh38
rs1047329505 Chr.11: 123630186-123630186 on Build GRCh38
rs949822608 Chr.11: 123630366-123630366 on Build GRCh38
rs886047888 Chr.11: 123630517-123630517 on Build GRCh38
rs750444172 Chr.11: 123630492-123630492 on Build GRCh38
rs1046714313 Chr.11: 123630087-123630087 on Build GRCh38
rs1038269619 Chr.11: 123630565-123630565 on Build GRCh38
rs770600572 Chr.11: 123630247-123630247 on Build GRCh38
rs941618283 Chr.11: 123630473-123630473 on Build GRCh38
rs190602957 Chr.11: 123630251-123630251 on Build GRCh38
rs939570830 Chr.11: 123630162-123630162 on Build GRCh38
rs563098976 Chr.11: 123630090-123630090 on Build GRCh38
rs886047887 Chr.11: 123630493-123630493 on Build GRCh38
rs1050154875 Chr.11: 123630516-123630516 on Build GRCh38
rs941566867 Chr.11: 123630190-123630190 on Build GRCh38
rs1019265426 Chr.11: 123630432-123630432 on Build GRCh38
rs927959841 Chr.11: 123630134-123630134 on Build GRCh38
rs1052781623 Chr.11: 123630570-123630570 on Build GRCh38
rs182838795 Chr.11: 123630183-123630183 on Build GRCh38
rs983425940 Chr.11: 123630433-123630433 on Build GRCh38
rs188355898 Chr.11: 123630234-123630234 on Build GRCh38

View Full Product Details