Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44885196-44885770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGM4
Entrez Gene ID
7047
Gene Name
transglutaminase 4
Gene Aliases
TGP, hTGP
UniGene
Hs.438265
Gene Chromosome Location
Chr.3: 44874592-44914601 on Build GRCh38
Transcripts
NM_003241.3 XM_011534042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5974796 c.112C>T p.R38* Chr.3: 44885417-44885417 on Build GRCh38
5719170 c.147C>A p.N49K Chr.3: 44885452-44885452 on Build GRCh38
3067430 c.48G>C p.L16F Chr.3: 44885353-44885353 on Build GRCh38
5004817 c.63C>T p.A21A Chr.3: 44885368-44885368 on Build GRCh38
3067435 c.151C>T p.P51S Chr.3: 44885456-44885456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750069289 Chr.3: 44885338-44885338 on Build GRCh38
rs201384600 Chr.3: 44885413-44885413 on Build GRCh38
rs764787433 Chr.3: 44885336-44885336 on Build GRCh38
rs1007312476 Chr.3: 44885707-44885707 on Build GRCh38
rs904701344 Chr.3: 44885239-44885239 on Build GRCh38
rs751626855 Chr.3: 44885432-44885433 on Build GRCh38
rs751487088 Chr.3: 44885278-44885278 on Build GRCh38
rs994531940 Chr.3: 44885694-44885694 on Build GRCh38
rs781658460 Chr.3: 44885449-44885449 on Build GRCh38
rs373455840 Chr.3: 44885508-44885508 on Build GRCh38
rs748832317 Chr.3: 44885452-44885452 on Build GRCh38
rs770407946 Chr.3: 44885450-44885450 on Build GRCh38
rs781726203 Chr.3: 44885521-44885521 on Build GRCh38
rs879135776 Chr.3: 44885435-44885435 on Build GRCh38
rs1036245476 Chr.3: 44885682-44885682 on Build GRCh38
rs985588015 Chr.3: 44885294-44885294 on Build GRCh38
rs765842579 Chr.3: 44885735-44885735 on Build GRCh38
rs906374769 Chr.3: 44885650-44885650 on Build GRCh38
rs761414312 Chr.3: 44885333-44885333 on Build GRCh38
rs759149143 Chr.3: 44885462-44885462 on Build GRCh38
rs1027410248 Chr.3: 44885490-44885490 on Build GRCh38
rs746053672 Chr.3: 44885309-44885309 on Build GRCh38
rs199591140 Chr.3: 44885402-44885402 on Build GRCh38
rs776349690 Chr.3: 44885327-44885327 on Build GRCh38
rs751865519 Chr.3: 44885513-44885513 on Build GRCh38
rs36081973 Chr.3: 44885365-44885365 on Build GRCh38
rs552193015 Chr.3: 44885474-44885474 on Build GRCh38
rs756015035 Chr.3: 44885438-44885438 on Build GRCh38
rs915340301 Chr.3: 44885600-44885600 on Build GRCh38
rs747413936 Chr.3: 44885314-44885314 on Build GRCh38
rs535311644 Chr.3: 44885283-44885283 on Build GRCh38
rs534397213 Chr.3: 44885517-44885517 on Build GRCh38
rs758615516 Chr.3: 44885441-44885441 on Build GRCh38
rs1054170121 Chr.3: 44885595-44885595 on Build GRCh38
rs752662091 Chr.3: 44885434-44885434 on Build GRCh38
rs767251214 Chr.3: 44885431-44885431 on Build GRCh38
rs113058339 Chr.3: 44885614-44885614 on Build GRCh38
rs369242368 Chr.3: 44885423-44885423 on Build GRCh38
rs775244330 Chr.3: 44885477-44885477 on Build GRCh38
rs766325168 Chr.3: 44885511-44885511 on Build GRCh38
rs754934190 Chr.3: 44885363-44885363 on Build GRCh38
rs762569716 Chr.3: 44885418-44885418 on Build GRCh38
rs751353474 Chr.3: 44885352-44885352 on Build GRCh38
rs1027705513 Chr.3: 44885702-44885702 on Build GRCh38
rs372280514 Chr.3: 44885306-44885306 on Build GRCh38
rs759822246 Chr.3: 44885462-44885462 on Build GRCh38
rs141141838 Chr.3: 44885369-44885369 on Build GRCh38
rs369927794 Chr.3: 44885648-44885648 on Build GRCh38
rs201282380 Chr.3: 44885445-44885445 on Build GRCh38
rs200524990 Chr.3: 44885509-44885509 on Build GRCh38
rs770288413 Chr.3: 44885457-44885457 on Build GRCh38
rs762632161 Chr.3: 44885347-44885347 on Build GRCh38
rs768073730 Chr.3: 44885750-44885778 on Build GRCh38
rs777885331 Chr.3: 44885290-44885290 on Build GRCh38
rs148245944 Chr.3: 44885414-44885414 on Build GRCh38
rs202085487 Chr.3: 44885451-44885451 on Build GRCh38
rs574601519 Chr.3: 44885230-44885230 on Build GRCh38
rs138221415 Chr.3: 44885383-44885383 on Build GRCh38
rs772318632 Chr.3: 44885390-44885390 on Build GRCh38
rs201921648 Chr.3: 44885316-44885316 on Build GRCh38
rs115097986 Chr.3: 44885439-44885439 on Build GRCh38
rs201445192 Chr.3: 44885472-44885472 on Build GRCh38
rs34630572 Chr.3: 44885368-44885368 on Build GRCh38
rs546645441 Chr.3: 44885527-44885527 on Build GRCh38
rs769273451 Chr.3: 44885320-44885320 on Build GRCh38
rs747144892 Chr.3: 44885447-44885447 on Build GRCh38
rs191466796 Chr.3: 44885635-44885635 on Build GRCh38
rs368559848 Chr.3: 44885295-44885295 on Build GRCh38
rs766549776 Chr.3: 44885279-44885279 on Build GRCh38
rs752570416 Chr.3: 44885366-44885366 on Build GRCh38
rs924152556 Chr.3: 44885551-44885551 on Build GRCh38
rs769465408 Chr.3: 44885416-44885416 on Build GRCh38
rs556737663 Chr.3: 44885663-44885663 on Build GRCh38
rs962507437 Chr.3: 44885569-44885569 on Build GRCh38
rs900059591 Chr.3: 44885453-44885453 on Build GRCh38
rs770929696 Chr.3: 44885296-44885296 on Build GRCh38
rs368986340 Chr.3: 44885537-44885537 on Build GRCh38
rs530630256 Chr.3: 44885703-44885703 on Build GRCh38
rs1014533446 Chr.3: 44885221-44885221 on Build GRCh38
rs201971224 Chr.3: 44885425-44885425 on Build GRCh38
rs554382271 Chr.3: 44885675-44885675 on Build GRCh38
rs113353665 Chr.3: 44885565-44885565 on Build GRCh38
rs897697706 Chr.3: 44885685-44885685 on Build GRCh38
rs747399684 Chr.3: 44885398-44885398 on Build GRCh38
rs557718516 Chr.3: 44885747-44885747 on Build GRCh38
rs920123258 Chr.3: 44885246-44885246 on Build GRCh38
rs760663947 Chr.3: 44885494-44885494 on Build GRCh38
rs556800894 Chr.3: 44885313-44885313 on Build GRCh38
rs768219492 Chr.3: 44885727-44885727 on Build GRCh38
rs1003400018 Chr.3: 44885433-44885433 on Build GRCh38
rs772751863 Chr.3: 44885417-44885417 on Build GRCh38
rs766047776 Chr.3: 44885422-44885422 on Build GRCh38
rs143546694 Chr.3: 44885351-44885351 on Build GRCh38
rs768970538 Chr.3: 44885634-44885634 on Build GRCh38
rs973176267 Chr.3: 44885236-44885236 on Build GRCh38
rs926470134 Chr.3: 44885649-44885649 on Build GRCh38
rs774074356 Chr.3: 44885458-44885458 on Build GRCh38
rs867774133 Chr.3: 44885448-44885451 on Build GRCh38
rs775665195 Chr.3: 44885397-44885397 on Build GRCh38
rs909678172 Chr.3: 44885356-44885356 on Build GRCh38
rs750505124 Chr.3: 44885440-44885440 on Build GRCh38

View Full Product Details