Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 6642300-6642898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRFAP1
Entrez Gene ID
93621
Gene Name
Morf4 family associated protein 1
Gene Aliases
PAM14, PGR1
UniGene
Hs.406590
Gene Chromosome Location
Chr.4: 6640091-6642745 on Build GRCh38
Transcripts
NM_001272053.1 NM_001272054.1 NM_033296.2

SNP Detail

SNP ID Chromosome Location
rs868514213 Chr.4: 6642859-6642859 on Build GRCh38
rs1000253392 Chr.4: 6642517-6642517 on Build GRCh38
rs1051519256 Chr.4: 6642483-6642483 on Build GRCh38
rs185670294 Chr.4: 6642834-6642834 on Build GRCh38
rs950694229 Chr.4: 6642373-6642373 on Build GRCh38
rs890629725 Chr.4: 6642453-6642453 on Build GRCh38
rs924292424 Chr.4: 6642459-6642459 on Build GRCh38
rs538246226 Chr.4: 6642602-6642602 on Build GRCh38
rs546963661 Chr.4: 6642511-6642511 on Build GRCh38
rs756949244 Chr.4: 6642348-6642348 on Build GRCh38
rs987739792 Chr.4: 6642650-6642650 on Build GRCh38
rs886448620 Chr.4: 6642568-6642568 on Build GRCh38
rs866395614 Chr.4: 6642532-6642532 on Build GRCh38
rs935799841 Chr.4: 6642714-6642714 on Build GRCh38
rs890205967 Chr.4: 6642492-6642492 on Build GRCh38
rs1059220 Chr.4: 6642664-6642664 on Build GRCh38
rs755993557 Chr.4: 6642801-6642801 on Build GRCh38
rs866663004 Chr.4: 6642609-6642609 on Build GRCh38
rs1007846698 Chr.4: 6642540-6642540 on Build GRCh38
rs891463841 Chr.4: 6642747-6642747 on Build GRCh38
rs559047901 Chr.4: 6642565-6642565 on Build GRCh38
rs181362943 Chr.4: 6642828-6642828 on Build GRCh38
rs982162447 Chr.4: 6642374-6642374 on Build GRCh38
rs753512964 Chr.4: 6642878-6642878 on Build GRCh38
rs1022762747 Chr.4: 6642873-6642873 on Build GRCh38
rs539208149 Chr.4: 6642711-6642711 on Build GRCh38
rs968216787 Chr.4: 6642875-6642875 on Build GRCh38
rs146846138 Chr.4: 6642877-6642877 on Build GRCh38
rs919269459 Chr.4: 6642618-6642618 on Build GRCh38
rs915502771 Chr.4: 6642438-6642438 on Build GRCh38
rs993068285 Chr.4: 6642324-6642324 on Build GRCh38
rs567641220 Chr.4: 6642454-6642454 on Build GRCh38
rs560257481 Chr.4: 6642376-6642376 on Build GRCh38
rs12695 Chr.4: 6642546-6642546 on Build GRCh38
rs924156768 Chr.4: 6642683-6642683 on Build GRCh38
rs1001320778 Chr.4: 6642819-6642819 on Build GRCh38
rs904269211 Chr.4: 6642687-6642687 on Build GRCh38
rs1003521794 Chr.4: 6642855-6642855 on Build GRCh38
rs530295771 Chr.4: 6642479-6642479 on Build GRCh38
rs991424893 Chr.4: 6642629-6642629 on Build GRCh38
rs899108479 Chr.4: 6642550-6642550 on Build GRCh38
rs1035014892 Chr.4: 6642857-6642857 on Build GRCh38
rs891699346 Chr.4: 6642531-6642531 on Build GRCh38
rs151108558 Chr.4: 6642361-6642361 on Build GRCh38
rs141106213 Chr.4: 6642732-6642732 on Build GRCh38
rs567082856 Chr.4: 6642526-6642526 on Build GRCh38
rs1051898412 Chr.4: 6642433-6642433 on Build GRCh38
rs112442636 Chr.4: 6642475-6642475 on Build GRCh38
rs7660166 Chr.4: 6642420-6642420 on Build GRCh38
rs968185882 Chr.4: 6642652-6642652 on Build GRCh38
rs879723701 Chr.4: 6642793-6642793 on Build GRCh38
rs575113251 Chr.4: 6642680-6642683 on Build GRCh38
rs574796879 Chr.4: 6642610-6642610 on Build GRCh38
rs547740137 Chr.4: 6642449-6642449 on Build GRCh38
rs546321867 Chr.4: 6642400-6642400 on Build GRCh38
rs886355209 Chr.4: 6642853-6642853 on Build GRCh38
rs943506362 Chr.4: 6642667-6642667 on Build GRCh38
rs7660424 Chr.4: 6642600-6642600 on Build GRCh38
rs763139054 Chr.4: 6642572-6642572 on Build GRCh38
rs866145892 Chr.4: 6642733-6642733 on Build GRCh38
rs554971245 Chr.4: 6642607-6642607 on Build GRCh38
rs540553231 Chr.4: 6642644-6642644 on Build GRCh38
rs994824498 Chr.4: 6642325-6642325 on Build GRCh38
rs905811826 Chr.4: 6642804-6642804 on Build GRCh38
rs552917936 Chr.4: 6642340-6642340 on Build GRCh38
rs568732348 Chr.4: 6642446-6642446 on Build GRCh38
rs1024161583 Chr.4: 6642798-6642798 on Build GRCh38
rs912132599 Chr.4: 6642873-6642874 on Build GRCh38
rs767240893 Chr.4: 6642697-6642697 on Build GRCh38
rs934276402 Chr.4: 6642460-6642460 on Build GRCh38
rs926596394 Chr.4: 6642385-6642385 on Build GRCh38
rs374603605 Chr.4: 6642849-6642849 on Build GRCh38
rs553758871 Chr.4: 6642649-6642649 on Build GRCh38
rs1014096046 Chr.4: 6642751-6642751 on Build GRCh38
rs895108603 Chr.4: 6642869-6642869 on Build GRCh38
rs771407217 Chr.4: 6642558-6642558 on Build GRCh38
rs1031375380 Chr.4: 6642518-6642518 on Build GRCh38
rs535325295 Chr.4: 6642398-6642398 on Build GRCh38
rs550403434 Chr.4: 6642378-6642378 on Build GRCh38
rs899003480 Chr.4: 6642833-6642833 on Build GRCh38
rs919498631 Chr.4: 6642404-6642404 on Build GRCh38
rs1012351292 Chr.4: 6642872-6642872 on Build GRCh38
rs149814133 Chr.4: 6642831-6642831 on Build GRCh38
rs571112921 Chr.4: 6642360-6642360 on Build GRCh38

View Full Product Details