Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26545872-26546423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGN4
Entrez Gene ID
10473
Gene Name
high mobility group nucleosomal binding domain 4
Gene Aliases
HMG17L3, NHC
UniGene
Hs.236774
Gene Chromosome Location
Chr.6: 26538344-26546937 on Build GRCh38
Transcripts
NM_006353.2 XM_017010171.1 XM_017010172.1 XM_017010173.1

SNP Detail

SNP ID Chromosome Location
rs909113009 Chr.6: 26545975-26545975 on Build GRCh38
rs1056893393 Chr.6: 26546087-26546087 on Build GRCh38
rs746786857 Chr.6: 26546067-26546067 on Build GRCh38
rs745447446 Chr.6: 26545974-26545974 on Build GRCh38
rs571818122 Chr.6: 26545895-26545895 on Build GRCh38
rs763034341 Chr.6: 26546284-26546284 on Build GRCh38
rs1030405597 Chr.6: 26546249-26546249 on Build GRCh38
rs903783320 Chr.6: 26546120-26546120 on Build GRCh38
rs1043554986 Chr.6: 26546112-26546112 on Build GRCh38
rs904105384 Chr.6: 26546117-26546117 on Build GRCh38
rs768625602 Chr.6: 26546102-26546102 on Build GRCh38
rs972275423 Chr.6: 26546031-26546031 on Build GRCh38
rs928677249 Chr.6: 26546037-26546037 on Build GRCh38
rs1016416884 Chr.6: 26546329-26546329 on Build GRCh38
rs1804835 Chr.6: 26546266-26546266 on Build GRCh38
rs1029153664 Chr.6: 26545897-26545897 on Build GRCh38
rs999470304 Chr.6: 26546131-26546131 on Build GRCh38
rs779779308 Chr.6: 26546028-26546028 on Build GRCh38
rs138074103 Chr.6: 26546038-26546038 on Build GRCh38
rs773511632 Chr.6: 26546258-26546258 on Build GRCh38
rs962166909 Chr.6: 26546355-26546355 on Build GRCh38
rs972329292 Chr.6: 26546358-26546358 on Build GRCh38
rs909082137 Chr.6: 26546319-26546319 on Build GRCh38
rs986400041 Chr.6: 26545941-26545941 on Build GRCh38
rs962240891 Chr.6: 26546023-26546023 on Build GRCh38
rs769692276 Chr.6: 26546255-26546255 on Build GRCh38
rs901546598 Chr.6: 26546180-26546180 on Build GRCh38
rs765175769 Chr.6: 26546089-26546089 on Build GRCh38
rs941967777 Chr.6: 26545977-26545977 on Build GRCh38
rs928874862 Chr.6: 26546392-26546392 on Build GRCh38
rs953614030 Chr.6: 26546273-26546273 on Build GRCh38
rs193042267 Chr.6: 26546349-26546349 on Build GRCh38
rs145535311 Chr.6: 26545928-26545928 on Build GRCh38
rs953227604 Chr.6: 26546294-26546294 on Build GRCh38
rs553043550 Chr.6: 26546110-26546110 on Build GRCh38
rs752559038 Chr.6: 26546309-26546309 on Build GRCh38
rs1052761934 Chr.6: 26546132-26546132 on Build GRCh38
rs976544153 Chr.6: 26546241-26546241 on Build GRCh38
rs11966754 Chr.6: 26546036-26546036 on Build GRCh38
rs931847392 Chr.6: 26545940-26545940 on Build GRCh38
rs771934523 Chr.6: 26546013-26546013 on Build GRCh38
rs1055917040 Chr.6: 26546043-26546043 on Build GRCh38
rs550856870 Chr.6: 26546000-26546000 on Build GRCh38
rs956749914 Chr.6: 26546138-26546138 on Build GRCh38
rs761892159 Chr.6: 26546224-26546224 on Build GRCh38
rs976196477 Chr.6: 26545915-26545915 on Build GRCh38
rs536498208 Chr.6: 26546103-26546103 on Build GRCh38
rs1036023005 Chr.6: 26546032-26546032 on Build GRCh38

View Full Product Details