Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28907279-28907781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF12
Entrez Gene ID
57649
Gene Name
PHD finger protein 12
Gene Aliases
PF1
UniGene
Hs.444173
Gene Chromosome Location
Chr.17: 28905250-28951490 on Build GRCh38
Transcripts
NM_001033561.1 XM_005258015.3 XM_005258016.3 XM_011525078.2 XM_011525079.2 XM_017024903.1 XM_017024904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1324092 c.2525G>A p.C842Y Chr.17: 28907606-28907606 on Build GRCh38
1679676 c.2518C>G p.H840D Chr.17: 28907613-28907613 on Build GRCh38
4065087 c.2529A>G p.I843M Chr.17: 28907602-28907602 on Build GRCh38
3819167 c.2539G>A p.E847K Chr.17: 28907592-28907592 on Build GRCh38
4984088 c.2532C>T p.F844F Chr.17: 28907599-28907599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770469264 Chr.17: 28907637-28907637 on Build GRCh38
rs957607188 Chr.17: 28907579-28907579 on Build GRCh38
rs749629069 Chr.17: 28907668-28907668 on Build GRCh38
rs776255265 Chr.17: 28907708-28907708 on Build GRCh38
rs1036196250 Chr.17: 28907747-28907747 on Build GRCh38
rs767229168 Chr.17: 28907551-28907551 on Build GRCh38
rs769146676 Chr.17: 28907689-28907689 on Build GRCh38
rs752639694 Chr.17: 28907721-28907721 on Build GRCh38
rs577897325 Chr.17: 28907732-28907732 on Build GRCh38
rs760433256 Chr.17: 28907700-28907700 on Build GRCh38
rs751540249 Chr.17: 28907316-28907316 on Build GRCh38
rs765121157 Chr.17: 28907718-28907718 on Build GRCh38
rs1011449225 Chr.17: 28907584-28907584 on Build GRCh38
rs539810511 Chr.17: 28907421-28907421 on Build GRCh38
rs906951451 Chr.17: 28907347-28907347 on Build GRCh38
rs753715529 Chr.17: 28907571-28907571 on Build GRCh38
rs1016880358 Chr.17: 28907532-28907532 on Build GRCh38
rs772273487 Chr.17: 28907521-28907522 on Build GRCh38
rs887638822 Chr.17: 28907725-28907725 on Build GRCh38
rs928377136 Chr.17: 28907687-28907687 on Build GRCh38
rs372423347 Chr.17: 28907735-28907735 on Build GRCh38
rs776033292 Chr.17: 28907545-28907545 on Build GRCh38
rs755096425 Chr.17: 28907433-28907433 on Build GRCh38
rs772681017 Chr.17: 28907691-28907691 on Build GRCh38
rs1036325618 Chr.17: 28907427-28907428 on Build GRCh38
rs372161716 Chr.17: 28907717-28907717 on Build GRCh38
rs541236615 Chr.17: 28907638-28907638 on Build GRCh38
rs781482539 Chr.17: 28907500-28907500 on Build GRCh38
rs756452000 Chr.17: 28907619-28907619 on Build GRCh38
rs933334299 Chr.17: 28907748-28907748 on Build GRCh38
rs909425980 Chr.17: 28907399-28907399 on Build GRCh38
rs539725471 Chr.17: 28907342-28907342 on Build GRCh38
rs765924916 Chr.17: 28907564-28907564 on Build GRCh38
rs778656865 Chr.17: 28907573-28907573 on Build GRCh38
rs559265173 Chr.17: 28907730-28907730 on Build GRCh38
rs909385951 Chr.17: 28907709-28907709 on Build GRCh38
rs933386568 Chr.17: 28907528-28907528 on Build GRCh38
rs981125052 Chr.17: 28907664-28907664 on Build GRCh38
rs368783828 Chr.17: 28907581-28907581 on Build GRCh38
rs1018880959 Chr.17: 28907577-28907577 on Build GRCh38
rs770905812 Chr.17: 28907620-28907620 on Build GRCh38
rs897126834 Chr.17: 28907534-28907534 on Build GRCh38
rs183014573 Chr.17: 28907560-28907560 on Build GRCh38
rs913495476 Chr.17: 28907618-28907618 on Build GRCh38
rs765777345 Chr.17: 28907702-28907702 on Build GRCh38
rs761531957 Chr.17: 28907547-28907547 on Build GRCh38
rs752441350 Chr.17: 28907580-28907580 on Build GRCh38
rs191375479 Chr.17: 28907557-28907557 on Build GRCh38
rs937168966 Chr.17: 28907374-28907374 on Build GRCh38
rs1035877126 Chr.17: 28907690-28907690 on Build GRCh38
rs199533270 Chr.17: 28907572-28907572 on Build GRCh38
rs746327338 Chr.17: 28907632-28907632 on Build GRCh38
rs1006130815 Chr.17: 28907517-28907517 on Build GRCh38
rs942316864 Chr.17: 28907404-28907404 on Build GRCh38
rs750190531 Chr.17: 28907569-28907569 on Build GRCh38
rs967049646 Chr.17: 28907647-28907647 on Build GRCh38
rs140797913 Chr.17: 28907489-28907489 on Build GRCh38
rs150109867 Chr.17: 28907495-28907495 on Build GRCh38

View Full Product Details