Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36546788-36547336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF529
Entrez Gene ID
57711
Gene Name
zinc finger protein 529
Gene Aliases
-
UniGene
Hs.654960
Gene Chromosome Location
Chr.19: 36534491-36605411 on Build GRCh38
Transcripts
NM_001145649.1 NM_001145650.1 NM_001321351.1 NM_020951.4 XM_006723302.3 XM_006723304.2 XM_011527164.2 XM_011527165.2 XM_011527166.2 XM_011527167.2 XM_011527169.2 XM_011527170.1 XM_017027039.1 XM_017027040.1 XM_017027041.1 XM_017027042.1 XM_017027043.1 XM_017027044.1 XM_017027045.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1304473 c.1513C>T p.P505S Chr.19: 36547045-36547045 on Build GRCh38
6282088 c.1385C>T p.T462M Chr.19: 36547173-36547173 on Build GRCh38
7109735 c.1439G>T p.C480F Chr.19: 36547119-36547119 on Build GRCh38
6326262 c.1642C>A p.H548N Chr.19: 36546916-36546916 on Build GRCh38
711817 c.1517A>G p.Y506C Chr.19: 36547041-36547041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754340489 Chr.19: 36546896-36546896 on Build GRCh38
rs2278168 Chr.19: 36547190-36547190 on Build GRCh38
rs944040453 Chr.19: 36547310-36547310 on Build GRCh38
rs770746097 Chr.19: 36546919-36546919 on Build GRCh38
rs372576892 Chr.19: 36546924-36546924 on Build GRCh38
rs750759014 Chr.19: 36546871-36546871 on Build GRCh38
rs370804865 Chr.19: 36547185-36547185 on Build GRCh38
rs868440523 Chr.19: 36547219-36547219 on Build GRCh38
rs760023755 Chr.19: 36547002-36547002 on Build GRCh38
rs754354128 Chr.19: 36546992-36546992 on Build GRCh38
rs756086805 Chr.19: 36547210-36547212 on Build GRCh38
rs761966616 Chr.19: 36547256-36547256 on Build GRCh38
rs748392973 Chr.19: 36546902-36546902 on Build GRCh38
rs755968227 Chr.19: 36547172-36547172 on Build GRCh38
rs766333845 Chr.19: 36547290-36547290 on Build GRCh38
rs766699853 Chr.19: 36546982-36546982 on Build GRCh38
rs751823405 Chr.19: 36547044-36547044 on Build GRCh38
rs375503527 Chr.19: 36546957-36546957 on Build GRCh38
rs759049681 Chr.19: 36546848-36546848 on Build GRCh38
rs745813355 Chr.19: 36547046-36547046 on Build GRCh38
rs867056137 Chr.19: 36547246-36547246 on Build GRCh38
rs777934586 Chr.19: 36547041-36547041 on Build GRCh38
rs530802385 Chr.19: 36547162-36547162 on Build GRCh38
rs762231513 Chr.19: 36547195-36547195 on Build GRCh38
rs778914917 Chr.19: 36546900-36546900 on Build GRCh38
rs775956348 Chr.19: 36547233-36547233 on Build GRCh38
rs775187673 Chr.19: 36546858-36546858 on Build GRCh38
rs771319475 Chr.19: 36547088-36547088 on Build GRCh38
rs762571412 Chr.19: 36546867-36546867 on Build GRCh38
rs529869961 Chr.19: 36546981-36546981 on Build GRCh38
rs762992067 Chr.19: 36547112-36547112 on Build GRCh38
rs796075805 Chr.19: 36547206-36547206 on Build GRCh38
rs1049789748 Chr.19: 36547225-36547225 on Build GRCh38
rs764210844 Chr.19: 36547140-36547140 on Build GRCh38
rs375029468 Chr.19: 36547040-36547040 on Build GRCh38
rs750128723 Chr.19: 36547170-36547170 on Build GRCh38
rs769335591 Chr.19: 36546955-36546955 on Build GRCh38
rs377492229 Chr.19: 36547173-36547173 on Build GRCh38
rs879034805 Chr.19: 36547052-36547052 on Build GRCh38
rs527440575 Chr.19: 36546914-36546914 on Build GRCh38
rs749285886 Chr.19: 36547179-36547179 on Build GRCh38
rs776790047 Chr.19: 36547090-36547090 on Build GRCh38
rs752665319 Chr.19: 36547021-36547021 on Build GRCh38
rs372917053 Chr.19: 36546979-36546979 on Build GRCh38
rs780062358 Chr.19: 36547055-36547055 on Build GRCh38
rs774930403 Chr.19: 36546972-36546972 on Build GRCh38
rs767990256 Chr.19: 36546869-36546869 on Build GRCh38
rs867983848 Chr.19: 36547158-36547158 on Build GRCh38
rs764694417 Chr.19: 36546856-36546856 on Build GRCh38
rs371280643 Chr.19: 36547245-36547245 on Build GRCh38
rs762558120 Chr.19: 36546977-36546977 on Build GRCh38
rs1008214706 Chr.19: 36546923-36546923 on Build GRCh38
rs765774965 Chr.19: 36547018-36547018 on Build GRCh38
rs879029463 Chr.19: 36546901-36546901 on Build GRCh38
rs769553021 Chr.19: 36547049-36547049 on Build GRCh38
rs746762046 Chr.19: 36546910-36546910 on Build GRCh38
rs763511485 Chr.19: 36547242-36547242 on Build GRCh38
rs770236184 Chr.19: 36547210-36547210 on Build GRCh38
rs371970294 Chr.19: 36547035-36547035 on Build GRCh38
rs367645333 Chr.19: 36547142-36547142 on Build GRCh38
rs776457645 Chr.19: 36546823-36546823 on Build GRCh38
rs528447108 Chr.19: 36547298-36547298 on Build GRCh38
rs754520818 Chr.19: 36547181-36547181 on Build GRCh38
rs761613672 Chr.19: 36547075-36547075 on Build GRCh38
rs992526533 Chr.19: 36547089-36547089 on Build GRCh38
rs1039828217 Chr.19: 36547230-36547230 on Build GRCh38
rs539319778 Chr.19: 36547108-36547108 on Build GRCh38
rs778629820 Chr.19: 36547182-36547182 on Build GRCh38
rs756569304 Chr.19: 36546878-36546878 on Build GRCh38
rs920106920 Chr.19: 36547157-36547157 on Build GRCh38
rs766951206 Chr.19: 36546883-36546883 on Build GRCh38
rs774347676 Chr.19: 36547255-36547255 on Build GRCh38
rs972898208 Chr.19: 36547149-36547149 on Build GRCh38
rs759955182 Chr.19: 36547100-36547100 on Build GRCh38
rs753818666 Chr.19: 36547296-36547296 on Build GRCh38
rs768319359 Chr.19: 36547066-36547066 on Build GRCh38
rs540776276 Chr.19: 36546909-36546909 on Build GRCh38
rs764403428 Chr.19: 36547225-36547226 on Build GRCh38
rs750019478 Chr.19: 36547206-36547207 on Build GRCh38
rs751768594 Chr.19: 36547150-36547150 on Build GRCh38
rs767041476 Chr.19: 36547173-36547174 on Build GRCh38
rs770801284 Chr.19: 36546818-36546818 on Build GRCh38
rs186457740 Chr.19: 36546899-36546899 on Build GRCh38
rs979905442 Chr.19: 36547048-36547048 on Build GRCh38
rs866070720 Chr.19: 36546935-36546935 on Build GRCh38
rs745843383 Chr.19: 36546938-36546938 on Build GRCh38
rs753238573 Chr.19: 36547109-36547109 on Build GRCh38
rs746182254 Chr.19: 36547209-36547209 on Build GRCh38
rs757025877 Chr.19: 36547045-36547045 on Build GRCh38
rs377579218 Chr.19: 36547270-36547270 on Build GRCh38
rs774123182 Chr.19: 36547067-36547067 on Build GRCh38
rs563607868 Chr.19: 36547161-36547161 on Build GRCh38
rs750697067 Chr.19: 36547279-36547279 on Build GRCh38
rs775298631 Chr.19: 36547197-36547197 on Build GRCh38
rs761386356 Chr.19: 36546932-36546932 on Build GRCh38
rs781481584 Chr.19: 36547166-36547166 on Build GRCh38
rs542011441 Chr.19: 36547058-36547058 on Build GRCh38
rs961938384 Chr.19: 36547137-36547137 on Build GRCh38

View Full Product Details