Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 12214215-12214722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM106B
Entrez Gene ID
54664
Gene Name
transmembrane protein 106B
Gene Aliases
-
UniGene
Hs.396358
Gene Chromosome Location
Chr.7: 12211222-12237264 on Build GRCh38
Transcripts
NM_001134232.1 NM_018374.3 XM_005249789.1

SNP Detail

SNP ID Chromosome Location
rs564186377 Chr.7: 12214581-12214581 on Build GRCh38
rs1040041303 Chr.7: 12214586-12214586 on Build GRCh38
rs370325647 Chr.7: 12214527-12214527 on Build GRCh38
rs1040435978 Chr.7: 12214362-12214362 on Build GRCh38
rs944252685 Chr.7: 12214570-12214570 on Build GRCh38
rs369745081 Chr.7: 12214483-12214483 on Build GRCh38
rs546036789 Chr.7: 12214671-12214671 on Build GRCh38
rs764680543 Chr.7: 12214613-12214613 on Build GRCh38
rs761322268 Chr.7: 12214572-12214572 on Build GRCh38
rs1054159569 Chr.7: 12214658-12214658 on Build GRCh38
rs531416060 Chr.7: 12214284-12214284 on Build GRCh38
rs907949223 Chr.7: 12214360-12214360 on Build GRCh38
rs927428666 Chr.7: 12214393-12214393 on Build GRCh38
rs950296119 Chr.7: 12214370-12214370 on Build GRCh38
rs902368259 Chr.7: 12214281-12214281 on Build GRCh38
rs1020504938 Chr.7: 12214673-12214675 on Build GRCh38
rs6959892 Chr.7: 12214642-12214642 on Build GRCh38
rs760373882 Chr.7: 12214403-12214403 on Build GRCh38
rs568182644 Chr.7: 12214344-12214344 on Build GRCh38
rs533749478 Chr.7: 12214368-12214368 on Build GRCh38
rs189232140 Chr.7: 12214537-12214537 on Build GRCh38
rs62435714 Chr.7: 12214386-12214386 on Build GRCh38
rs1042571595 Chr.7: 12214486-12214486 on Build GRCh38
rs902253325 Chr.7: 12214533-12214533 on Build GRCh38
rs952179563 Chr.7: 12214600-12214600 on Build GRCh38
rs778564900 Chr.7: 12214605-12214606 on Build GRCh38
rs900257536 Chr.7: 12214366-12214366 on Build GRCh38
rs958837882 Chr.7: 12214494-12214494 on Build GRCh38
rs541016309 Chr.7: 12214559-12214559 on Build GRCh38
rs1004875895 Chr.7: 12214602-12214602 on Build GRCh38
rs183483017 Chr.7: 12214280-12214280 on Build GRCh38
rs892866821 Chr.7: 12214661-12214661 on Build GRCh38
rs185689027 Chr.7: 12214542-12214542 on Build GRCh38
rs892940650 Chr.7: 12214392-12214392 on Build GRCh38
rs2043539 Chr.7: 12214254-12214254 on Build GRCh38
rs1018755133 Chr.7: 12214290-12214290 on Build GRCh38
rs540174934 Chr.7: 12214452-12214452 on Build GRCh38
rs1015304315 Chr.7: 12214634-12214634 on Build GRCh38
rs562658233 Chr.7: 12214672-12214672 on Build GRCh38
rs937615068 Chr.7: 12214398-12214398 on Build GRCh38
rs556584304 Chr.7: 12214475-12214475 on Build GRCh38
rs974126016 Chr.7: 12214499-12214499 on Build GRCh38
rs749572159 Chr.7: 12214376-12214376 on Build GRCh38
rs2043538 Chr.7: 12214267-12214267 on Build GRCh38
rs763727757 Chr.7: 12214408-12214408 on Build GRCh38
rs375976215 Chr.7: 12214524-12214524 on Build GRCh38
rs571152574 Chr.7: 12214413-12214413 on Build GRCh38
rs2043540 Chr.7: 12214241-12214241 on Build GRCh38
rs891354438 Chr.7: 12214360-12214363 on Build GRCh38
rs182275337 Chr.7: 12214539-12214539 on Build GRCh38
rs758241344 Chr.7: 12214665-12214665 on Build GRCh38
rs73301022 Chr.7: 12214320-12214320 on Build GRCh38
rs952064474 Chr.7: 12214337-12214337 on Build GRCh38
rs1028082075 Chr.7: 12214329-12214329 on Build GRCh38
rs1020690242 Chr.7: 12214278-12214278 on Build GRCh38
rs971714947 Chr.7: 12214425-12214425 on Build GRCh38
rs974632509 Chr.7: 12214322-12214322 on Build GRCh38

View Full Product Details