Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57098378-57098952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT6
Entrez Gene ID
6778
Gene Name
signal transducer and activator of transcription 6
Gene Aliases
D12S1644, IL-4-STAT, STAT6B, STAT6C
UniGene
Hs.524518
Gene Chromosome Location
Chr.12: 57095404-57111433 on Build GRCh38
Transcripts
NM_001178078.1 NM_001178079.1 NM_001178080.1 NM_001178081.1 NM_003153.4 NR_033659.1 XM_006719575.2 XM_011538703.2 XM_011538704.2 XM_011538705.2 XM_011538706.2 XM_011538707.2 XM_011538708.2 XM_011538709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5920555 c.2066+4A>G p.? Chr.12: 57098788-57098788 on Build GRCh38
4476495 c.2071C>T p.Q691* Chr.12: 57098593-57098593 on Build GRCh38
4832731 c.2006C>G p.S669C Chr.12: 57098852-57098852 on Build GRCh38
1227823 c.1970C>A p.P657H Chr.12: 57098888-57098888 on Build GRCh38
5841577 c.2159+1G>A p.? Chr.12: 57098504-57098504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553935766 Chr.12: 57098918-57098918 on Build GRCh38
rs57453825 Chr.12: 57098602-57098602 on Build GRCh38
rs1057319072 Chr.12: 57098733-57098733 on Build GRCh38
rs750178630 Chr.12: 57098621-57098621 on Build GRCh38
rs143892572 Chr.12: 57098549-57098549 on Build GRCh38
rs777374334 Chr.12: 57098771-57098771 on Build GRCh38
rs751981673 Chr.12: 57098906-57098906 on Build GRCh38
rs182950040 Chr.12: 57098631-57098631 on Build GRCh38
rs200378618 Chr.12: 57098909-57098909 on Build GRCh38
rs868253949 Chr.12: 57098889-57098889 on Build GRCh38
rs187584500 Chr.12: 57098765-57098765 on Build GRCh38
rs747595519 Chr.12: 57098577-57098577 on Build GRCh38
rs565775861 Chr.12: 57098653-57098653 on Build GRCh38
rs936885470 Chr.12: 57098786-57098786 on Build GRCh38
rs377599531 Chr.12: 57098624-57098625 on Build GRCh38
rs761861730 Chr.12: 57098835-57098835 on Build GRCh38
rs371752088 Chr.12: 57098538-57098538 on Build GRCh38
rs777431511 Chr.12: 57098596-57098596 on Build GRCh38
rs775896121 Chr.12: 57098635-57098637 on Build GRCh38
rs761900246 Chr.12: 57098572-57098573 on Build GRCh38
rs763200057 Chr.12: 57098758-57098758 on Build GRCh38
rs529610542 Chr.12: 57098696-57098696 on Build GRCh38
rs745600545 Chr.12: 57098816-57098816 on Build GRCh38
rs765280377 Chr.12: 57098616-57098617 on Build GRCh38
rs760639049 Chr.12: 57098558-57098558 on Build GRCh38
rs771750209 Chr.12: 57098817-57098817 on Build GRCh38
rs934855028 Chr.12: 57098418-57098418 on Build GRCh38
rs757485322 Chr.12: 57098561-57098561 on Build GRCh38
rs776697703 Chr.12: 57098641-57098641 on Build GRCh38
rs759503637 Chr.12: 57098540-57098540 on Build GRCh38
rs745626705 Chr.12: 57098618-57098618 on Build GRCh38
rs878904507 Chr.12: 57098651-57098651 on Build GRCh38
rs1056796023 Chr.12: 57098428-57098428 on Build GRCh38
rs765520388 Chr.12: 57098566-57098566 on Build GRCh38
rs778623543 Chr.12: 57098925-57098925 on Build GRCh38
rs201859128 Chr.12: 57098880-57098880 on Build GRCh38
rs868020316 Chr.12: 57098793-57098793 on Build GRCh38
rs921269621 Chr.12: 57098636-57098636 on Build GRCh38
rs200507162 Chr.12: 57098559-57098559 on Build GRCh38
rs201983882 Chr.12: 57098640-57098640 on Build GRCh38
rs768613456 Chr.12: 57098828-57098828 on Build GRCh38
rs1014836394 Chr.12: 57098655-57098655 on Build GRCh38
rs547163909 Chr.12: 57098643-57098643 on Build GRCh38
rs778676581 Chr.12: 57098814-57098814 on Build GRCh38
rs769398935 Chr.12: 57098490-57098490 on Build GRCh38
rs200879762 Chr.12: 57098564-57098564 on Build GRCh38
rs750819181 Chr.12: 57098569-57098569 on Build GRCh38
rs775545726 Chr.12: 57098554-57098554 on Build GRCh38
rs756822613 Chr.12: 57098919-57098919 on Build GRCh38
rs527765739 Chr.12: 57098856-57098856 on Build GRCh38
rs916946593 Chr.12: 57098746-57098746 on Build GRCh38
rs779913165 Chr.12: 57098820-57098820 on Build GRCh38
rs765469174 Chr.12: 57098646-57098646 on Build GRCh38
rs1051450159 Chr.12: 57098840-57098840 on Build GRCh38
rs775284729 Chr.12: 57098629-57098629 on Build GRCh38
rs537391733 Chr.12: 57098731-57098731 on Build GRCh38
rs372077113 Chr.12: 57098751-57098751 on Build GRCh38
rs766466274 Chr.12: 57098645-57098645 on Build GRCh38
rs368837773 Chr.12: 57098864-57098864 on Build GRCh38
rs570552272 Chr.12: 57098879-57098879 on Build GRCh38
rs770737057 Chr.12: 57098601-57098601 on Build GRCh38
rs113040923 Chr.12: 57098831-57098831 on Build GRCh38
rs768112977 Chr.12: 57098913-57098913 on Build GRCh38
rs990914306 Chr.12: 57098497-57098497 on Build GRCh38
rs995854853 Chr.12: 57098616-57098616 on Build GRCh38
rs978966254 Chr.12: 57098669-57098669 on Build GRCh38
rs374780089 Chr.12: 57098800-57098800 on Build GRCh38
rs866152566 Chr.12: 57098565-57098565 on Build GRCh38
rs138299683 Chr.12: 57098533-57098533 on Build GRCh38
rs898827521 Chr.12: 57098784-57098784 on Build GRCh38
rs923967360 Chr.12: 57098776-57098776 on Build GRCh38
rs758302126 Chr.12: 57098623-57098624 on Build GRCh38
rs758835971 Chr.12: 57098469-57098469 on Build GRCh38
rs12314983 Chr.12: 57098525-57098525 on Build GRCh38
rs748880577 Chr.12: 57098597-57098597 on Build GRCh38
rs922079958 Chr.12: 57098795-57098795 on Build GRCh38
rs780166951 Chr.12: 57098898-57098898 on Build GRCh38
rs200069633 Chr.12: 57098510-57098510 on Build GRCh38
rs776644607 Chr.12: 57098834-57098834 on Build GRCh38
rs756874074 Chr.12: 57098813-57098813 on Build GRCh38
rs959408454 Chr.12: 57098451-57098451 on Build GRCh38
rs771940811 Chr.12: 57098619-57098619 on Build GRCh38
rs367613358 Chr.12: 57098468-57098468 on Build GRCh38
rs147007768 Chr.12: 57098703-57098703 on Build GRCh38
rs758891439 Chr.12: 57098576-57098576 on Build GRCh38
rs147655222 Chr.12: 57098524-57098524 on Build GRCh38
rs774237943 Chr.12: 57098612-57098612 on Build GRCh38
rs994762240 Chr.12: 57098681-57098681 on Build GRCh38
rs201585470 Chr.12: 57098571-57098571 on Build GRCh38
rs766581379 Chr.12: 57098888-57098888 on Build GRCh38
rs780715662 Chr.12: 57098472-57098472 on Build GRCh38
rs747637426 Chr.12: 57098489-57098489 on Build GRCh38
rs145469282 Chr.12: 57098434-57098434 on Build GRCh38
rs766837542 Chr.12: 57098764-57098764 on Build GRCh38
rs755510327 Chr.12: 57098769-57098769 on Build GRCh38
rs565114056 Chr.12: 57098588-57098588 on Build GRCh38
rs200837080 Chr.12: 57098824-57098824 on Build GRCh38
rs899119289 Chr.12: 57098671-57098671 on Build GRCh38
rs763298931 Chr.12: 57098885-57098885 on Build GRCh38

View Full Product Details