Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 16844722-16845315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP7
Entrez Gene ID
5931
Gene Name
RB binding protein 7, chromatin remodeling factor
Gene Aliases
RbAp46
UniGene
Hs.495755
Gene Chromosome Location
Chr.X: 16844650-16870411 on Build GRCh38
Transcripts
NM_001198719.1 NM_002893.3

SNP Detail

SNP ID Chromosome Location
rs930827918 Chr.X: 16844818-16844818 on Build GRCh38
rs966478203 Chr.X: 16845155-16845155 on Build GRCh38
rs928513330 Chr.X: 16845161-16845161 on Build GRCh38
rs141936185 Chr.X: 16845071-16845071 on Build GRCh38
rs746250238 Chr.X: 16845008-16845013 on Build GRCh38
rs763029875 Chr.X: 16845064-16845064 on Build GRCh38
rs201656534 Chr.X: 16844999-16844999 on Build GRCh38
rs780455163 Chr.X: 16845131-16845132 on Build GRCh38
rs907989245 Chr.X: 16844877-16844877 on Build GRCh38
rs758732327 Chr.X: 16845013-16845013 on Build GRCh38
rs750937673 Chr.X: 16844745-16844746 on Build GRCh38
rs773248438 Chr.X: 16845061-16845061 on Build GRCh38
rs371441921 Chr.X: 16845045-16845045 on Build GRCh38
rs181164513 Chr.X: 16844923-16844923 on Build GRCh38
rs138778228 Chr.X: 16845047-16845047 on Build GRCh38
rs952461073 Chr.X: 16845159-16845159 on Build GRCh38
rs200756039 Chr.X: 16845066-16845066 on Build GRCh38
rs966109178 Chr.X: 16845267-16845267 on Build GRCh38
rs140423015 Chr.X: 16845286-16845286 on Build GRCh38
rs747785211 Chr.X: 16844880-16844880 on Build GRCh38
rs938407983 Chr.X: 16845173-16845173 on Build GRCh38
rs911534923 Chr.X: 16844746-16844746 on Build GRCh38
rs199796174 Chr.X: 16844993-16844993 on Build GRCh38
rs779380972 Chr.X: 16845003-16845003 on Build GRCh38
rs11542192 Chr.X: 16844783-16844783 on Build GRCh38
rs756188425 Chr.X: 16844894-16844894 on Build GRCh38
rs199744009 Chr.X: 16844752-16844752 on Build GRCh38
rs769447719 Chr.X: 16844881-16844881 on Build GRCh38
rs999674178 Chr.X: 16845233-16845233 on Build GRCh38
rs375962521 Chr.X: 16845132-16845132 on Build GRCh38
rs900777575 Chr.X: 16844747-16844747 on Build GRCh38
rs189795519 Chr.X: 16844887-16844887 on Build GRCh38
rs760740099 Chr.X: 16845149-16845149 on Build GRCh38
rs1010760609 Chr.X: 16845065-16845065 on Build GRCh38
rs770770607 Chr.X: 16844903-16844903 on Build GRCh38
rs975397381 Chr.X: 16844751-16844751 on Build GRCh38
rs954813960 Chr.X: 16844832-16844833 on Build GRCh38
rs907196786 Chr.X: 16844972-16844972 on Build GRCh38
rs1018650249 Chr.X: 16845238-16845238 on Build GRCh38
rs966352898 Chr.X: 16844749-16844749 on Build GRCh38
rs780450117 Chr.X: 16845024-16845024 on Build GRCh38
rs769493754 Chr.X: 16845060-16845060 on Build GRCh38
rs747677478 Chr.X: 16845050-16845050 on Build GRCh38
rs775802793 Chr.X: 16844851-16844851 on Build GRCh38
rs367866935 Chr.X: 16845012-16845012 on Build GRCh38
rs746597238 Chr.X: 16845030-16845030 on Build GRCh38
rs1030968378 Chr.X: 16844943-16844943 on Build GRCh38
rs780598436 Chr.X: 16844796-16844796 on Build GRCh38
rs770234647 Chr.X: 16845086-16845088 on Build GRCh38

View Full Product Details