Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31120331-31120873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDSN
Entrez Gene ID
1041
Gene Name
corneodesmosin
Gene Aliases
HTSS, HTSS1, HYPT2, PSS, PSS1, S
UniGene
Hs.310958
Gene Chromosome Location
Chr.6: 31115088-31120475 on Build GRCh38
Transcripts
NM_001264.4 NM_014068.2

Gene Symbol
PSORS1C1
Entrez Gene ID
170679
Gene Name
psoriasis susceptibility 1 candidate 1
Gene Aliases
C6orf16, SEEK1
UniGene
Hs.310958
Gene Chromosome Location
Chr.6: 31114831-31140092 on Build GRCh38
Transcripts
NM_001264.4 NM_014068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948978 c.53T>A p.M18K Chr.6: 31120367-31120367 on Build GRCh38
7229991 c.52A>T p.M18L Chr.6: 31120368-31120368 on Build GRCh38
1443219 c.30delG p.R11fs*7 Chr.6: 31120390-31120390 on Build GRCh38
306940 c.9G>A p.S3S Chr.6: 31120411-31120411 on Build GRCh38
7330051 c.85+1G>A p.? Chr.6: 31120334-31120334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753558327 Chr.6: 31120436-31120436 on Build GRCh38
rs1052462010 Chr.6: 31120675-31120675 on Build GRCh38
rs3095318 Chr.6: 31120368-31120368 on Build GRCh38
rs371334209 Chr.6: 31120388-31120388 on Build GRCh38
rs939146576 Chr.6: 31120647-31120647 on Build GRCh38
rs772462110 Chr.6: 31120390-31120390 on Build GRCh38
rs186630661 Chr.6: 31120646-31120646 on Build GRCh38
rs771299175 Chr.6: 31120407-31120407 on Build GRCh38
rs889247144 Chr.6: 31120746-31120746 on Build GRCh38
rs946594904 Chr.6: 31120845-31120845 on Build GRCh38
rs1022054881 Chr.6: 31120494-31120494 on Build GRCh38
rs1008183544 Chr.6: 31120525-31120525 on Build GRCh38
rs954301418 Chr.6: 31120567-31120567 on Build GRCh38
rs371644590 Chr.6: 31120389-31120389 on Build GRCh38
rs755757570 Chr.6: 31120375-31120375 on Build GRCh38
rs1006197363 Chr.6: 31120771-31120771 on Build GRCh38
rs901267715 Chr.6: 31120745-31120745 on Build GRCh38
rs1028439441 Chr.6: 31120761-31120761 on Build GRCh38
rs895031302 Chr.6: 31120668-31120668 on Build GRCh38
rs972446634 Chr.6: 31120800-31120800 on Build GRCh38
rs374928892 Chr.6: 31120412-31120412 on Build GRCh38
rs764702522 Chr.6: 31120447-31120447 on Build GRCh38
rs916415611 Chr.6: 31120841-31120841 on Build GRCh38
rs753363882 Chr.6: 31120377-31120377 on Build GRCh38
rs1028491781 Chr.6: 31120566-31120566 on Build GRCh38
rs2302397 Chr.6: 31120464-31120464 on Build GRCh38
rs747213653 Chr.6: 31120406-31120406 on Build GRCh38
rs754406601 Chr.6: 31120628-31120628 on Build GRCh38
rs750064832 Chr.6: 31120374-31120374 on Build GRCh38
rs544236343 Chr.6: 31120728-31120728 on Build GRCh38
rs1017212823 Chr.6: 31120777-31120777 on Build GRCh38
rs751243454 Chr.6: 31120354-31120354 on Build GRCh38
rs932296324 Chr.6: 31120380-31120380 on Build GRCh38
rs998896272 Chr.6: 31120710-31120710 on Build GRCh38
rs1043688319 Chr.6: 31120686-31120686 on Build GRCh38
rs545243512 Chr.6: 31120465-31120465 on Build GRCh38
rs946618683 Chr.6: 31120677-31120677 on Build GRCh38
rs772997751 Chr.6: 31120467-31120467 on Build GRCh38
rs904558225 Chr.6: 31120509-31120509 on Build GRCh38
rs118054930 Chr.6: 31120835-31120835 on Build GRCh38
rs757486236 Chr.6: 31120454-31120454 on Build GRCh38
rs1048238270 Chr.6: 31120382-31120382 on Build GRCh38
rs567699625 Chr.6: 31120657-31120657 on Build GRCh38
rs927939655 Chr.6: 31120662-31120662 on Build GRCh38
rs2302398 Chr.6: 31120455-31120455 on Build GRCh38
rs776243511 Chr.6: 31120471-31120471 on Build GRCh38
rs753219514 Chr.6: 31120451-31120451 on Build GRCh38
rs2302396 Chr.6: 31120548-31120548 on Build GRCh38
rs984421863 Chr.6: 31120576-31120576 on Build GRCh38
rs754472181 Chr.6: 31120446-31120446 on Build GRCh38
rs909535306 Chr.6: 31120626-31120626 on Build GRCh38
rs778040184 Chr.6: 31120395-31120395 on Build GRCh38
rs369172404 Chr.6: 31120722-31120722 on Build GRCh38
rs200160014 Chr.6: 31120432-31120432 on Build GRCh38
rs778148740 Chr.6: 31120452-31120452 on Build GRCh38
rs774327734 Chr.6: 31120423-31120423 on Build GRCh38
rs760265373 Chr.6: 31120431-31120431 on Build GRCh38
rs766220565 Chr.6: 31120470-31120470 on Build GRCh38
rs939347236 Chr.6: 31120663-31120663 on Build GRCh38
rs990604957 Chr.6: 31120840-31120840 on Build GRCh38
rs961302962 Chr.6: 31120799-31120799 on Build GRCh38
rs1010971414 Chr.6: 31120716-31120716 on Build GRCh38
rs769969765 Chr.6: 31120462-31120462 on Build GRCh38
rs761659956 Chr.6: 31120359-31120359 on Build GRCh38
rs947916329 Chr.6: 31120647-31120647 on Build GRCh38
rs918315942 Chr.6: 31120595-31120595 on Build GRCh38
rs141945415 Chr.6: 31120411-31120411 on Build GRCh38
rs904504659 Chr.6: 31120705-31120705 on Build GRCh38
rs999343746 Chr.6: 31120528-31120528 on Build GRCh38
rs3132546 Chr.6: 31120802-31120802 on Build GRCh38
rs767309535 Chr.6: 31120424-31120424 on Build GRCh38
rs2302399 Chr.6: 31120425-31120425 on Build GRCh38
rs554319664 Chr.6: 31120489-31120489 on Build GRCh38
rs892596130 Chr.6: 31120437-31120437 on Build GRCh38
rs144887769 Chr.6: 31120536-31120536 on Build GRCh38
rs771919596 Chr.6: 31120466-31120466 on Build GRCh38
rs760461777 Chr.6: 31120469-31120469 on Build GRCh38
rs972879567 Chr.6: 31120640-31120640 on Build GRCh38
rs573633731 Chr.6: 31120715-31120715 on Build GRCh38
rs954274483 Chr.6: 31120763-31120763 on Build GRCh38
rs997793805 Chr.6: 31120803-31120803 on Build GRCh38
rs998274032 Chr.6: 31120760-31120760 on Build GRCh38
rs770062672 Chr.6: 31120422-31120422 on Build GRCh38
rs965505192 Chr.6: 31120547-31120547 on Build GRCh38

View Full Product Details