Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7446761-7447286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB1
Entrez Gene ID
1140
Gene Name
cholinergic receptor nicotinic beta 1 subunit
Gene Aliases
ACHRB, CHRNB, CMS1D, CMS2A, CMS2C, SCCMS
UniGene
Hs.330386
Gene Chromosome Location
Chr.17: 7445087-7457613 on Build GRCh38
Transcripts
NM_000747.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5853480 c.280G>A p.E94K Chr.17: 7446869-7446869 on Build GRCh38
4954020 c.340G>T p.V114L Chr.17: 7446929-7446929 on Build GRCh38
6628756 c.420T>A p.R140R Chr.17: 7447109-7447109 on Build GRCh38
4397334 c.462G>A p.Q154Q Chr.17: 7447151-7447151 on Build GRCh38
4396947 c.363G>A p.G121G Chr.17: 7447052-7447052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028687450 Chr.17: 7446891-7446891 on Build GRCh38
rs555170218 Chr.17: 7446859-7446859 on Build GRCh38
rs1054753793 Chr.17: 7447266-7447266 on Build GRCh38
rs369128541 Chr.17: 7447015-7447015 on Build GRCh38
rs747533360 Chr.17: 7447035-7447035 on Build GRCh38
rs761442846 Chr.17: 7447060-7447060 on Build GRCh38
rs750202077 Chr.17: 7446946-7446946 on Build GRCh38
rs775883407 Chr.17: 7446979-7446979 on Build GRCh38
rs2302766 Chr.17: 7447244-7447244 on Build GRCh38
rs762571476 Chr.17: 7447200-7447200 on Build GRCh38
rs774639221 Chr.17: 7446973-7446973 on Build GRCh38
rs376099204 Chr.17: 7446889-7446889 on Build GRCh38
rs750195665 Chr.17: 7446991-7446991 on Build GRCh38
rs769786794 Chr.17: 7446983-7446984 on Build GRCh38
rs758291015 Chr.17: 7446949-7446949 on Build GRCh38
rs748309137 Chr.17: 7446945-7446947 on Build GRCh38
rs755005617 Chr.17: 7446881-7446881 on Build GRCh38
rs953970739 Chr.17: 7446893-7446893 on Build GRCh38
rs368014931 Chr.17: 7446992-7446992 on Build GRCh38
rs753874135 Chr.17: 7447116-7447116 on Build GRCh38
rs764988154 Chr.17: 7446989-7446989 on Build GRCh38
rs936242017 Chr.17: 7447250-7447250 on Build GRCh38
rs779173795 Chr.17: 7447121-7447121 on Build GRCh38
rs950143800 Chr.17: 7447214-7447214 on Build GRCh38
rs897148871 Chr.17: 7447029-7447029 on Build GRCh38
rs776217572 Chr.17: 7447053-7447053 on Build GRCh38
rs376712316 Chr.17: 7446988-7446988 on Build GRCh38
rs201915086 Chr.17: 7446894-7446894 on Build GRCh38
rs778968747 Chr.17: 7446875-7446875 on Build GRCh38
rs370657868 Chr.17: 7446898-7446898 on Build GRCh38
rs776540918 Chr.17: 7446943-7446943 on Build GRCh38
rs752110148 Chr.17: 7446851-7446851 on Build GRCh38
rs998450498 Chr.17: 7447079-7447079 on Build GRCh38
rs745997895 Chr.17: 7446985-7446985 on Build GRCh38
rs75019736 Chr.17: 7446931-7446931 on Build GRCh38
rs766221698 Chr.17: 7447115-7447116 on Build GRCh38
rs763274076 Chr.17: 7446990-7446991 on Build GRCh38
rs745838344 Chr.17: 7446878-7446878 on Build GRCh38
rs1020753062 Chr.17: 7447045-7447045 on Build GRCh38
rs769248655 Chr.17: 7447161-7447161 on Build GRCh38
rs976605479 Chr.17: 7446888-7446888 on Build GRCh38
rs760206481 Chr.17: 7446981-7446981 on Build GRCh38
rs747388705 Chr.17: 7447158-7447158 on Build GRCh38
rs772793418 Chr.17: 7447074-7447074 on Build GRCh38
rs781770923 Chr.17: 7447181-7447181 on Build GRCh38
rs760149212 Chr.17: 7446910-7446910 on Build GRCh38
rs17856698 Chr.17: 7447059-7447059 on Build GRCh38
rs958911348 Chr.17: 7446976-7446992 on Build GRCh38
rs780337044 Chr.17: 7447157-7447157 on Build GRCh38
rs757401724 Chr.17: 7447006-7447006 on Build GRCh38
rs983967827 Chr.17: 7447097-7447097 on Build GRCh38
rs950296052 Chr.17: 7446980-7446980 on Build GRCh38
rs1028135504 Chr.17: 7447087-7447087 on Build GRCh38
rs965264425 Chr.17: 7447191-7447191 on Build GRCh38
rs377462093 Chr.17: 7447088-7447088 on Build GRCh38
rs146117123 Chr.17: 7447095-7447095 on Build GRCh38
rs577977209 Chr.17: 7447002-7447002 on Build GRCh38
rs555518291 Chr.17: 7447098-7447098 on Build GRCh38
rs578069408 Chr.17: 7447080-7447080 on Build GRCh38
rs376365098 Chr.17: 7446871-7446871 on Build GRCh38
rs748183383 Chr.17: 7446892-7446892 on Build GRCh38
rs772367417 Chr.17: 7447017-7447017 on Build GRCh38
rs976624570 Chr.17: 7447201-7447201 on Build GRCh38
rs370702418 Chr.17: 7447096-7447096 on Build GRCh38
rs759413080 Chr.17: 7447103-7447103 on Build GRCh38
rs142549791 Chr.17: 7446902-7446902 on Build GRCh38
rs763734305 Chr.17: 7446983-7446983 on Build GRCh38
rs773503570 Chr.17: 7446819-7446819 on Build GRCh38
rs373764198 Chr.17: 7446870-7446870 on Build GRCh38
rs140086721 Chr.17: 7447108-7447108 on Build GRCh38
rs751491164 Chr.17: 7446997-7446997 on Build GRCh38
rs750646520 Chr.17: 7447129-7447129 on Build GRCh38
rs763342607 Chr.17: 7446835-7446835 on Build GRCh38
rs768266029 Chr.17: 7447037-7447037 on Build GRCh38
rs779033360 Chr.17: 7447010-7447010 on Build GRCh38
rs373219508 Chr.17: 7446996-7446996 on Build GRCh38
rs777940832 Chr.17: 7446964-7446964 on Build GRCh38
rs774997603 Chr.17: 7446916-7446916 on Build GRCh38
rs760133867 Chr.17: 7446919-7446919 on Build GRCh38
rs149433073 Chr.17: 7446929-7446929 on Build GRCh38
rs370010578 Chr.17: 7446802-7446802 on Build GRCh38
rs866965972 Chr.17: 7446903-7446903 on Build GRCh38
rs553745903 Chr.17: 7446991-7446991 on Build GRCh38
rs755732708 Chr.17: 7446860-7446860 on Build GRCh38
rs776841381 Chr.17: 7447242-7447242 on Build GRCh38
rs766823872 Chr.17: 7446837-7446837 on Build GRCh38
rs917329065 Chr.17: 7446933-7446933 on Build GRCh38
rs780576388 Chr.17: 7447022-7447022 on Build GRCh38
rs771326232 Chr.17: 7446901-7446901 on Build GRCh38
rs375214106 Chr.17: 7447148-7447148 on Build GRCh38
rs779891398 Chr.17: 7446962-7446962 on Build GRCh38
rs909856677 Chr.17: 7446928-7446928 on Build GRCh38
rs765188115 Chr.17: 7446945-7446945 on Build GRCh38
rs142983471 Chr.17: 7447093-7447093 on Build GRCh38
rs545335596 Chr.17: 7447005-7447005 on Build GRCh38
rs756198240 Chr.17: 7446963-7446963 on Build GRCh38
rs556323311 Chr.17: 7447000-7447000 on Build GRCh38
rs2302767 Chr.17: 7447225-7447225 on Build GRCh38
rs148584502 Chr.17: 7447062-7447062 on Build GRCh38
rs771273323 Chr.17: 7446965-7446965 on Build GRCh38
rs527663969 Chr.17: 7447185-7447185 on Build GRCh38
rs757349737 Chr.17: 7447118-7447118 on Build GRCh38
rs749632247 Chr.17: 7446900-7446900 on Build GRCh38
rs369771393 Chr.17: 7447199-7447199 on Build GRCh38
rs767921705 Chr.17: 7446915-7446915 on Build GRCh38
rs886053392 Chr.17: 7447040-7447040 on Build GRCh38
rs772591945 Chr.17: 7446978-7446978 on Build GRCh38
rs753646145 Chr.17: 7446867-7446867 on Build GRCh38
rs757084012 Chr.17: 7446932-7446932 on Build GRCh38
rs866659276 Chr.17: 7446984-7446984 on Build GRCh38
rs774348522 Chr.17: 7447190-7447191 on Build GRCh38
rs766155515 Chr.17: 7447090-7447090 on Build GRCh38
rs183537253 Chr.17: 7447194-7447194 on Build GRCh38
rs762779280 Chr.17: 7447075-7447075 on Build GRCh38
rs758939802 Chr.17: 7446856-7446856 on Build GRCh38

View Full Product Details