Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19078875-19079398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ7
Entrez Gene ID
10229
Gene Name
coenzyme Q7, hydroxylase
Gene Aliases
CAT5, CLK-1, CLK1, COQ10D8
UniGene
Hs.157113
Gene Chromosome Location
Chr.16: 19067595-19083097 on Build GRCh38
Transcripts
NM_001190983.1 NM_016138.4

SNP Detail

SNP ID Chromosome Location
rs1035368530 Chr.16: 19078973-19078973 on Build GRCh38
rs564821208 Chr.16: 19079227-19079227 on Build GRCh38
rs1034337615 Chr.16: 19079198-19079198 on Build GRCh38
rs759242262 Chr.16: 19078967-19078967 on Build GRCh38
rs140878310 Chr.16: 19079271-19079271 on Build GRCh38
rs186924963 Chr.16: 19079146-19079146 on Build GRCh38
rs990414023 Chr.16: 19079073-19079073 on Build GRCh38
rs146504690 Chr.16: 19079248-19079248 on Build GRCh38
rs932238150 Chr.16: 19079360-19079360 on Build GRCh38
rs762626817 Chr.16: 19079050-19079050 on Build GRCh38
rs566603880 Chr.16: 19078947-19078947 on Build GRCh38
rs946359942 Chr.16: 19079009-19079009 on Build GRCh38
rs538204851 Chr.16: 19079027-19079027 on Build GRCh38
rs976230967 Chr.16: 19079369-19079369 on Build GRCh38
rs958902970 Chr.16: 19079200-19079200 on Build GRCh38
rs914410413 Chr.16: 19079217-19079217 on Build GRCh38
rs533145902 Chr.16: 19078904-19078904 on Build GRCh38
rs75907431 Chr.16: 19079064-19079064 on Build GRCh38
rs72777500 Chr.16: 19079100-19079100 on Build GRCh38
rs370288419 Chr.16: 19078995-19078995 on Build GRCh38
rs920924653 Chr.16: 19079359-19079359 on Build GRCh38
rs571301498 Chr.16: 19079063-19079063 on Build GRCh38
rs956735464 Chr.16: 19079209-19079209 on Build GRCh38
rs1041193762 Chr.16: 19079066-19079066 on Build GRCh38
rs905254859 Chr.16: 19079172-19079172 on Build GRCh38
rs1023635987 Chr.16: 19078964-19078964 on Build GRCh38
rs900788243 Chr.16: 19079117-19079117 on Build GRCh38
rs781258375 Chr.16: 19079158-19079158 on Build GRCh38
rs556809566 Chr.16: 19079055-19079055 on Build GRCh38
rs528211784 Chr.16: 19078996-19078996 on Build GRCh38
rs892917264 Chr.16: 19078905-19078906 on Build GRCh38
rs772482468 Chr.16: 19079060-19079060 on Build GRCh38
rs937272288 Chr.16: 19079071-19079071 on Build GRCh38
rs914400037 Chr.16: 19079078-19079078 on Build GRCh38
rs756422660 Chr.16: 19079354-19079354 on Build GRCh38
rs983485301 Chr.16: 19078997-19078997 on Build GRCh38
rs554467776 Chr.16: 19079173-19079173 on Build GRCh38
rs1047301655 Chr.16: 19079102-19079102 on Build GRCh38
rs576180149 Chr.16: 19079183-19079183 on Build GRCh38
rs543392701 Chr.16: 19079208-19079208 on Build GRCh38
rs994593037 Chr.16: 19078900-19078900 on Build GRCh38
rs1025674509 Chr.16: 19078917-19078917 on Build GRCh38
rs527746963 Chr.16: 19078949-19078949 on Build GRCh38
rs866229907 Chr.16: 19079299-19079299 on Build GRCh38
rs1041465980 Chr.16: 19079096-19079096 on Build GRCh38
rs559525939 Chr.16: 19078991-19078991 on Build GRCh38
rs1014891257 Chr.16: 19079045-19079045 on Build GRCh38
rs1023691272 Chr.16: 19079157-19079157 on Build GRCh38
rs945801155 Chr.16: 19079083-19079083 on Build GRCh38
rs988449194 Chr.16: 19078992-19078992 on Build GRCh38
rs770596454 Chr.16: 19078926-19078926 on Build GRCh38
rs912346225 Chr.16: 19079301-19079301 on Build GRCh38
rs376500045 Chr.16: 19079144-19079144 on Build GRCh38
rs764785904 Chr.16: 19078969-19078969 on Build GRCh38
rs985764709 Chr.16: 19079314-19079314 on Build GRCh38
rs73528967 Chr.16: 19078960-19078960 on Build GRCh38
rs141953305 Chr.16: 19079134-19079134 on Build GRCh38
rs560075062 Chr.16: 19079308-19079308 on Build GRCh38
rs887921623 Chr.16: 19079138-19079138 on Build GRCh38
rs943764685 Chr.16: 19079311-19079311 on Build GRCh38
rs1049293556 Chr.16: 19079370-19079370 on Build GRCh38
rs1049723356 Chr.16: 19079138-19079139 on Build GRCh38
rs551492362 Chr.16: 19078924-19078924 on Build GRCh38
rs764071448 Chr.16: 19079098-19079098 on Build GRCh38
rs1009173976 Chr.16: 19078943-19078943 on Build GRCh38
rs140482392 Chr.16: 19079109-19079109 on Build GRCh38
rs910321474 Chr.16: 19079331-19079331 on Build GRCh38
rs956671513 Chr.16: 19078976-19078976 on Build GRCh38
rs1022179689 Chr.16: 19079285-19079285 on Build GRCh38

View Full Product Details