Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7226282-7226776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL2
Entrez Gene ID
1856
Gene Name
dishevelled segment polarity protein 2
Gene Aliases
-
UniGene
Hs.118640
Gene Chromosome Location
Chr.17: 7225342-7234548 on Build GRCh38
Transcripts
NM_004422.2 XM_005256502.1 XM_017024298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6016442 c.1616T>C p.L539P Chr.17: 7226567-7226567 on Build GRCh38
6650467 c.1624G>A p.A542T Chr.17: 7226559-7226559 on Build GRCh38
4769108 c.1724C>A p.T575N Chr.17: 7226459-7226459 on Build GRCh38
1242211 c.1632C>A p.P544P Chr.17: 7226551-7226551 on Build GRCh38
3691765 c.1685G>A p.S562N Chr.17: 7226498-7226498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054177258 Chr.17: 7226735-7226735 on Build GRCh38
rs972749353 Chr.17: 7226716-7226716 on Build GRCh38
rs2074222 Chr.17: 7226655-7226655 on Build GRCh38
rs371112172 Chr.17: 7226426-7226426 on Build GRCh38
rs770057085 Chr.17: 7226470-7226470 on Build GRCh38
rs762111480 Chr.17: 7226378-7226378 on Build GRCh38
rs35594616 Chr.17: 7226521-7226521 on Build GRCh38
rs372921737 Chr.17: 7226357-7226357 on Build GRCh38
rs766840095 Chr.17: 7226403-7226403 on Build GRCh38
rs140545048 Chr.17: 7226702-7226702 on Build GRCh38
rs985536911 Chr.17: 7226700-7226700 on Build GRCh38
rs758400019 Chr.17: 7226537-7226537 on Build GRCh38
rs759057007 Chr.17: 7226504-7226504 on Build GRCh38
rs918547537 Chr.17: 7226719-7226719 on Build GRCh38
rs756324626 Chr.17: 7226551-7226551 on Build GRCh38
rs747210791 Chr.17: 7226325-7226325 on Build GRCh38
rs773513204 Chr.17: 7226381-7226381 on Build GRCh38
rs746276617 Chr.17: 7226682-7226682 on Build GRCh38
rs867656789 Chr.17: 7226722-7226722 on Build GRCh38
rs796500594 Chr.17: 7226544-7226544 on Build GRCh38
rs777885516 Chr.17: 7226552-7226552 on Build GRCh38
rs752499366 Chr.17: 7226308-7226308 on Build GRCh38
rs565161291 Chr.17: 7226525-7226525 on Build GRCh38
rs777036769 Chr.17: 7226643-7226644 on Build GRCh38
rs752409376 Chr.17: 7226415-7226415 on Build GRCh38
rs765167402 Chr.17: 7226529-7226529 on Build GRCh38
rs756165808 Chr.17: 7226656-7226656 on Build GRCh38
rs755993922 Chr.17: 7226321-7226321 on Build GRCh38
rs772667459 Chr.17: 7226684-7226684 on Build GRCh38
rs941356299 Chr.17: 7226428-7226428 on Build GRCh38
rs761606568 Chr.17: 7226611-7226611 on Build GRCh38
rs377522537 Chr.17: 7226405-7226405 on Build GRCh38
rs371737565 Chr.17: 7226330-7226330 on Build GRCh38
rs138493643 Chr.17: 7226427-7226427 on Build GRCh38
rs186932260 Chr.17: 7226538-7226538 on Build GRCh38
rs774854172 Chr.17: 7226574-7226574 on Build GRCh38
rs567571945 Chr.17: 7226587-7226587 on Build GRCh38
rs201803378 Chr.17: 7226478-7226478 on Build GRCh38
rs757188541 Chr.17: 7226444-7226444 on Build GRCh38
rs751086029 Chr.17: 7226304-7226304 on Build GRCh38
rs779400293 Chr.17: 7226679-7226679 on Build GRCh38
rs375432216 Chr.17: 7226346-7226346 on Build GRCh38
rs528578173 Chr.17: 7226506-7226506 on Build GRCh38
rs750368235 Chr.17: 7226535-7226535 on Build GRCh38
rs774742299 Chr.17: 7226495-7226495 on Build GRCh38
rs760382893 Chr.17: 7226512-7226512 on Build GRCh38
rs754651383 Chr.17: 7226307-7226307 on Build GRCh38
rs911550351 Chr.17: 7226657-7226657 on Build GRCh38
rs377013969 Chr.17: 7226545-7226545 on Build GRCh38
rs773601337 Chr.17: 7226477-7226477 on Build GRCh38
rs771212668 Chr.17: 7226571-7226571 on Build GRCh38
rs778747234 Chr.17: 7226451-7226451 on Build GRCh38
rs373873616 Chr.17: 7226358-7226358 on Build GRCh38
rs781328615 Chr.17: 7226466-7226466 on Build GRCh38
rs771429420 Chr.17: 7226489-7226489 on Build GRCh38
rs766341126 Chr.17: 7226652-7226652 on Build GRCh38
rs954190070 Chr.17: 7226690-7226690 on Build GRCh38
rs762608292 Chr.17: 7226743-7226743 on Build GRCh38
rs373332227 Chr.17: 7226461-7226461 on Build GRCh38
rs750278603 Chr.17: 7226628-7226628 on Build GRCh38
rs973083666 Chr.17: 7226456-7226456 on Build GRCh38
rs902996625 Chr.17: 7226370-7226371 on Build GRCh38
rs370320396 Chr.17: 7226467-7226467 on Build GRCh38
rs374057583 Chr.17: 7226673-7226673 on Build GRCh38
rs763500518 Chr.17: 7226392-7226392 on Build GRCh38
rs755903914 Chr.17: 7226421-7226421 on Build GRCh38
rs764939670 Chr.17: 7226620-7226620 on Build GRCh38
rs72839768 Chr.17: 7226579-7226579 on Build GRCh38
rs753626462 Chr.17: 7226431-7226431 on Build GRCh38
rs760294769 Chr.17: 7226585-7226585 on Build GRCh38
rs747066639 Chr.17: 7226459-7226459 on Build GRCh38
rs200484159 Chr.17: 7226349-7226349 on Build GRCh38
rs566656706 Chr.17: 7226712-7226712 on Build GRCh38
rs776165646 Chr.17: 7226593-7226593 on Build GRCh38
rs139855538 Chr.17: 7226583-7226583 on Build GRCh38
rs777993321 Chr.17: 7226661-7226661 on Build GRCh38
rs375513655 Chr.17: 7226380-7226380 on Build GRCh38
rs963072186 Chr.17: 7226709-7226709 on Build GRCh38
rs867174928 Chr.17: 7226663-7226663 on Build GRCh38
rs867386566 Chr.17: 7226653-7226653 on Build GRCh38
rs763694344 Chr.17: 7226516-7226516 on Build GRCh38
rs998811978 Chr.17: 7226658-7226658 on Build GRCh38
rs768740247 Chr.17: 7226465-7226465 on Build GRCh38
rs749474221 Chr.17: 7226664-7226664 on Build GRCh38
rs200099877 Chr.17: 7226407-7226407 on Build GRCh38
rs145634625 Chr.17: 7226736-7226736 on Build GRCh38
rs770832523 Chr.17: 7226333-7226333 on Build GRCh38
rs767139067 Chr.17: 7226505-7226505 on Build GRCh38
rs749615262 Chr.17: 7226563-7226563 on Build GRCh38
rs370419589 Chr.17: 7226494-7226494 on Build GRCh38
rs758307598 Chr.17: 7226643-7226643 on Build GRCh38

View Full Product Details