Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41508440-41508992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP3
Entrez Gene ID
59269
Gene Name
human immunodeficiency virus type I enhancer binding protein 3
Gene Aliases
KBP-1, KBP1, KRC, SHN3, Schnurri-3, ZAS3, ZNF40C
UniGene
Hs.403972
Gene Chromosome Location
Chr.1: 41506365-42035925 on Build GRCh38
Transcripts
NM_001127714.2 NM_024503.4 XM_011541884.2 XM_017001992.1 XM_017001993.1 XM_017001994.1

SNP Detail

SNP ID Chromosome Location
rs375109999 Chr.1: 41508798-41508798 on Build GRCh38
rs978501975 Chr.1: 41508912-41508912 on Build GRCh38
rs545656450 Chr.1: 41508712-41508712 on Build GRCh38
rs11586958 Chr.1: 41508493-41508493 on Build GRCh38
rs2151674 Chr.1: 41508587-41508587 on Build GRCh38
rs772390867 Chr.1: 41508870-41508870 on Build GRCh38
rs28496731 Chr.1: 41508934-41508934 on Build GRCh38
rs145544548 Chr.1: 41508842-41508842 on Build GRCh38
rs530109422 Chr.1: 41508872-41508872 on Build GRCh38
rs764234907 Chr.1: 41508660-41508660 on Build GRCh38
rs1007440782 Chr.1: 41508519-41508519 on Build GRCh38
rs114064779 Chr.1: 41508524-41508524 on Build GRCh38
rs12076179 Chr.1: 41508813-41508813 on Build GRCh38
rs902909902 Chr.1: 41508832-41508832 on Build GRCh38
rs542926639 Chr.1: 41508684-41508684 on Build GRCh38
rs147530329 Chr.1: 41508565-41508565 on Build GRCh38
rs188296749 Chr.1: 41508894-41508894 on Build GRCh38
rs991731076 Chr.1: 41508727-41508727 on Build GRCh38
rs150655963 Chr.1: 41508967-41508967 on Build GRCh38
rs193195885 Chr.1: 41508581-41508581 on Build GRCh38
rs967051213 Chr.1: 41508627-41508627 on Build GRCh38
rs1054093760 Chr.1: 41508844-41508844 on Build GRCh38
rs968315832 Chr.1: 41508888-41508888 on Build GRCh38
rs183682179 Chr.1: 41508869-41508869 on Build GRCh38
rs572147690 Chr.1: 41508680-41508680 on Build GRCh38
rs1007221403 Chr.1: 41508878-41508878 on Build GRCh38
rs1047804741 Chr.1: 41508830-41508830 on Build GRCh38
rs951456361 Chr.1: 41508951-41508951 on Build GRCh38
rs971897501 Chr.1: 41508568-41508568 on Build GRCh38
rs917543787 Chr.1: 41508750-41508750 on Build GRCh38
rs548965969 Chr.1: 41508880-41508880 on Build GRCh38
rs1018801901 Chr.1: 41508879-41508879 on Build GRCh38
rs899154980 Chr.1: 41508815-41508815 on Build GRCh38
rs955970649 Chr.1: 41508960-41508960 on Build GRCh38
rs901799180 Chr.1: 41508889-41508889 on Build GRCh38
rs538054811 Chr.1: 41508547-41508547 on Build GRCh38
rs975130937 Chr.1: 41508512-41508512 on Build GRCh38
rs191367229 Chr.1: 41508691-41508691 on Build GRCh38
rs917401723 Chr.1: 41508964-41508964 on Build GRCh38
rs182992633 Chr.1: 41508664-41508664 on Build GRCh38
rs993313804 Chr.1: 41508895-41508895 on Build GRCh38
rs187280237 Chr.1: 41508690-41508690 on Build GRCh38
rs193017768 Chr.1: 41508963-41508963 on Build GRCh38
rs991510832 Chr.1: 41508968-41508968 on Build GRCh38
rs185844850 Chr.1: 41508965-41508965 on Build GRCh38
rs28512399 Chr.1: 41508677-41508677 on Build GRCh38
rs965850578 Chr.1: 41508535-41508535 on Build GRCh38
rs549274555 Chr.1: 41508505-41508505 on Build GRCh38
rs565373583 Chr.1: 41508797-41508797 on Build GRCh38
rs950179254 Chr.1: 41508810-41508810 on Build GRCh38
rs749335981 Chr.1: 41508743-41508743 on Build GRCh38

View Full Product Details