Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33084935-33085517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DPB1
Entrez Gene ID
3115
Gene Name
major histocompatibility complex, class II, DP beta 1
Gene Aliases
DPB1, HLA-DP, HLA-DP1B, HLA-DPB
UniGene
Hs.485130
Gene Chromosome Location
Chr.6: 33075926-33089696 on Build GRCh38
Transcripts
NM_002121.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211646 c.410T>G p.L137R Chr.6: 33085064-33085064 on Build GRCh38
4354540 c.389G>C p.S130T Chr.6: 33085043-33085043 on Build GRCh38
6106854 c.495G>T p.L165L Chr.6: 33085149-33085149 on Build GRCh38
1195852 c.458G>C p.S153T Chr.6: 33085043-33085043 on Build GRCh38
4338259 c.503delC p.Q170fs*35 Chr.6: 33085157-33085157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs9277462 Chr.6: 33085494-33085494 on Build GRCh38
rs61736935 Chr.6: 33085040-33085040 on Build GRCh38
rs751221886 Chr.6: 33085094-33085094 on Build GRCh38
rs140447065 Chr.6: 33085043-33085043 on Build GRCh38
rs912128450 Chr.6: 33084970-33084971 on Build GRCh38
rs1071597 Chr.6: 33085209-33085209 on Build GRCh38
rs894270562 Chr.6: 33085128-33085128 on Build GRCh38
rs969428264 Chr.6: 33085485-33085503 on Build GRCh38
rs1042335 Chr.6: 33085181-33085181 on Build GRCh38
rs750357586 Chr.6: 33085116-33085116 on Build GRCh38
rs748284821 Chr.6: 33085149-33085149 on Build GRCh38
rs755847836 Chr.6: 33085231-33085231 on Build GRCh38
rs370864722 Chr.6: 33085200-33085200 on Build GRCh38
rs773409536 Chr.6: 33085054-33085054 on Build GRCh38
rs61736936 Chr.6: 33085016-33085016 on Build GRCh38
rs760040959 Chr.6: 33085068-33085068 on Build GRCh38
rs948601779 Chr.6: 33085459-33085459 on Build GRCh38
rs9277458 Chr.6: 33085390-33085390 on Build GRCh38
rs747228585 Chr.6: 33085259-33085259 on Build GRCh38
rs986312685 Chr.6: 33085324-33085324 on Build GRCh38
rs756011172 Chr.6: 33085118-33085118 on Build GRCh38
rs372905365 Chr.6: 33085089-33085089 on Build GRCh38
rs144181653 Chr.6: 33085171-33085171 on Build GRCh38
rs80315764 Chr.6: 33085344-33085344 on Build GRCh38
rs759617109 Chr.6: 33085372-33085372 on Build GRCh38
rs143269376 Chr.6: 33085400-33085400 on Build GRCh38
rs879761623 Chr.6: 33085493-33085494 on Build GRCh38
rs764687991 Chr.6: 33085072-33085072 on Build GRCh38
rs553069394 Chr.6: 33085479-33085479 on Build GRCh38
rs879025225 Chr.6: 33085364-33085364 on Build GRCh38
rs772872005 Chr.6: 33085182-33085182 on Build GRCh38
rs527862621 Chr.6: 33085443-33085443 on Build GRCh38
rs757047904 Chr.6: 33085111-33085111 on Build GRCh38
rs747234343 Chr.6: 33085162-33085162 on Build GRCh38
rs1003152165 Chr.6: 33085357-33085357 on Build GRCh38
rs750173090 Chr.6: 33085222-33085222 on Build GRCh38
rs751346150 Chr.6: 33085002-33085002 on Build GRCh38
rs539695276 Chr.6: 33085248-33085248 on Build GRCh38
rs758087295 Chr.6: 33084983-33084983 on Build GRCh38
rs9277459 Chr.6: 33085471-33085471 on Build GRCh38
rs1031689463 Chr.6: 33085423-33085423 on Build GRCh38
rs142567249 Chr.6: 33085303-33085303 on Build GRCh38
rs780989395 Chr.6: 33085112-33085112 on Build GRCh38
rs41559424 Chr.6: 33085221-33085221 on Build GRCh38
rs764783267 Chr.6: 33084968-33084968 on Build GRCh38
rs14362 Chr.6: 33085204-33085204 on Build GRCh38
rs534162897 Chr.6: 33085090-33085090 on Build GRCh38
rs778977608 Chr.6: 33085146-33085146 on Build GRCh38
rs183308747 Chr.6: 33085411-33085411 on Build GRCh38
rs1042331 Chr.6: 33085173-33085173 on Build GRCh38
rs549282061 Chr.6: 33085446-33085446 on Build GRCh38
rs781427144 Chr.6: 33085277-33085277 on Build GRCh38
rs34802875 Chr.6: 33085416-33085416 on Build GRCh38
rs9277460 Chr.6: 33085475-33085475 on Build GRCh38
rs368841277 Chr.6: 33085055-33085055 on Build GRCh38
rs562166558 Chr.6: 33085025-33085025 on Build GRCh38
rs770371240 Chr.6: 33085174-33085174 on Build GRCh38
rs9277457 Chr.6: 33085249-33085249 on Build GRCh38
rs762588336 Chr.6: 33085185-33085185 on Build GRCh38
rs200465564 Chr.6: 33085493-33085494 on Build GRCh38
rs61736937 Chr.6: 33084986-33084986 on Build GRCh38
rs376906908 Chr.6: 33085137-33085137 on Build GRCh38
rs566678973 Chr.6: 33085075-33085075 on Build GRCh38
rs772135792 Chr.6: 33085158-33085158 on Build GRCh38
rs754366191 Chr.6: 33084960-33084960 on Build GRCh38
rs757067798 Chr.6: 33085008-33085008 on Build GRCh38
rs778922500 Chr.6: 33085037-33085037 on Build GRCh38
rs1003872854 Chr.6: 33084992-33084992 on Build GRCh38
rs780393867 Chr.6: 33085157-33085157 on Build GRCh38
rs75656943 Chr.6: 33085315-33085315 on Build GRCh38
rs771222927 Chr.6: 33085168-33085168 on Build GRCh38
rs529551304 Chr.6: 33085420-33085420 on Build GRCh38
rs9277461 Chr.6: 33085493-33085493 on Build GRCh38
rs752423599 Chr.6: 33084978-33084978 on Build GRCh38
rs778011110 Chr.6: 33085159-33085159 on Build GRCh38
rs61736938 Chr.6: 33085142-33085142 on Build GRCh38
rs1003099995 Chr.6: 33085028-33085028 on Build GRCh38
rs772223745 Chr.6: 33085053-33085053 on Build GRCh38
rs386699882 Chr.6: 33085493-33085494 on Build GRCh38
rs766369841 Chr.6: 33085233-33085233 on Build GRCh38
rs1045563293 Chr.6: 33085461-33085461 on Build GRCh38
rs61736934 Chr.6: 33085059-33085059 on Build GRCh38
rs1048124057 Chr.6: 33084963-33084963 on Build GRCh38
rs757437801 Chr.6: 33085273-33085273 on Build GRCh38
rs1042212 Chr.6: 33085026-33085026 on Build GRCh38
rs907128816 Chr.6: 33085467-33085467 on Build GRCh38

View Full Product Details