Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136224813-136225322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP7D3
Entrez Gene ID
79649
Gene Name
MAP7 domain containing 3
Gene Aliases
MDP3
UniGene
Hs.446275
Gene Chromosome Location
Chr.X: 136213220-136256482 on Build GRCh38
Transcripts
NM_001173516.1 NM_001173517.1 NM_024597.3 XM_005262472.1 XM_017029843.1

SNP Detail

SNP ID Chromosome Location
rs574941368 Chr.X: 136224904-136224904 on Build GRCh38
rs756504007 Chr.X: 136225036-136225036 on Build GRCh38
rs144908649 Chr.X: 136225150-136225150 on Build GRCh38
rs780430699 Chr.X: 136225231-136225231 on Build GRCh38
rs140985130 Chr.X: 136225091-136225091 on Build GRCh38
rs201140656 Chr.X: 136224877-136224877 on Build GRCh38
rs1042188356 Chr.X: 136224983-136224983 on Build GRCh38
rs377132183 Chr.X: 136224840-136224840 on Build GRCh38
rs751705386 Chr.X: 136224947-136224947 on Build GRCh38
rs774025753 Chr.X: 136224853-136224853 on Build GRCh38
rs779424942 Chr.X: 136225185-136225185 on Build GRCh38
rs1026169415 Chr.X: 136225206-136225206 on Build GRCh38
rs936850943 Chr.X: 136225009-136225009 on Build GRCh38
rs926757899 Chr.X: 136224956-136224956 on Build GRCh38
rs369314225 Chr.X: 136225276-136225276 on Build GRCh38
rs191340545 Chr.X: 136225027-136225027 on Build GRCh38
rs747107771 Chr.X: 136225248-136225248 on Build GRCh38
rs199755641 Chr.X: 136224860-136224860 on Build GRCh38
rs781503446 Chr.X: 136225007-136225007 on Build GRCh38
rs949098959 Chr.X: 136224916-136224916 on Build GRCh38
rs916961743 Chr.X: 136225076-136225076 on Build GRCh38
rs1007129969 Chr.X: 136225095-136225095 on Build GRCh38
rs902924263 Chr.X: 136225165-136225165 on Build GRCh38
rs376639097 Chr.X: 136224928-136224928 on Build GRCh38
rs761170815 Chr.X: 136224874-136224874 on Build GRCh38
rs765697551 Chr.X: 136224875-136224876 on Build GRCh38
rs999877699 Chr.X: 136225184-136225184 on Build GRCh38
rs778300824 Chr.X: 136225069-136225069 on Build GRCh38
rs767040493 Chr.X: 136224859-136224859 on Build GRCh38
rs938286919 Chr.X: 136224974-136224974 on Build GRCh38
rs5929742 Chr.X: 136224890-136224890 on Build GRCh38
rs186237042 Chr.X: 136224991-136224991 on Build GRCh38
rs949714093 Chr.X: 136225090-136225090 on Build GRCh38
rs996778340 Chr.X: 136225249-136225249 on Build GRCh38
rs758882401 Chr.X: 136225146-136225146 on Build GRCh38
rs761282315 Chr.X: 136224858-136224858 on Build GRCh38

View Full Product Details