Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141202891-141203417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB11
Entrez Gene ID
56125
Gene Name
protocadherin beta 11
Gene Aliases
ME2, PCDH-BETA11
UniGene
Hs.283084
Gene Chromosome Location
Chr.5: 141199775-141203045 on Build GRCh38
Transcripts
NM_018931.2

SNP Detail

SNP ID Chromosome Location
rs956476147 Chr.5: 141203338-141203338 on Build GRCh38
rs533978807 Chr.5: 141203311-141203311 on Build GRCh38
rs781880320 Chr.5: 141203327-141203327 on Build GRCh38
rs372483088 Chr.5: 141203064-141203064 on Build GRCh38
rs986580206 Chr.5: 141203379-141203379 on Build GRCh38
rs928834622 Chr.5: 141203018-141203018 on Build GRCh38
rs547324190 Chr.5: 141203334-141203334 on Build GRCh38
rs1043433599 Chr.5: 141203115-141203115 on Build GRCh38
rs981616855 Chr.5: 141203003-141203003 on Build GRCh38
rs1035899330 Chr.5: 141203322-141203322 on Build GRCh38
rs556054445 Chr.5: 141203377-141203377 on Build GRCh38
rs528901664 Chr.5: 141203061-141203061 on Build GRCh38
rs1022313672 Chr.5: 141203356-141203356 on Build GRCh38
rs782131825 Chr.5: 141203098-141203098 on Build GRCh38
rs185619642 Chr.5: 141202939-141202939 on Build GRCh38
rs782714387 Chr.5: 141203261-141203261 on Build GRCh38
rs1023072954 Chr.5: 141202964-141202964 on Build GRCh38
rs190036085 Chr.5: 141203114-141203114 on Build GRCh38
rs562431076 Chr.5: 141203168-141203168 on Build GRCh38
rs995519348 Chr.5: 141203247-141203247 on Build GRCh38
rs201791995 Chr.5: 141203360-141203360 on Build GRCh38
rs1004618848 Chr.5: 141202920-141202920 on Build GRCh38
rs1037419330 Chr.5: 141203214-141203214 on Build GRCh38
rs970186556 Chr.5: 141202996-141202996 on Build GRCh38
rs921009810 Chr.5: 141203375-141203375 on Build GRCh38
rs913597867 Chr.5: 141203100-141203100 on Build GRCh38
rs572259695 Chr.5: 141202951-141202951 on Build GRCh38
rs956358195 Chr.5: 141203031-141203031 on Build GRCh38
rs1023023792 Chr.5: 141203263-141203263 on Build GRCh38
rs545068655 Chr.5: 141202917-141202917 on Build GRCh38
rs112928015 Chr.5: 141203173-141203173 on Build GRCh38
rs530352659 Chr.5: 141203154-141203154 on Build GRCh38
rs912418911 Chr.5: 141203386-141203386 on Build GRCh38
rs995612697 Chr.5: 141202955-141202955 on Build GRCh38
rs561621286 Chr.5: 141203143-141203143 on Build GRCh38
rs989100490 Chr.5: 141203345-141203345 on Build GRCh38
rs182541044 Chr.5: 141203367-141203367 on Build GRCh38
rs1003041251 Chr.5: 141203321-141203321 on Build GRCh38
rs979068952 Chr.5: 141203370-141203370 on Build GRCh38
rs112044047 Chr.5: 141203245-141203245 on Build GRCh38
rs782740736 Chr.5: 141203142-141203142 on Build GRCh38
rs939865986 Chr.5: 141203390-141203390 on Build GRCh38
rs113034719 Chr.5: 141203164-141203164 on Build GRCh38
rs571373208 Chr.5: 141203299-141203299 on Build GRCh38
rs932446932 Chr.5: 141203161-141203161 on Build GRCh38
rs560572871 Chr.5: 141203103-141203104 on Build GRCh38
rs963103272 Chr.5: 141202941-141202941 on Build GRCh38

View Full Product Details