Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 20177239-20177781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXDC2
Entrez Gene ID
84898
Gene Name
plexin domain containing 2
Gene Aliases
TEM7R
UniGene
Hs.658134
Gene Chromosome Location
Chr.10: 19816215-20289856 on Build GRCh38
Transcripts
NM_001282736.1 NM_032812.8 XM_011519750.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6517727 c.984C>A p.C328* Chr.10: 20177332-20177332 on Build GRCh38
3866963 c.1016C>T p.S339F Chr.10: 20177364-20177364 on Build GRCh38
235218 c.984C>T p.C328C Chr.10: 20177332-20177332 on Build GRCh38
6896433 c.1001G>C p.C334S Chr.10: 20177349-20177349 on Build GRCh38
916937 c.1061+2T>C p.? Chr.10: 20177411-20177411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981228676 Chr.10: 20177426-20177426 on Build GRCh38
rs763613047 Chr.10: 20177298-20177298 on Build GRCh38
rs537901904 Chr.10: 20177630-20177630 on Build GRCh38
rs548825737 Chr.10: 20177580-20177580 on Build GRCh38
rs770448367 Chr.10: 20177281-20177281 on Build GRCh38
rs1046059201 Chr.10: 20177485-20177485 on Build GRCh38
rs940561440 Chr.10: 20177463-20177463 on Build GRCh38
rs143248975 Chr.10: 20177566-20177566 on Build GRCh38
rs984220831 Chr.10: 20177687-20177687 on Build GRCh38
rs1036301220 Chr.10: 20177471-20177471 on Build GRCh38
rs926718296 Chr.10: 20177433-20177433 on Build GRCh38
rs200191125 Chr.10: 20177301-20177301 on Build GRCh38
rs745631807 Chr.10: 20177442-20177442 on Build GRCh38
rs756578181 Chr.10: 20177312-20177312 on Build GRCh38
rs766797332 Chr.10: 20177317-20177317 on Build GRCh38
rs779075601 Chr.10: 20177372-20177372 on Build GRCh38
rs142318150 Chr.10: 20177552-20177552 on Build GRCh38
rs954659959 Chr.10: 20177591-20177591 on Build GRCh38
rs747354180 Chr.10: 20177346-20177346 on Build GRCh38
rs776057528 Chr.10: 20177375-20177375 on Build GRCh38
rs552607149 Chr.10: 20177586-20177586 on Build GRCh38
rs74972937 Chr.10: 20177641-20177641 on Build GRCh38
rs764965476 Chr.10: 20177284-20177284 on Build GRCh38
rs571918739 Chr.10: 20177749-20177749 on Build GRCh38
rs750909838 Chr.10: 20177309-20177309 on Build GRCh38
rs1000599646 Chr.10: 20177569-20177569 on Build GRCh38
rs938908868 Chr.10: 20177505-20177505 on Build GRCh38
rs904840193 Chr.10: 20177592-20177592 on Build GRCh38
rs781187184 Chr.10: 20177441-20177441 on Build GRCh38
rs919012473 Chr.10: 20177685-20177685 on Build GRCh38
rs398074996 Chr.10: 20177759-20177759 on Build GRCh38
rs986048956 Chr.10: 20177618-20177618 on Build GRCh38
rs541664049 Chr.10: 20177272-20177272 on Build GRCh38
rs193176905 Chr.10: 20177584-20177584 on Build GRCh38
rs79921091 Chr.10: 20177759-20177760 on Build GRCh38
rs34150429 Chr.10: 20177683-20177683 on Build GRCh38
rs777849258 Chr.10: 20177423-20177423 on Build GRCh38
rs762358183 Chr.10: 20177696-20177696 on Build GRCh38
rs754188191 Chr.10: 20177352-20177352 on Build GRCh38
rs781511314 Chr.10: 20177426-20177426 on Build GRCh38
rs553814706 Chr.10: 20177729-20177729 on Build GRCh38
rs772406318 Chr.10: 20177364-20177364 on Build GRCh38
rs762404378 Chr.10: 20177291-20177291 on Build GRCh38
rs563744336 Chr.10: 20177570-20177570 on Build GRCh38
rs988365177 Chr.10: 20177349-20177349 on Build GRCh38
rs746922053 Chr.10: 20177428-20177428 on Build GRCh38
rs559900603 Chr.10: 20177277-20177277 on Build GRCh38
rs927602720 Chr.10: 20177754-20177754 on Build GRCh38
rs10764210 Chr.10: 20177590-20177590 on Build GRCh38
rs771326272 Chr.10: 20177418-20177420 on Build GRCh38
rs770984632 Chr.10: 20177431-20177431 on Build GRCh38
rs55837147 Chr.10: 20177752-20177752 on Build GRCh38
rs147644062 Chr.10: 20177445-20177445 on Build GRCh38
rs775184355 Chr.10: 20177287-20177287 on Build GRCh38
rs10827983 Chr.10: 20177534-20177534 on Build GRCh38
rs377043785 Chr.10: 20177643-20177643 on Build GRCh38
rs929037590 Chr.10: 20177695-20177695 on Build GRCh38
rs372437836 Chr.10: 20177417-20177417 on Build GRCh38
rs567088869 Chr.10: 20177582-20177582 on Build GRCh38
rs970921314 Chr.10: 20177397-20177397 on Build GRCh38
rs564365115 Chr.10: 20177451-20177451 on Build GRCh38
rs981808402 Chr.10: 20177747-20177747 on Build GRCh38
rs755242147 Chr.10: 20177370-20177370 on Build GRCh38
rs908575202 Chr.10: 20177450-20177450 on Build GRCh38
rs941800796 Chr.10: 20177416-20177416 on Build GRCh38
rs907493817 Chr.10: 20177501-20177501 on Build GRCh38
rs908522632 Chr.10: 20177716-20177716 on Build GRCh38

View Full Product Details