Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 6917162-6917661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF144A
Entrez Gene ID
9781
Gene Name
ring finger protein 144A
Gene Aliases
RNF144, UBCE7IP4
UniGene
Hs.22146
Gene Chromosome Location
Chr.2: 6917392-7076886 on Build GRCh38
Transcripts
NM_014746.4 XM_017005398.1 XM_017005399.1 XM_017005400.1 XM_017005401.1 XM_017005402.1 XM_017005403.1 XM_017005404.1 NR_033997.1

Gene Symbol
RNF144A-AS1
Entrez Gene ID
386597
Gene Name
RNF144A antisense RNA 1
Gene Aliases
-
UniGene
Hs.559010
Gene Chromosome Location
Chr.2: 6912276-6918682 on Build GRCh38
Transcripts
NM_014746.4 XM_017005398.1 XM_017005399.1 XM_017005400.1 XM_017005401.1 XM_017005402.1 XM_017005403.1 XM_017005404.1 NR_033997.1

SNP Detail

SNP ID Chromosome Location
rs996407783 Chr.2: 6917601-6917601 on Build GRCh38
rs968126432 Chr.2: 6917311-6917311 on Build GRCh38
rs955864251 Chr.2: 6917479-6917479 on Build GRCh38
rs1027786058 Chr.2: 6917253-6917253 on Build GRCh38
rs903841270 Chr.2: 6917620-6917620 on Build GRCh38
rs907665684 Chr.2: 6917568-6917568 on Build GRCh38
rs1028966547 Chr.2: 6917459-6917459 on Build GRCh38
rs556046747 Chr.2: 6917333-6917333 on Build GRCh38
rs917083162 Chr.2: 6917518-6917518 on Build GRCh38
rs930327645 Chr.2: 6917547-6917547 on Build GRCh38
rs988642390 Chr.2: 6917516-6917516 on Build GRCh38
rs886589620 Chr.2: 6917442-6917442 on Build GRCh38
rs988675015 Chr.2: 6917346-6917346 on Build GRCh38
rs112524533 Chr.2: 6917532-6917532 on Build GRCh38
rs934518986 Chr.2: 6917339-6917339 on Build GRCh38
rs931071271 Chr.2: 6917455-6917467 on Build GRCh38
rs886616825 Chr.2: 6917286-6917286 on Build GRCh38
rs554702295 Chr.2: 6917197-6917197 on Build GRCh38
rs866777619 Chr.2: 6917467-6917467 on Build GRCh38
rs902059697 Chr.2: 6917453-6917453 on Build GRCh38
rs930378901 Chr.2: 6917409-6917409 on Build GRCh38
rs762573450 Chr.2: 6917289-6917289 on Build GRCh38
rs74619211 Chr.2: 6917254-6917254 on Build GRCh38
rs926953775 Chr.2: 6917541-6917541 on Build GRCh38
rs933323417 Chr.2: 6917245-6917245 on Build GRCh38
rs971252956 Chr.2: 6917524-6917524 on Build GRCh38
rs956749113 Chr.2: 6917641-6917641 on Build GRCh38
rs902025578 Chr.2: 6917597-6917597 on Build GRCh38
rs917197603 Chr.2: 6917352-6917352 on Build GRCh38
rs968471144 Chr.2: 6917462-6917462 on Build GRCh38
rs979938578 Chr.2: 6917321-6917321 on Build GRCh38
rs1031080614 Chr.2: 6917633-6917633 on Build GRCh38
rs1005112301 Chr.2: 6917287-6917287 on Build GRCh38
rs1044027036 Chr.2: 6917387-6917387 on Build GRCh38
rs940427175 Chr.2: 6917570-6917570 on Build GRCh38
rs1044117281 Chr.2: 6917232-6917232 on Build GRCh38
rs1001307245 Chr.2: 6917626-6917626 on Build GRCh38
rs915546058 Chr.2: 6917309-6917310 on Build GRCh38
rs1040533569 Chr.2: 6917578-6917578 on Build GRCh38
rs926957663 Chr.2: 6917390-6917390 on Build GRCh38
rs565080701 Chr.2: 6917566-6917566 on Build GRCh38
rs1051142934 Chr.2: 6917611-6917611 on Build GRCh38
rs949718558 Chr.2: 6917364-6917364 on Build GRCh38
rs527428692 Chr.2: 6917612-6917612 on Build GRCh38
rs1049302703 Chr.2: 6917435-6917435 on Build GRCh38
rs191078726 Chr.2: 6917451-6917451 on Build GRCh38
rs1013631658 Chr.2: 6917642-6917642 on Build GRCh38
rs35378104 Chr.2: 6917307-6917307 on Build GRCh38
rs1031533366 Chr.2: 6917474-6917474 on Build GRCh38
rs949749698 Chr.2: 6917201-6917201 on Build GRCh38
rs527528919 Chr.2: 6917517-6917532 on Build GRCh38
rs1032708195 Chr.2: 6917379-6917379 on Build GRCh38
rs925786707 Chr.2: 6917325-6917325 on Build GRCh38
rs1048798290 Chr.2: 6917558-6917558 on Build GRCh38
rs994997637 Chr.2: 6917247-6917247 on Build GRCh38
rs1005089526 Chr.2: 6917449-6917449 on Build GRCh38
rs187210495 Chr.2: 6917200-6917200 on Build GRCh38
rs543376747 Chr.2: 6917310-6917310 on Build GRCh38
rs535111113 Chr.2: 6917190-6917190 on Build GRCh38
rs923299475 Chr.2: 6917222-6917222 on Build GRCh38
rs966027914 Chr.2: 6917305-6917305 on Build GRCh38

View Full Product Details