Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74845064-74845561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAMP2
Entrez Gene ID
10066
Gene Name
secretory carrier membrane protein 2
Gene Aliases
-
UniGene
Hs.458917
Gene Chromosome Location
Chr.15: 74843730-74873379 on Build GRCh38
Transcripts
NM_001320778.1 NM_005697.4 XM_017021856.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7449225 c.778G>A p.A260T Chr.15: 74845550-74845550 on Build GRCh38
6772406 c.874C>T p.R292W Chr.15: 74845199-74845199 on Build GRCh38
3370260 c.832C>T p.L278F Chr.15: 74845496-74845496 on Build GRCh38
5322802 c.785C>G p.S262* Chr.15: 74845543-74845543 on Build GRCh38
4056935 c.829G>A p.V277M Chr.15: 74845499-74845499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754112307 Chr.15: 74845130-74845130 on Build GRCh38
rs747565961 Chr.15: 74845427-74845427 on Build GRCh38
rs765268439 Chr.15: 74845380-74845380 on Build GRCh38
rs773066760 Chr.15: 74845229-74845229 on Build GRCh38
rs754577900 Chr.15: 74845423-74845423 on Build GRCh38
rs751957239 Chr.15: 74845405-74845405 on Build GRCh38
rs775737312 Chr.15: 74845384-74845384 on Build GRCh38
rs750963093 Chr.15: 74845413-74845413 on Build GRCh38
rs959312458 Chr.15: 74845257-74845257 on Build GRCh38
rs781737537 Chr.15: 74845326-74845326 on Build GRCh38
rs374810081 Chr.15: 74845425-74845425 on Build GRCh38
rs745361131 Chr.15: 74845298-74845298 on Build GRCh38
rs770602744 Chr.15: 74845236-74845236 on Build GRCh38
rs373991079 Chr.15: 74845087-74845087 on Build GRCh38
rs771815418 Chr.15: 74845340-74845340 on Build GRCh38
rs919337601 Chr.15: 74845300-74845300 on Build GRCh38
rs10220747 Chr.15: 74845527-74845527 on Build GRCh38
rs763258211 Chr.15: 74845385-74845385 on Build GRCh38
rs759676763 Chr.15: 74845379-74845379 on Build GRCh38
rs148434437 Chr.15: 74845102-74845102 on Build GRCh38
rs758104845 Chr.15: 74845432-74845432 on Build GRCh38
rs985230716 Chr.15: 74845312-74845315 on Build GRCh38
rs774424649 Chr.15: 74845494-74845494 on Build GRCh38
rs367749039 Chr.15: 74845263-74845263 on Build GRCh38
rs750363423 Chr.15: 74845106-74845106 on Build GRCh38
rs768429330 Chr.15: 74845214-74845214 on Build GRCh38
rs762307284 Chr.15: 74845498-74845498 on Build GRCh38
rs113637464 Chr.15: 74845507-74845507 on Build GRCh38
rs759550153 Chr.15: 74845242-74845242 on Build GRCh38
rs755855763 Chr.15: 74845128-74845128 on Build GRCh38
rs528663367 Chr.15: 74845448-74845448 on Build GRCh38
rs778346051 Chr.15: 74845286-74845286 on Build GRCh38
rs779638713 Chr.15: 74845210-74845210 on Build GRCh38
rs1024913591 Chr.15: 74845512-74845512 on Build GRCh38
rs770910702 Chr.15: 74845369-74845369 on Build GRCh38
rs769235979 Chr.15: 74845202-74845202 on Build GRCh38
rs776587369 Chr.15: 74845458-74845458 on Build GRCh38
rs1013478728 Chr.15: 74845496-74845496 on Build GRCh38
rs754524723 Chr.15: 74845331-74845331 on Build GRCh38
rs773085891 Chr.15: 74845352-74845352 on Build GRCh38
rs768937069 Chr.15: 74845488-74845488 on Build GRCh38
rs746876056 Chr.15: 74845442-74845442 on Build GRCh38
rs114328139 Chr.15: 74845282-74845282 on Build GRCh38
rs551116869 Chr.15: 74845406-74845406 on Build GRCh38
rs757814505 Chr.15: 74845321-74845321 on Build GRCh38
rs1033367442 Chr.15: 74845429-74845429 on Build GRCh38
rs755219920 Chr.15: 74845149-74845149 on Build GRCh38
rs562411151 Chr.15: 74845233-74845233 on Build GRCh38
rs771612545 Chr.15: 74845393-74845393 on Build GRCh38
rs751019735 Chr.15: 74845508-74845508 on Build GRCh38
rs778777033 Chr.15: 74845335-74845335 on Build GRCh38
rs987856573 Chr.15: 74845287-74845287 on Build GRCh38
rs761759236 Chr.15: 74845222-74845222 on Build GRCh38
rs199748703 Chr.15: 74845464-74845464 on Build GRCh38
rs1033570722 Chr.15: 74845237-74845237 on Build GRCh38
rs767371103 Chr.15: 74845104-74845104 on Build GRCh38
rs561124420 Chr.15: 74845089-74845089 on Build GRCh38
rs747518031 Chr.15: 74845199-74845199 on Build GRCh38
rs747774503 Chr.15: 74845337-74845337 on Build GRCh38
rs766455179 Chr.15: 74845127-74845127 on Build GRCh38
rs767965502 Chr.15: 74845500-74845500 on Build GRCh38
rs371228428 Chr.15: 74845264-74845264 on Build GRCh38
rs376862263 Chr.15: 74845110-74845110 on Build GRCh38
rs751820605 Chr.15: 74845267-74845267 on Build GRCh38
rs771899563 Chr.15: 74845227-74845227 on Build GRCh38
rs548821463 Chr.15: 74845474-74845474 on Build GRCh38
rs764181886 Chr.15: 74845392-74845392 on Build GRCh38
rs746711560 Chr.15: 74845367-74845367 on Build GRCh38
rs779036778 Chr.15: 74845152-74845152 on Build GRCh38
rs768374541 Chr.15: 74845099-74845099 on Build GRCh38
rs760642146 Chr.15: 74845230-74845230 on Build GRCh38
rs777002891 Chr.15: 74845441-74845441 on Build GRCh38
rs748821730 Chr.15: 74845327-74845327 on Build GRCh38
rs776533978 Chr.15: 74845370-74845370 on Build GRCh38
rs370339997 Chr.15: 74845424-74845424 on Build GRCh38
rs753070982 Chr.15: 74845179-74845179 on Build GRCh38
rs550586142 Chr.15: 74845232-74845232 on Build GRCh38
rs765497955 Chr.15: 74845249-74845249 on Build GRCh38
rs748833322 Chr.15: 74845215-74845215 on Build GRCh38
rs762444069 Chr.15: 74845475-74845475 on Build GRCh38
rs113655984 Chr.15: 74845219-74845219 on Build GRCh38
rs758746473 Chr.15: 74845186-74845186 on Build GRCh38
rs930625988 Chr.15: 74845304-74845304 on Build GRCh38
rs752909419 Chr.15: 74845252-74845252 on Build GRCh38
rs762874433 Chr.15: 74845482-74845482 on Build GRCh38
rs142904321 Chr.15: 74845198-74845198 on Build GRCh38
rs748768188 Chr.15: 74845213-74845213 on Build GRCh38

View Full Product Details