Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74222838-74223340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC4A5
Entrez Gene ID
57835
Gene Name
solute carrier family 4 member 5
Gene Aliases
NBC4, NBCe2
UniGene
Hs.594099
Gene Chromosome Location
Chr.2: 74216242-74343407 on Build GRCh38
Transcripts
NM_021196.3 NM_133478.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6782856 c.3315T>C p.G1105G Chr.2: 74222884-74222884 on Build GRCh38
5451676 c.3267G>A p.S1089S Chr.2: 74222932-74222932 on Build GRCh38
5451675 c.3315G>A p.S1105S Chr.2: 74222932-74222932 on Build GRCh38
5397400 c.3307C>T p.Q1103* Chr.2: 74222892-74222892 on Build GRCh38
4986337 c.3316G>A p.V1106I Chr.2: 74222931-74222931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757085721 Chr.2: 74222915-74222915 on Build GRCh38
rs577260178 Chr.2: 74223194-74223194 on Build GRCh38
rs1001223816 Chr.2: 74223246-74223246 on Build GRCh38
rs988894280 Chr.2: 74223050-74223050 on Build GRCh38
rs937619536 Chr.2: 74223096-74223097 on Build GRCh38
rs1038541680 Chr.2: 74223114-74223114 on Build GRCh38
rs375755484 Chr.2: 74223271-74223271 on Build GRCh38
rs774641469 Chr.2: 74222874-74222874 on Build GRCh38
rs930728721 Chr.2: 74223151-74223151 on Build GRCh38
rs890890014 Chr.2: 74223236-74223236 on Build GRCh38
rs913787091 Chr.2: 74223007-74223007 on Build GRCh38
rs545559914 Chr.2: 74223152-74223152 on Build GRCh38
rs1028080625 Chr.2: 74222892-74222892 on Build GRCh38
rs138995029 Chr.2: 74222887-74222887 on Build GRCh38
rs546940709 Chr.2: 74223035-74223035 on Build GRCh38
rs574685882 Chr.2: 74223073-74223073 on Build GRCh38
rs994343126 Chr.2: 74223174-74223174 on Build GRCh38
rs1036505623 Chr.2: 74223149-74223149 on Build GRCh38
rs1049102285 Chr.2: 74223217-74223217 on Build GRCh38
rs149598291 Chr.2: 74222932-74222932 on Build GRCh38
rs530627975 Chr.2: 74223150-74223150 on Build GRCh38
rs764932863 Chr.2: 74223295-74223295 on Build GRCh38
rs991741300 Chr.2: 74223074-74223074 on Build GRCh38
rs766378680 Chr.2: 74222975-74222975 on Build GRCh38
rs901745589 Chr.2: 74223294-74223294 on Build GRCh38
rs777446708 Chr.2: 74223003-74223004 on Build GRCh38
rs968222173 Chr.2: 74222994-74222994 on Build GRCh38
rs767688760 Chr.2: 74222882-74222882 on Build GRCh38
rs539102054 Chr.2: 74223238-74223238 on Build GRCh38
rs765291329 Chr.2: 74222948-74222949 on Build GRCh38
rs6745054 Chr.2: 74222998-74222998 on Build GRCh38
rs56208981 Chr.2: 74223299-74223299 on Build GRCh38
rs759736499 Chr.2: 74223061-74223061 on Build GRCh38
rs762209450 Chr.2: 74222942-74222942 on Build GRCh38
rs762827480 Chr.2: 74222979-74222979 on Build GRCh38
rs773525340 Chr.2: 74222914-74222914 on Build GRCh38
rs539991705 Chr.2: 74222916-74222916 on Build GRCh38
rs766472355 Chr.2: 74222895-74222895 on Build GRCh38
rs746758842 Chr.2: 74222910-74222910 on Build GRCh38
rs767257391 Chr.2: 74222965-74222966 on Build GRCh38
rs979268515 Chr.2: 74223000-74223000 on Build GRCh38
rs557328300 Chr.2: 74223144-74223144 on Build GRCh38
rs937653432 Chr.2: 74223067-74223067 on Build GRCh38
rs536317044 Chr.2: 74223298-74223298 on Build GRCh38
rs752577356 Chr.2: 74222896-74222896 on Build GRCh38
rs957442634 Chr.2: 74223037-74223037 on Build GRCh38
rs762047487 Chr.2: 74222876-74222876 on Build GRCh38
rs202213981 Chr.2: 74223083-74223083 on Build GRCh38
rs201396252 Chr.2: 74222977-74222977 on Build GRCh38
rs767786134 Chr.2: 74223082-74223082 on Build GRCh38
rs752508779 Chr.2: 74222974-74222975 on Build GRCh38
rs79685968 Chr.2: 74223021-74223021 on Build GRCh38
rs751455476 Chr.2: 74222950-74222954 on Build GRCh38
rs751519868 Chr.2: 74222993-74222993 on Build GRCh38
rs781129527 Chr.2: 74222999-74222999 on Build GRCh38
rs36081793 Chr.2: 74222931-74222931 on Build GRCh38
rs774999444 Chr.2: 74222933-74222933 on Build GRCh38
rs926291658 Chr.2: 74223036-74223036 on Build GRCh38
rs542106980 Chr.2: 74223080-74223080 on Build GRCh38
rs750615240 Chr.2: 74222886-74222886 on Build GRCh38
rs769159779 Chr.2: 74222937-74222937 on Build GRCh38
rs763512633 Chr.2: 74222864-74222864 on Build GRCh38
rs1040345518 Chr.2: 74223293-74223293 on Build GRCh38
rs913295844 Chr.2: 74223297-74223297 on Build GRCh38
rs758294798 Chr.2: 74222898-74222898 on Build GRCh38
rs755949249 Chr.2: 74223003-74223006 on Build GRCh38
rs963979270 Chr.2: 74223245-74223245 on Build GRCh38
rs777729420 Chr.2: 74222902-74222902 on Build GRCh38
rs925146077 Chr.2: 74223028-74223029 on Build GRCh38
rs889153740 Chr.2: 74223256-74223256 on Build GRCh38
rs563860602 Chr.2: 74223266-74223266 on Build GRCh38
rs377272391 Chr.2: 74222970-74222970 on Build GRCh38
rs764166148 Chr.2: 74222983-74222983 on Build GRCh38
rs759472286 Chr.2: 74222960-74222966 on Build GRCh38
rs1018379391 Chr.2: 74223243-74223244 on Build GRCh38
rs764371682 Chr.2: 74222870-74222870 on Build GRCh38
rs767850457 Chr.2: 74222962-74222962 on Build GRCh38
rs181545717 Chr.2: 74223093-74223093 on Build GRCh38

View Full Product Details