Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64062895-64063409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRGAP1
Entrez Gene ID
57522
Gene Name
SLIT-ROBO Rho GTPase activating protein 1
Gene Aliases
ARHGAP13, NMTC2
UniGene
Hs.210751
Gene Chromosome Location
Chr.12: 63844372-64147863 on Build GRCh38
Transcripts
NM_020762.2 XM_005269042.3 XM_011538580.1 XM_011538581.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6042678 c.1023+7A>G p.? Chr.12: 64063145-64063145 on Build GRCh38
6790194 c.969T>A p.P323P Chr.12: 64063084-64063084 on Build GRCh38
4044122 c.975G>A p.A325A Chr.12: 64063090-64063090 on Build GRCh38
1363573 c.974C>T p.A325V Chr.12: 64063089-64063089 on Build GRCh38
942444 c.975G>A p.A325A Chr.12: 64063090-64063090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374079925 Chr.12: 64063114-64063114 on Build GRCh38
rs545431477 Chr.12: 64063185-64063185 on Build GRCh38
rs745493927 Chr.12: 64063339-64063339 on Build GRCh38
rs968239995 Chr.12: 64063355-64063355 on Build GRCh38
rs946340027 Chr.12: 64063256-64063256 on Build GRCh38
rs1005078673 Chr.12: 64063375-64063375 on Build GRCh38
rs763987787 Chr.12: 64063002-64063002 on Build GRCh38
rs776580465 Chr.12: 64063110-64063110 on Build GRCh38
rs780449158 Chr.12: 64063051-64063051 on Build GRCh38
rs978884308 Chr.12: 64063361-64063361 on Build GRCh38
rs746018881 Chr.12: 64063291-64063291 on Build GRCh38
rs367757661 Chr.12: 64063003-64063003 on Build GRCh38
rs998072040 Chr.12: 64063286-64063286 on Build GRCh38
rs564794053 Chr.12: 64063366-64063366 on Build GRCh38
rs554298746 Chr.12: 64062988-64062988 on Build GRCh38
rs200530268 Chr.12: 64062994-64062994 on Build GRCh38
rs145973371 Chr.12: 64063082-64063082 on Build GRCh38
rs773806400 Chr.12: 64063089-64063089 on Build GRCh38
rs797044990 Chr.12: 64062938-64062938 on Build GRCh38
rs371278918 Chr.12: 64063043-64063043 on Build GRCh38
rs542720477 Chr.12: 64063019-64063019 on Build GRCh38
rs1036544617 Chr.12: 64063385-64063385 on Build GRCh38
rs762637674 Chr.12: 64062970-64062970 on Build GRCh38
rs558573607 Chr.12: 64062934-64062934 on Build GRCh38
rs750534559 Chr.12: 64063040-64063040 on Build GRCh38
rs142448927 Chr.12: 64063076-64063076 on Build GRCh38
rs11829649 Chr.12: 64063167-64063167 on Build GRCh38
rs750481462 Chr.12: 64063126-64063126 on Build GRCh38
rs769440054 Chr.12: 64062966-64062966 on Build GRCh38
rs565428638 Chr.12: 64063212-64063212 on Build GRCh38
rs771658622 Chr.12: 64063098-64063098 on Build GRCh38
rs747601728 Chr.12: 64063054-64063054 on Build GRCh38
rs1013359299 Chr.12: 64063008-64063008 on Build GRCh38
rs754414509 Chr.12: 64062959-64062959 on Build GRCh38
rs114508252 Chr.12: 64062933-64062933 on Build GRCh38
rs947565208 Chr.12: 64062954-64062954 on Build GRCh38
rs760361145 Chr.12: 64062998-64062998 on Build GRCh38
rs765587555 Chr.12: 64063124-64063124 on Build GRCh38
rs767469843 Chr.12: 64062986-64062986 on Build GRCh38
rs11829640 Chr.12: 64063285-64063285 on Build GRCh38
rs759415074 Chr.12: 64062980-64062980 on Build GRCh38
rs139894551 Chr.12: 64063103-64063103 on Build GRCh38
rs747163022 Chr.12: 64063346-64063346 on Build GRCh38
rs137909926 Chr.12: 64062976-64062976 on Build GRCh38
rs768873949 Chr.12: 64063055-64063055 on Build GRCh38
rs888564636 Chr.12: 64063120-64063120 on Build GRCh38
rs772394387 Chr.12: 64063307-64063307 on Build GRCh38
rs758531451 Chr.12: 64063164-64063164 on Build GRCh38
rs373824078 Chr.12: 64063304-64063304 on Build GRCh38
rs781357821 Chr.12: 64063186-64063186 on Build GRCh38
rs770426933 Chr.12: 64062975-64062975 on Build GRCh38
rs556008574 Chr.12: 64063180-64063180 on Build GRCh38
rs775257660 Chr.12: 64063100-64063100 on Build GRCh38
rs944215842 Chr.12: 64063158-64063158 on Build GRCh38
rs563035863 Chr.12: 64063079-64063079 on Build GRCh38
rs77397108 Chr.12: 64062935-64062935 on Build GRCh38
rs757609288 Chr.12: 64063006-64063006 on Build GRCh38
rs754137240 Chr.12: 64062929-64062929 on Build GRCh38
rs772719836 Chr.12: 64062969-64062969 on Build GRCh38
rs374501455 Chr.12: 64062964-64062964 on Build GRCh38
rs763782085 Chr.12: 64063108-64063108 on Build GRCh38
rs780964806 Chr.12: 64063244-64063244 on Build GRCh38
rs1045216574 Chr.12: 64063260-64063260 on Build GRCh38
rs752007273 Chr.12: 64063174-64063174 on Build GRCh38
rs772720206 Chr.12: 64062937-64062937 on Build GRCh38
rs770588443 Chr.12: 64063086-64063086 on Build GRCh38
rs7135485 Chr.12: 64063308-64063308 on Build GRCh38
rs745401734 Chr.12: 64063090-64063090 on Build GRCh38
rs376728642 Chr.12: 64063203-64063203 on Build GRCh38

View Full Product Details