Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100738433-100738846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAN
Entrez Gene ID
7455
Gene Name
zonadhesin (gene/pseudogene)
Gene Aliases
-
UniGene
Hs.307004
Gene Chromosome Location
Chr.7: 100733599-100797797 on Build GRCh38
Transcripts
NM_003386.2 NM_173059.2 NR_111917.1 NR_111918.1 NR_111919.1 XM_011516555.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3411363 c.760C>T p.P254S Chr.7: 100738607-100738607 on Build GRCh38
3831307 c.742G>C p.D248H Chr.7: 100738589-100738589 on Build GRCh38
1082968 c.696G>A p.K232K Chr.7: 100738543-100738543 on Build GRCh38
5029625 c.614-1G>C p.? Chr.7: 100738460-100738460 on Build GRCh38
5672926 c.658T>A p.C220S Chr.7: 100738505-100738505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750283375 Chr.7: 100738526-100738526 on Build GRCh38
rs1030184198 Chr.7: 100738677-100738677 on Build GRCh38
rs780809483 Chr.7: 100738579-100738579 on Build GRCh38
rs747747456 Chr.7: 100738580-100738580 on Build GRCh38
rs962930901 Chr.7: 100738758-100738758 on Build GRCh38
rs889895942 Chr.7: 100738688-100738688 on Build GRCh38
rs764912674 Chr.7: 100738516-100738516 on Build GRCh38
rs555668643 Chr.7: 100738547-100738547 on Build GRCh38
rs927008800 Chr.7: 100738818-100738818 on Build GRCh38
rs780495267 Chr.7: 100738600-100738600 on Build GRCh38
rs772644738 Chr.7: 100738616-100738616 on Build GRCh38
rs560141733 Chr.7: 100738736-100738736 on Build GRCh38
rs73711157 Chr.7: 100738667-100738667 on Build GRCh38
rs755640042 Chr.7: 100738586-100738586 on Build GRCh38
rs971061065 Chr.7: 100738782-100738782 on Build GRCh38
rs927130320 Chr.7: 100738487-100738487 on Build GRCh38
rs981152039 Chr.7: 100738799-100738799 on Build GRCh38
rs746268486 Chr.7: 100738607-100738607 on Build GRCh38
rs372927060 Chr.7: 100738581-100738581 on Build GRCh38
rs776157900 Chr.7: 100738617-100738617 on Build GRCh38
rs370119698 Chr.7: 100738501-100738501 on Build GRCh38
rs994413971 Chr.7: 100738779-100738779 on Build GRCh38
rs775673718 Chr.7: 100738485-100738485 on Build GRCh38
rs1026365229 Chr.7: 100738546-100738546 on Build GRCh38
rs749912066 Chr.7: 100738481-100738481 on Build GRCh38
rs937149549 Chr.7: 100738498-100738498 on Build GRCh38
rs779996117 Chr.7: 100738534-100738534 on Build GRCh38
rs370624145 Chr.7: 100738474-100738474 on Build GRCh38
rs201633210 Chr.7: 100738528-100738528 on Build GRCh38
rs761921744 Chr.7: 100738638-100738638 on Build GRCh38
rs1042409580 Chr.7: 100738611-100738611 on Build GRCh38
rs779121452 Chr.7: 100738594-100738594 on Build GRCh38
rs1025252145 Chr.7: 100738780-100738780 on Build GRCh38
rs1046322223 Chr.7: 100738464-100738464 on Build GRCh38
rs572783687 Chr.7: 100738747-100738747 on Build GRCh38
rs183919117 Chr.7: 100738493-100738493 on Build GRCh38
rs267601185 Chr.7: 100738569-100738569 on Build GRCh38
rs556576698 Chr.7: 100738685-100738685 on Build GRCh38
rs747576918 Chr.7: 100738622-100738622 on Build GRCh38
rs776813615 Chr.7: 100738627-100738627 on Build GRCh38
rs377078924 Chr.7: 100738623-100738623 on Build GRCh38
rs751004444 Chr.7: 100738535-100738535 on Build GRCh38
rs760846665 Chr.7: 100738490-100738490 on Build GRCh38
rs964755878 Chr.7: 100738715-100738715 on Build GRCh38
rs754370642 Chr.7: 100738634-100738635 on Build GRCh38
rs147660500 Chr.7: 100738714-100738714 on Build GRCh38
rs373927197 Chr.7: 100738712-100738712 on Build GRCh38
rs760086332 Chr.7: 100738470-100738470 on Build GRCh38

View Full Product Details