Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9755486-9755984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC440028
Entrez Gene ID
440028
Gene Name
uncharacterized LOC440028
Gene Aliases
-
UniGene
Hs.677541
Gene Chromosome Location
Chr.11: 9754770-9759533 on Build GRCh38
Transcripts
NR_033972.1

SNP Detail

SNP ID Chromosome Location
rs979305597 Chr.11: 9755588-9755588 on Build GRCh38
rs950085496 Chr.11: 9755948-9755948 on Build GRCh38
rs1033169851 Chr.11: 9755935-9755935 on Build GRCh38
rs1046311394 Chr.11: 9755682-9755682 on Build GRCh38
rs537925861 Chr.11: 9755578-9755578 on Build GRCh38
rs557660305 Chr.11: 9755743-9755743 on Build GRCh38
rs927315269 Chr.11: 9755958-9755958 on Build GRCh38
rs888265712 Chr.11: 9755786-9755786 on Build GRCh38
rs950029031 Chr.11: 9755674-9755674 on Build GRCh38
rs932245680 Chr.11: 9755757-9755757 on Build GRCh38
rs574703101 Chr.11: 9755718-9755718 on Build GRCh38
rs926120153 Chr.11: 9755603-9755603 on Build GRCh38
rs982618667 Chr.11: 9755710-9755710 on Build GRCh38
rs1028582313 Chr.11: 9755908-9755908 on Build GRCh38
rs7928407 Chr.11: 9755580-9755580 on Build GRCh38
rs759601896 Chr.11: 9755935-9755937 on Build GRCh38
rs7939397 Chr.11: 9755874-9755874 on Build GRCh38
rs896802689 Chr.11: 9755791-9755791 on Build GRCh38
rs1003995275 Chr.11: 9755816-9755816 on Build GRCh38
rs915243042 Chr.11: 9755960-9755960 on Build GRCh38
rs530745908 Chr.11: 9755950-9755950 on Build GRCh38
rs917260364 Chr.11: 9755673-9755673 on Build GRCh38
rs1050517406 Chr.11: 9755759-9755764 on Build GRCh38
rs990685136 Chr.11: 9755686-9755686 on Build GRCh38
rs916425362 Chr.11: 9755695-9755695 on Build GRCh38
rs988841776 Chr.11: 9755659-9755659 on Build GRCh38
rs937933093 Chr.11: 9755736-9755736 on Build GRCh38
rs763679947 Chr.11: 9755922-9755922 on Build GRCh38
rs186025640 Chr.11: 9755742-9755742 on Build GRCh38
rs780048223 Chr.11: 9755797-9755797 on Build GRCh38
rs796720246 Chr.11: 9755759-9755759 on Build GRCh38
rs905072971 Chr.11: 9755721-9755721 on Build GRCh38
rs746432226 Chr.11: 9755597-9755597 on Build GRCh38
rs561822870 Chr.11: 9755942-9755942 on Build GRCh38
rs998222835 Chr.11: 9755896-9755896 on Build GRCh38
rs970762414 Chr.11: 9755701-9755701 on Build GRCh38
rs771401028 Chr.11: 9755510-9755510 on Build GRCh38
rs1007947234 Chr.11: 9755782-9755782 on Build GRCh38
rs979018122 Chr.11: 9755920-9755920 on Build GRCh38
rs35350639 Chr.11: 9755588-9755589 on Build GRCh38
rs144270844 Chr.11: 9755746-9755746 on Build GRCh38
rs917291340 Chr.11: 9755944-9755944 on Build GRCh38
rs935354406 Chr.11: 9755725-9755725 on Build GRCh38
rs1007936567 Chr.11: 9755585-9755585 on Build GRCh38
rs956177876 Chr.11: 9755936-9755936 on Build GRCh38
rs754009410 Chr.11: 9755534-9755535 on Build GRCh38
rs554697197 Chr.11: 9755669-9755669 on Build GRCh38
rs1026897146 Chr.11: 9755833-9755833 on Build GRCh38
rs545771272 Chr.11: 9755747-9755747 on Build GRCh38
rs1012173314 Chr.11: 9755919-9755919 on Build GRCh38
rs954476115 Chr.11: 9755915-9755915 on Build GRCh38
rs559433922 Chr.11: 9755805-9755805 on Build GRCh38
rs977045769 Chr.11: 9755551-9755551 on Build GRCh38
rs117778087 Chr.11: 9755658-9755658 on Build GRCh38
rs1054038968 Chr.11: 9755737-9755737 on Build GRCh38
rs534016123 Chr.11: 9755818-9755818 on Build GRCh38

View Full Product Details