Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 24583486-24583989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00477
Entrez Gene ID
144360
Gene Name
long intergenic non-protein coding RNA 477
Gene Aliases
C12orf67, FAM191B
UniGene
Hs.350668
Gene Chromosome Location
Chr.12: 24566964-24584168 on Build GRCh38
Transcripts
NR_029451.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6072213 c.483C>A p.P161P Chr.12: 24583637-24583637 on Build GRCh38
3459446 c.401C>T p.S134F Chr.12: 24583719-24583719 on Build GRCh38
6136532 c.287G>T p.G96V Chr.12: 24583833-24583833 on Build GRCh38
3871304 c.243C>T p.F81F Chr.12: 24583877-24583877 on Build GRCh38
6701402 c.363C>T p.S121S Chr.12: 24583757-24583757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374782178 Chr.12: 24583834-24583835 on Build GRCh38
rs372628957 Chr.12: 24583780-24583780 on Build GRCh38
rs753354985 Chr.12: 24583599-24583599 on Build GRCh38
rs779438257 Chr.12: 24583776-24583776 on Build GRCh38
rs759685106 Chr.12: 24583901-24583901 on Build GRCh38
rs757405655 Chr.12: 24583659-24583659 on Build GRCh38
rs758820947 Chr.12: 24583831-24583831 on Build GRCh38
rs767708944 Chr.12: 24583752-24583752 on Build GRCh38
rs369289029 Chr.12: 24583906-24583906 on Build GRCh38
rs765152672 Chr.12: 24583717-24583717 on Build GRCh38
rs759949584 Chr.12: 24583822-24583822 on Build GRCh38
rs369958757 Chr.12: 24583725-24583725 on Build GRCh38
rs748590273 Chr.12: 24583828-24583828 on Build GRCh38
rs776126085 Chr.12: 24583602-24583602 on Build GRCh38
rs762636138 Chr.12: 24583801-24583801 on Build GRCh38
rs558009001 Chr.12: 24583948-24583948 on Build GRCh38
rs528057305 Chr.12: 24583850-24583850 on Build GRCh38
rs753987744 Chr.12: 24583896-24583896 on Build GRCh38
rs779545495 Chr.12: 24583923-24583923 on Build GRCh38
rs369677772 Chr.12: 24583815-24583815 on Build GRCh38
rs770655292 Chr.12: 24583715-24583715 on Build GRCh38
rs748113118 Chr.12: 24583932-24583932 on Build GRCh38
rs779308874 Chr.12: 24583576-24583576 on Build GRCh38
rs750162840 Chr.12: 24583579-24583579 on Build GRCh38
rs377243854 Chr.12: 24583860-24583860 on Build GRCh38
rs373100495 Chr.12: 24583821-24583821 on Build GRCh38
rs778029312 Chr.12: 24583649-24583649 on Build GRCh38
rs759772313 Chr.12: 24583955-24583955 on Build GRCh38
rs150843007 Chr.12: 24583793-24583793 on Build GRCh38
rs766485149 Chr.12: 24583892-24583892 on Build GRCh38
rs185434134 Chr.12: 24583841-24583841 on Build GRCh38
rs1043931639 Chr.12: 24583876-24583876 on Build GRCh38
rs780918840 Chr.12: 24583725-24583725 on Build GRCh38
rs748867470 Chr.12: 24583924-24583924 on Build GRCh38
rs756131800 Chr.12: 24583731-24583731 on Build GRCh38
rs969829323 Chr.12: 24583774-24583774 on Build GRCh38
rs754697365 Chr.12: 24583701-24583701 on Build GRCh38
rs745425076 Chr.12: 24583791-24583791 on Build GRCh38
rs751822551 Chr.12: 24583912-24583912 on Build GRCh38
rs377120412 Chr.12: 24583783-24583783 on Build GRCh38
rs745915596 Chr.12: 24583660-24583660 on Build GRCh38
rs901853109 Chr.12: 24583652-24583652 on Build GRCh38
rs768625609 Chr.12: 24583861-24583861 on Build GRCh38
rs767360689 Chr.12: 24583875-24583875 on Build GRCh38
rs774178948 Chr.12: 24583947-24583948 on Build GRCh38
rs142357702 Chr.12: 24583735-24583735 on Build GRCh38
rs373094764 Chr.12: 24583929-24583929 on Build GRCh38
rs1007588130 Chr.12: 24583628-24583628 on Build GRCh38
rs750512441 Chr.12: 24583753-24583753 on Build GRCh38
rs753267764 Chr.12: 24583905-24583905 on Build GRCh38
rs760352552 Chr.12: 24583944-24583944 on Build GRCh38
rs546873041 Chr.12: 24583789-24583789 on Build GRCh38
rs752473330 Chr.12: 24583631-24583631 on Build GRCh38
rs770747280 Chr.12: 24583784-24583784 on Build GRCh38
rs747666873 Chr.12: 24583768-24583768 on Build GRCh38
rs760343896 Chr.12: 24583580-24583580 on Build GRCh38
rs755839231 Chr.12: 24583916-24583916 on Build GRCh38
rs780486140 Chr.12: 24583757-24583757 on Build GRCh38
rs776979120 Chr.12: 24583951-24583951 on Build GRCh38
rs762910246 Chr.12: 24583797-24583797 on Build GRCh38
rs769184878 Chr.12: 24583582-24583582 on Build GRCh38
rs745541313 Chr.12: 24583834-24583834 on Build GRCh38
rs535420855 Chr.12: 24583810-24583810 on Build GRCh38
rs771852353 Chr.12: 24583935-24583935 on Build GRCh38
rs763971052 Chr.12: 24583798-24583798 on Build GRCh38
rs139293562 Chr.12: 24583827-24583827 on Build GRCh38
rs757493507 Chr.12: 24583745-24583745 on Build GRCh38
rs772840554 Chr.12: 24583881-24583881 on Build GRCh38
rs749774717 Chr.12: 24583665-24583665 on Build GRCh38
rs765900854 Chr.12: 24583902-24583902 on Build GRCh38
rs761657408 Chr.12: 24583874-24583874 on Build GRCh38
rs143206776 Chr.12: 24583926-24583926 on Build GRCh38
rs774271695 Chr.12: 24583872-24583872 on Build GRCh38
rs774787771 Chr.12: 24583689-24583689 on Build GRCh38
rs773104308 Chr.12: 24583942-24583942 on Build GRCh38
rs769154444 Chr.12: 24583684-24583684 on Build GRCh38
rs753807964 Chr.12: 24583825-24583825 on Build GRCh38
rs775089371 Chr.12: 24583855-24583855 on Build GRCh38
rs756216426 Chr.12: 24583756-24583756 on Build GRCh38
rs755498928 Chr.12: 24583771-24583771 on Build GRCh38
rs550581343 Chr.12: 24583833-24583833 on Build GRCh38
rs758206884 Chr.12: 24583640-24583640 on Build GRCh38
rs1039100202 Chr.12: 24583940-24583940 on Build GRCh38
rs756220932 Chr.12: 24583820-24583820 on Build GRCh38
rs751795060 Chr.12: 24583650-24583650 on Build GRCh38
rs760971865 Chr.12: 24583891-24583891 on Build GRCh38
rs776340992 Chr.12: 24583785-24583785 on Build GRCh38
rs765198109 Chr.12: 24583629-24583629 on Build GRCh38
rs746412924 Chr.12: 24583628-24583628 on Build GRCh38
rs749684650 Chr.12: 24583679-24583679 on Build GRCh38
rs759414439 Chr.12: 24583611-24583611 on Build GRCh38
rs749803722 Chr.12: 24583767-24583767 on Build GRCh38
rs776330023 Chr.12: 24583716-24583716 on Build GRCh38
rs769379372 Chr.12: 24583800-24583801 on Build GRCh38
rs142121009 Chr.12: 24583674-24583674 on Build GRCh38
rs774797676 Chr.12: 24583794-24583794 on Build GRCh38
rs769293077 Chr.12: 24583846-24583846 on Build GRCh38
rs760431831 Chr.12: 24583714-24583714 on Build GRCh38
rs528317796 Chr.12: 24583739-24583739 on Build GRCh38
rs765988568 Chr.12: 24583587-24583587 on Build GRCh38
rs767796963 Chr.12: 24583813-24583813 on Build GRCh38

View Full Product Details